Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154214360-154214816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf43
Entrez Gene ID
25912
Gene Name
chromosome 1 open reading frame 43
Gene Aliases
HSPC012, NICE-3, NS5ATP4, S863-3
UniGene
Hs.287471
Gene Chromosome Location
Chr.1: 154206701-154220797 on Build GRCh38
Transcripts
NM_001098616.2 NM_001297717.1 NM_001297718.1 NM_001297720.1 NM_001297721.1 NM_001297723.1 NM_015449.3 NM_138740.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5754206 c.109G>C p.D37H Chr.1: 154214532-154214532 on Build GRCh38
6502971 c.281A>C p.N94T Chr.1: 154214462-154214462 on Build GRCh38
6502972 c.179A>C p.N60T Chr.1: 154214462-154214462 on Build GRCh38
458571 c.96C>G p.L32L Chr.1: 154214545-154214545 on Build GRCh38
1583463 c.268G>A p.E90K Chr.1: 154214475-154214475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999113234 Chr.1: 154214683-154214683 on Build GRCh38
rs751407213 Chr.1: 154214780-154214780 on Build GRCh38
rs776251536 Chr.1: 154214506-154214506 on Build GRCh38
rs143072730 Chr.1: 154214621-154214621 on Build GRCh38
rs1020874961 Chr.1: 154214421-154214421 on Build GRCh38
rs1003441116 Chr.1: 154214739-154214739 on Build GRCh38
rs777948186 Chr.1: 154214592-154214592 on Build GRCh38
rs1040714154 Chr.1: 154214647-154214647 on Build GRCh38
rs929491903 Chr.1: 154214534-154214534 on Build GRCh38
rs148001096 Chr.1: 154214552-154214552 on Build GRCh38
rs745820987 Chr.1: 154214597-154214597 on Build GRCh38
rs1041335023 Chr.1: 154214621-154214621 on Build GRCh38
rs1021242784 Chr.1: 154214719-154214719 on Build GRCh38
rs1031961290 Chr.1: 154214691-154214691 on Build GRCh38
rs377260078 Chr.1: 154214468-154214468 on Build GRCh38
rs200282453 Chr.1: 154214601-154214601 on Build GRCh38
rs908088379 Chr.1: 154214716-154214716 on Build GRCh38
rs201037348 Chr.1: 154214430-154214430 on Build GRCh38
rs911997997 Chr.1: 154214746-154214746 on Build GRCh38
rs1032566821 Chr.1: 154214382-154214382 on Build GRCh38
rs575069015 Chr.1: 154214631-154214631 on Build GRCh38
rs764127878 Chr.1: 154214480-154214480 on Build GRCh38
rs1009865272 Chr.1: 154214403-154214403 on Build GRCh38
rs768239212 Chr.1: 154214502-154214502 on Build GRCh38
rs1009402445 Chr.1: 154214718-154214718 on Build GRCh38
rs746880045 Chr.1: 154214615-154214615 on Build GRCh38
rs546677374 Chr.1: 154214779-154214779 on Build GRCh38
rs528528690 Chr.1: 154214735-154214735 on Build GRCh38
rs565408981 Chr.1: 154214782-154214782 on Build GRCh38
rs982322318 Chr.1: 154214712-154214712 on Build GRCh38
rs773848577 Chr.1: 154214584-154214585 on Build GRCh38
rs1023414009 Chr.1: 154214698-154214698 on Build GRCh38
rs542516464 Chr.1: 154214677-154214677 on Build GRCh38
rs781625435 Chr.1: 154214563-154214563 on Build GRCh38
rs750943198 Chr.1: 154214543-154214543 on Build GRCh38
rs748185707 Chr.1: 154214453-154214453 on Build GRCh38
rs777559873 Chr.1: 154214413-154214414 on Build GRCh38
rs769984201 Chr.1: 154214460-154214460 on Build GRCh38
rs758727765 Chr.1: 154214412-154214412 on Build GRCh38
rs781425976 Chr.1: 154214449-154214449 on Build GRCh38
rs143562217 Chr.1: 154214709-154214709 on Build GRCh38
rs12408458 Chr.1: 154214442-154214442 on Build GRCh38
rs752966984 Chr.1: 154214582-154214582 on Build GRCh38
rs563158193 Chr.1: 154214614-154214614 on Build GRCh38
rs749003623 Chr.1: 154214457-154214457 on Build GRCh38
rs770505358 Chr.1: 154214582-154214628 on Build GRCh38
rs920724890 Chr.1: 154214533-154214533 on Build GRCh38
rs932362049 Chr.1: 154214694-154214694 on Build GRCh38
rs756305524 Chr.1: 154214587-154214587 on Build GRCh38
rs150316876 Chr.1: 154214386-154214386 on Build GRCh38
rs577642353 Chr.1: 154214549-154214549 on Build GRCh38
rs899518259 Chr.1: 154214651-154214651 on Build GRCh38
rs370526509 Chr.1: 154214429-154214429 on Build GRCh38
rs760244619 Chr.1: 154214498-154214498 on Build GRCh38
rs932053813 Chr.1: 154214585-154214585 on Build GRCh38
rs764694541 Chr.1: 154214522-154214522 on Build GRCh38
rs1053549143 Chr.1: 154214695-154214695 on Build GRCh38
rs766467531 Chr.1: 154214612-154214612 on Build GRCh38
rs775312577 Chr.1: 154214484-154214484 on Build GRCh38
rs766685758 Chr.1: 154214550-154214550 on Build GRCh38
rs971276580 Chr.1: 154214711-154214711 on Build GRCh38
rs138030971 Chr.1: 154214478-154214478 on Build GRCh38
rs779062998 Chr.1: 154214467-154214467 on Build GRCh38
rs932028790 Chr.1: 154214738-154214738 on Build GRCh38
rs370941499 Chr.1: 154214557-154214557 on Build GRCh38
rs761379099 Chr.1: 154214520-154214520 on Build GRCh38
rs867073052 Chr.1: 154214452-154214452 on Build GRCh38
rs780445903 Chr.1: 154214417-154214417 on Build GRCh38

View Full Product Details