Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154220095-154220594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf43
Entrez Gene ID
25912
Gene Name
chromosome 1 open reading frame 43
Gene Aliases
HSPC012, NICE-3, NS5ATP4, S863-3
UniGene
Hs.287471
Gene Chromosome Location
Chr.1: 154206701-154220797 on Build GRCh38
Transcripts
NM_001098616.2 NM_001297717.1 NM_001297718.1 NM_001297720.1 NM_001297721.1 NM_001297723.1 NM_015449.3 NM_138740.3 NM_001127320.2

Gene Symbol
UBAP2L
Entrez Gene ID
9898
Gene Name
ubiquitin associated protein 2 like
Gene Aliases
NICE-4, NICE4
UniGene
Hs.490551
Gene Chromosome Location
Chr.1: 154220172-154271510 on Build GRCh38
Transcripts
NM_001098616.2 NM_001297717.1 NM_001297718.1 NM_001297720.1 NM_001297721.1 NM_001297723.1 NM_015449.3 NM_138740.3 NM_001127320.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6246685 c.18C>A p.N6K Chr.1: 154220390-154220390 on Build GRCh38
5462001 c.12C>T p.G4G Chr.1: 154220396-154220396 on Build GRCh38
6246686 c.18C>A p.N6K Chr.1: 154220390-154220390 on Build GRCh38
6121086 c.48G>A p.V16V Chr.1: 154220360-154220360 on Build GRCh38
4890014 c.46G>T p.V16L Chr.1: 154220362-154220362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544910425 Chr.1: 154220530-154220530 on Build GRCh38
rs778792352 Chr.1: 154220447-154220447 on Build GRCh38
rs372405388 Chr.1: 154220176-154220176 on Build GRCh38
rs546863511 Chr.1: 154220147-154220147 on Build GRCh38
rs1042343624 Chr.1: 154220256-154220256 on Build GRCh38
rs765392017 Chr.1: 154220391-154220391 on Build GRCh38
rs982418153 Chr.1: 154220149-154220149 on Build GRCh38
rs1027686538 Chr.1: 154220228-154220228 on Build GRCh38
rs772134342 Chr.1: 154220511-154220511 on Build GRCh38
rs1034178381 Chr.1: 154220289-154220289 on Build GRCh38
rs768583049 Chr.1: 154220326-154220326 on Build GRCh38
rs755982410 Chr.1: 154220427-154220427 on Build GRCh38
rs199795177 Chr.1: 154220313-154220313 on Build GRCh38
rs1015595808 Chr.1: 154220161-154220162 on Build GRCh38
rs767502435 Chr.1: 154220420-154220420 on Build GRCh38
rs761891258 Chr.1: 154220385-154220385 on Build GRCh38
rs945255163 Chr.1: 154220512-154220512 on Build GRCh38
rs1007565566 Chr.1: 154220471-154220471 on Build GRCh38
rs965159531 Chr.1: 154220496-154220496 on Build GRCh38
rs775402311 Chr.1: 154220298-154220298 on Build GRCh38
rs761535520 Chr.1: 154220383-154220383 on Build GRCh38
rs752629676 Chr.1: 154220421-154220421 on Build GRCh38
rs994990903 Chr.1: 154220293-154220293 on Build GRCh38
rs577368300 Chr.1: 154220523-154220523 on Build GRCh38
rs59310765 Chr.1: 154220121-154220121 on Build GRCh38
rs765207763 Chr.1: 154220298-154220298 on Build GRCh38
rs908226153 Chr.1: 154220151-154220151 on Build GRCh38
rs750437861 Chr.1: 154220407-154220407 on Build GRCh38
rs973609295 Chr.1: 154220129-154220129 on Build GRCh38
rs906664009 Chr.1: 154220374-154220374 on Build GRCh38
rs939330791 Chr.1: 154220525-154220525 on Build GRCh38
rs142062765 Chr.1: 154220384-154220384 on Build GRCh38
rs893710882 Chr.1: 154220238-154220238 on Build GRCh38
rs945305853 Chr.1: 154220252-154220252 on Build GRCh38
rs1055161232 Chr.1: 154220526-154220526 on Build GRCh38
rs921058487 Chr.1: 154220221-154220221 on Build GRCh38
rs755890291 Chr.1: 154220326-154220326 on Build GRCh38
rs187712710 Chr.1: 154220269-154220269 on Build GRCh38
rs368781363 Chr.1: 154220329-154220329 on Build GRCh38
rs746379089 Chr.1: 154220338-154220338 on Build GRCh38
rs776368653 Chr.1: 154220334-154220336 on Build GRCh38
rs751222962 Chr.1: 154220141-154220141 on Build GRCh38
rs756790555 Chr.1: 154220440-154220440 on Build GRCh38
rs895193293 Chr.1: 154220527-154220527 on Build GRCh38
rs753554183 Chr.1: 154220435-154220435 on Build GRCh38
rs147570400 Chr.1: 154220346-154220346 on Build GRCh38
rs989242164 Chr.1: 154220510-154220510 on Build GRCh38
rs879114206 Chr.1: 154220436-154220436 on Build GRCh38
rs563231443 Chr.1: 154220534-154220534 on Build GRCh38
rs549495292 Chr.1: 154220122-154220122 on Build GRCh38
rs765475824 Chr.1: 154220411-154220412 on Build GRCh38
rs747881686 Chr.1: 154220317-154220317 on Build GRCh38
rs879325787 Chr.1: 154220234-154220234 on Build GRCh38
rs771700046 Chr.1: 154220454-154220454 on Build GRCh38
rs143438903 Chr.1: 154220306-154220306 on Build GRCh38
rs747008979 Chr.1: 154220320-154220320 on Build GRCh38
rs530979752 Chr.1: 154220536-154220536 on Build GRCh38
rs995262222 Chr.1: 154220188-154220188 on Build GRCh38
rs751518489 Chr.1: 154220300-154220300 on Build GRCh38
rs763832724 Chr.1: 154220132-154220132 on Build GRCh38
rs370850706 Chr.1: 154220143-154220143 on Build GRCh38
rs115612322 Chr.1: 154220134-154220134 on Build GRCh38
rs576117896 Chr.1: 154220277-154220277 on Build GRCh38
rs182456089 Chr.1: 154220192-154220192 on Build GRCh38
rs773688709 Chr.1: 154220334-154220334 on Build GRCh38
rs1018989643 Chr.1: 154220490-154220490 on Build GRCh38
rs373644399 Chr.1: 154220430-154220430 on Build GRCh38
rs375001187 Chr.1: 154220461-154220461 on Build GRCh38
rs906529324 Chr.1: 154220268-154220268 on Build GRCh38
rs574997743 Chr.1: 154220567-154220567 on Build GRCh38
rs12087877 Chr.1: 154220133-154220133 on Build GRCh38
rs367742696 Chr.1: 154220453-154220453 on Build GRCh38
rs757397163 Chr.1: 154220294-154220294 on Build GRCh38
rs761188540 Chr.1: 154220351-154220351 on Build GRCh38
rs932473761 Chr.1: 154220233-154220233 on Build GRCh38
rs756060044 Chr.1: 154220307-154220307 on Build GRCh38
rs781036756 Chr.1: 154220172-154220172 on Build GRCh38
rs757019890 Chr.1: 154220168-154220168 on Build GRCh38
rs932430331 Chr.1: 154220501-154220501 on Build GRCh38
rs75107105 Chr.1: 154220292-154220292 on Build GRCh38
rs370072625 Chr.1: 154220418-154220418 on Build GRCh38
rs748288657 Chr.1: 154220489-154220489 on Build GRCh38
rs886616516 Chr.1: 154220538-154220538 on Build GRCh38
rs573636657 Chr.1: 154220330-154220330 on Build GRCh38
rs753532276 Chr.1: 154220304-154220304 on Build GRCh38
rs555056807 Chr.1: 154220369-154220369 on Build GRCh38
rs373879246 Chr.1: 154220474-154220474 on Build GRCh38
rs772645038 Chr.1: 154220344-154220344 on Build GRCh38
rs1042292365 Chr.1: 154220520-154220520 on Build GRCh38
rs778576651 Chr.1: 154220445-154220445 on Build GRCh38
rs750347960 Chr.1: 154220236-154220236 on Build GRCh38
rs539363198 Chr.1: 154220211-154220211 on Build GRCh38
rs923597050 Chr.1: 154220126-154220126 on Build GRCh38

View Full Product Details