Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42101104-42101640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf132
Entrez Gene ID
647024
Gene Name
chromosome 6 open reading frame 132
Gene Aliases
bA7K24.2
UniGene
Hs.444277
Gene Chromosome Location
Chr.6: 42101119-42142977 on Build GRCh38
Transcripts
NM_001164446.2

SNP Detail

SNP ID Chromosome Location
rs577712667 Chr.6: 42101164-42101164 on Build GRCh38
rs941481219 Chr.6: 42101436-42101436 on Build GRCh38
rs1031583790 Chr.6: 42101321-42101321 on Build GRCh38
rs190365161 Chr.6: 42101320-42101320 on Build GRCh38
rs967961164 Chr.6: 42101272-42101272 on Build GRCh38
rs918808556 Chr.6: 42101475-42101475 on Build GRCh38
rs150207869 Chr.6: 42101303-42101303 on Build GRCh38
rs185787054 Chr.6: 42101311-42101311 on Build GRCh38
rs933993863 Chr.6: 42101490-42101490 on Build GRCh38
rs942459406 Chr.6: 42101533-42101533 on Build GRCh38
rs764366787 Chr.6: 42101323-42101326 on Build GRCh38
rs972716415 Chr.6: 42101430-42101430 on Build GRCh38
rs191210555 Chr.6: 42101238-42101238 on Build GRCh38
rs938593405 Chr.6: 42101198-42101198 on Build GRCh38
rs928515375 Chr.6: 42101126-42101126 on Build GRCh38
rs546796005 Chr.6: 42101338-42101338 on Build GRCh38
rs1019252880 Chr.6: 42101259-42101261 on Build GRCh38
rs910041238 Chr.6: 42101420-42101420 on Build GRCh38
rs893949970 Chr.6: 42101568-42101568 on Build GRCh38
rs149389561 Chr.6: 42101536-42101536 on Build GRCh38
rs530393235 Chr.6: 42101376-42101378 on Build GRCh38
rs537438540 Chr.6: 42101537-42101537 on Build GRCh38
rs1043569922 Chr.6: 42101564-42101564 on Build GRCh38
rs557688767 Chr.6: 42101546-42101546 on Build GRCh38
rs535676064 Chr.6: 42101450-42101450 on Build GRCh38
rs969165044 Chr.6: 42101365-42101365 on Build GRCh38
rs1037063844 Chr.6: 42101187-42101187 on Build GRCh38
rs1052732672 Chr.6: 42101562-42101562 on Build GRCh38
rs1000176416 Chr.6: 42101556-42101556 on Build GRCh38
rs767492122 Chr.6: 42101228-42101228 on Build GRCh38
rs972998493 Chr.6: 42101455-42101455 on Build GRCh38
rs9369335 Chr.6: 42101569-42101569 on Build GRCh38
rs539953699 Chr.6: 42101220-42101220 on Build GRCh38
rs549282948 Chr.6: 42101487-42101487 on Build GRCh38
rs931054980 Chr.6: 42101229-42101229 on Build GRCh38
rs747847182 Chr.6: 42101470-42101470 on Build GRCh38
rs941577772 Chr.6: 42101171-42101171 on Build GRCh38
rs34180099 Chr.6: 42101597-42101598 on Build GRCh38
rs138794174 Chr.6: 42101366-42101366 on Build GRCh38
rs754872799 Chr.6: 42101362-42101362 on Build GRCh38
rs982081603 Chr.6: 42101369-42101369 on Build GRCh38
rs562081939 Chr.6: 42101264-42101264 on Build GRCh38
rs999289346 Chr.6: 42101570-42101570 on Build GRCh38
rs771283799 Chr.6: 42101615-42101615 on Build GRCh38
rs1030877513 Chr.6: 42101591-42101591 on Build GRCh38
rs564328805 Chr.6: 42101317-42101317 on Build GRCh38
rs1048721855 Chr.6: 42101465-42101465 on Build GRCh38
rs754144702 Chr.6: 42101337-42101337 on Build GRCh38
rs896368075 Chr.6: 42101602-42101602 on Build GRCh38
rs763002783 Chr.6: 42101158-42101158 on Build GRCh38
rs4413610 Chr.6: 42101237-42101237 on Build GRCh38
rs141946074 Chr.6: 42101246-42101246 on Build GRCh38
rs746550924 Chr.6: 42101612-42101612 on Build GRCh38
rs916351857 Chr.6: 42101359-42101359 on Build GRCh38

View Full Product Details