Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42101607-42102103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf132
Entrez Gene ID
647024
Gene Name
chromosome 6 open reading frame 132
Gene Aliases
bA7K24.2
UniGene
Hs.444277
Gene Chromosome Location
Chr.6: 42101119-42142977 on Build GRCh38
Transcripts
NM_001164446.2

SNP Detail

SNP ID Chromosome Location
rs573076800 Chr.6: 42101648-42101648 on Build GRCh38
rs146274449 Chr.6: 42102077-42102077 on Build GRCh38
rs983935932 Chr.6: 42101698-42101698 on Build GRCh38
rs567885726 Chr.6: 42101636-42101636 on Build GRCh38
rs775828865 Chr.6: 42101874-42101874 on Build GRCh38
rs34066163 Chr.6: 42101796-42101796 on Build GRCh38
rs1040803814 Chr.6: 42101733-42101733 on Build GRCh38
rs984481902 Chr.6: 42102032-42102032 on Build GRCh38
rs1023947078 Chr.6: 42102026-42102026 on Build GRCh38
rs1013493682 Chr.6: 42102015-42102015 on Build GRCh38
rs894031129 Chr.6: 42101768-42101768 on Build GRCh38
rs182135667 Chr.6: 42101643-42101643 on Build GRCh38
rs1035017430 Chr.6: 42101652-42101652 on Build GRCh38
rs141812997 Chr.6: 42101915-42101915 on Build GRCh38
rs935037152 Chr.6: 42101748-42101748 on Build GRCh38
rs896418618 Chr.6: 42101972-42101972 on Build GRCh38
rs962456668 Chr.6: 42101966-42101966 on Build GRCh38
rs1005852375 Chr.6: 42102048-42102048 on Build GRCh38
rs934257728 Chr.6: 42101799-42101799 on Build GRCh38
rs1018053540 Chr.6: 42102034-42102034 on Build GRCh38
rs911211953 Chr.6: 42101832-42101832 on Build GRCh38
rs942448232 Chr.6: 42101717-42101720 on Build GRCh38
rs555639374 Chr.6: 42101694-42101694 on Build GRCh38
rs770248247 Chr.6: 42101843-42101843 on Build GRCh38
rs904875480 Chr.6: 42102046-42102046 on Build GRCh38
rs984552573 Chr.6: 42101693-42101693 on Build GRCh38
rs763305112 Chr.6: 42101875-42101875 on Build GRCh38
rs962875127 Chr.6: 42101740-42101740 on Build GRCh38
rs987228877 Chr.6: 42101800-42101800 on Build GRCh38
rs1052550962 Chr.6: 42101750-42101750 on Build GRCh38
rs1016030166 Chr.6: 42102052-42102052 on Build GRCh38
rs969575340 Chr.6: 42101678-42101678 on Build GRCh38
rs1052273983 Chr.6: 42101916-42101916 on Build GRCh38
rs1045217320 Chr.6: 42101842-42101842 on Build GRCh38
rs972862773 Chr.6: 42101769-42101769 on Build GRCh38
rs993484256 Chr.6: 42101992-42101992 on Build GRCh38
rs962072537 Chr.6: 42101654-42101654 on Build GRCh38
rs139550136 Chr.6: 42101704-42101704 on Build GRCh38
rs903594200 Chr.6: 42101650-42101650 on Build GRCh38
rs186789928 Chr.6: 42101797-42101797 on Build GRCh38
rs146295373 Chr.6: 42101663-42101663 on Build GRCh38
rs758872180 Chr.6: 42101756-42101756 on Build GRCh38
rs556920493 Chr.6: 42101852-42101852 on Build GRCh38

View Full Product Details