Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42105350-42105850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf132
Entrez Gene ID
647024
Gene Name
chromosome 6 open reading frame 132
Gene Aliases
bA7K24.2
UniGene
Hs.444277
Gene Chromosome Location
Chr.6: 42101119-42142977 on Build GRCh38
Transcripts
NM_001164446.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4595012 c.2409G>A p.V803V Chr.6: 42105503-42105503 on Build GRCh38
6201078 c.2481G>A p.P827P Chr.6: 42105431-42105431 on Build GRCh38
6402465 c.2271G>T p.K757N Chr.6: 42105641-42105641 on Build GRCh38
5589936 c.2262C>T p.F754F Chr.6: 42105650-42105650 on Build GRCh38
6513958 c.2272A>T p.T758S Chr.6: 42105640-42105640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780727301 Chr.6: 42105667-42105667 on Build GRCh38
rs780997452 Chr.6: 42105616-42105617 on Build GRCh38
rs977052851 Chr.6: 42105708-42105708 on Build GRCh38
rs889363913 Chr.6: 42105627-42105627 on Build GRCh38
rs532154873 Chr.6: 42105496-42105496 on Build GRCh38
rs1019086883 Chr.6: 42105682-42105682 on Build GRCh38
rs765974828 Chr.6: 42105401-42105401 on Build GRCh38
rs1040769614 Chr.6: 42105573-42105573 on Build GRCh38
rs938474899 Chr.6: 42105469-42105469 on Build GRCh38
rs778785250 Chr.6: 42105572-42105572 on Build GRCh38
rs991315068 Chr.6: 42105816-42105816 on Build GRCh38
rs754761217 Chr.6: 42105568-42105568 on Build GRCh38
rs189132528 Chr.6: 42105613-42105613 on Build GRCh38
rs928473391 Chr.6: 42105452-42105452 on Build GRCh38
rs550034359 Chr.6: 42105534-42105536 on Build GRCh38
rs1022032283 Chr.6: 42105717-42105717 on Build GRCh38
rs745479779 Chr.6: 42105681-42105681 on Build GRCh38
rs754043462 Chr.6: 42105562-42105562 on Build GRCh38
rs762238354 Chr.6: 42105463-42105463 on Build GRCh38
rs959871864 Chr.6: 42105813-42105813 on Build GRCh38
rs770665248 Chr.6: 42105666-42105666 on Build GRCh38
rs773295551 Chr.6: 42105738-42105738 on Build GRCh38
rs1035740198 Chr.6: 42105803-42105803 on Build GRCh38
rs977310911 Chr.6: 42105434-42105434 on Build GRCh38
rs977132742 Chr.6: 42105431-42105431 on Build GRCh38
rs751562155 Chr.6: 42105440-42105442 on Build GRCh38
rs534680828 Chr.6: 42105790-42105790 on Build GRCh38
rs977930010 Chr.6: 42105771-42105771 on Build GRCh38
rs994751066 Chr.6: 42105646-42105646 on Build GRCh38
rs1055935250 Chr.6: 42105528-42105529 on Build GRCh38
rs769203924 Chr.6: 42105407-42105407 on Build GRCh38
rs953381778 Chr.6: 42105402-42105402 on Build GRCh38
rs1008629799 Chr.6: 42105675-42105675 on Build GRCh38
rs777490227 Chr.6: 42105620-42105620 on Build GRCh38
rs760755874 Chr.6: 42105768-42105768 on Build GRCh38
rs967167935 Chr.6: 42105706-42105706 on Build GRCh38
rs114870393 Chr.6: 42105619-42105619 on Build GRCh38
rs997736703 Chr.6: 42105398-42105398 on Build GRCh38
rs28666916 Chr.6: 42105503-42105503 on Build GRCh38
rs548054679 Chr.6: 42105579-42105579 on Build GRCh38
rs539058752 Chr.6: 42105767-42105767 on Build GRCh38
rs1028774845 Chr.6: 42105655-42105655 on Build GRCh38
rs953129772 Chr.6: 42105374-42105374 on Build GRCh38
rs550474773 Chr.6: 42105680-42105680 on Build GRCh38
rs770087436 Chr.6: 42105807-42105809 on Build GRCh38
rs373176623 Chr.6: 42105733-42105733 on Build GRCh38
rs916123187 Chr.6: 42105547-42105547 on Build GRCh38
rs572419947 Chr.6: 42105523-42105523 on Build GRCh38
rs761001773 Chr.6: 42105510-42105510 on Build GRCh38
rs569931106 Chr.6: 42105732-42105732 on Build GRCh38
rs767594617 Chr.6: 42105483-42105483 on Build GRCh38
rs559521854 Chr.6: 42105511-42105511 on Build GRCh38
rs764467174 Chr.6: 42105380-42105380 on Build GRCh38
rs991408405 Chr.6: 42105475-42105475 on Build GRCh38
rs997601228 Chr.6: 42105617-42105617 on Build GRCh38
rs745658503 Chr.6: 42105800-42105800 on Build GRCh38
rs762377172 Chr.6: 42105815-42105815 on Build GRCh38
rs987662058 Chr.6: 42105388-42105388 on Build GRCh38
rs886884647 Chr.6: 42105512-42105512 on Build GRCh38
rs915799324 Chr.6: 42105491-42105491 on Build GRCh38
rs746470368 Chr.6: 42105639-42105639 on Build GRCh38
rs774866054 Chr.6: 42105409-42105409 on Build GRCh38
rs181548689 Chr.6: 42105818-42105818 on Build GRCh38
rs967847983 Chr.6: 42105429-42105429 on Build GRCh38
rs957607413 Chr.6: 42105728-42105728 on Build GRCh38
rs201084953 Chr.6: 42105536-42105538 on Build GRCh38
rs527877555 Chr.6: 42105574-42105574 on Build GRCh38

View Full Product Details