Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42890184-42890681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf226
Entrez Gene ID
441150
Gene Name
chromosome 6 open reading frame 226
Gene Aliases
-
UniGene
Hs.571098
Gene Chromosome Location
Chr.6: 42890265-42890816 on Build GRCh38
Transcripts
NM_001008739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1150410 c.270G>C p.A90A Chr.6: 42890519-42890519 on Build GRCh38
6616469 c.222C>T p.T74T Chr.6: 42890567-42890567 on Build GRCh38
6616467 c.186G>T p.E62D Chr.6: 42890603-42890603 on Build GRCh38
6854872 c.210T>C p.L70L Chr.6: 42890579-42890579 on Build GRCh38
1193536 c.145C>T p.P49S Chr.6: 42890644-42890644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368560537 Chr.6: 42890429-42890429 on Build GRCh38
rs1056182526 Chr.6: 42890534-42890534 on Build GRCh38
rs752601575 Chr.6: 42890491-42890491 on Build GRCh38
rs372621973 Chr.6: 42890636-42890636 on Build GRCh38
rs753213869 Chr.6: 42890634-42890634 on Build GRCh38
rs765146498 Chr.6: 42890490-42890490 on Build GRCh38
rs369458563 Chr.6: 42890543-42890543 on Build GRCh38
rs374407918 Chr.6: 42890651-42890651 on Build GRCh38
rs893781994 Chr.6: 42890224-42890224 on Build GRCh38
rs752534789 Chr.6: 42890553-42890553 on Build GRCh38
rs770311584 Chr.6: 42890445-42890445 on Build GRCh38
rs552908108 Chr.6: 42890558-42890558 on Build GRCh38
rs375235600 Chr.6: 42890525-42890525 on Build GRCh38
rs570180761 Chr.6: 42890390-42890390 on Build GRCh38
rs541705086 Chr.6: 42890588-42890588 on Build GRCh38
rs368790615 Chr.6: 42890511-42890511 on Build GRCh38
rs753739740 Chr.6: 42890482-42890482 on Build GRCh38
rs377669808 Chr.6: 42890658-42890658 on Build GRCh38
rs200252004 Chr.6: 42890493-42890493 on Build GRCh38
rs748475121 Chr.6: 42890556-42890556 on Build GRCh38
rs777333094 Chr.6: 42890437-42890437 on Build GRCh38
rs777992267 Chr.6: 42890637-42890637 on Build GRCh38
rs375877873 Chr.6: 42890466-42890466 on Build GRCh38
rs771036494 Chr.6: 42890643-42890643 on Build GRCh38
rs769822643 Chr.6: 42890578-42890578 on Build GRCh38
rs369342145 Chr.6: 42890597-42890597 on Build GRCh38
rs759739353 Chr.6: 42890577-42890577 on Build GRCh38
rs761359843 Chr.6: 42890599-42890599 on Build GRCh38
rs777338565 Chr.6: 42890495-42890495 on Build GRCh38
rs1045218145 Chr.6: 42890282-42890282 on Build GRCh38
rs970793456 Chr.6: 42890372-42890372 on Build GRCh38
rs763760093 Chr.6: 42890526-42890526 on Build GRCh38
rs751186487 Chr.6: 42890531-42890531 on Build GRCh38
rs780563854 Chr.6: 42890541-42890541 on Build GRCh38
rs772131590 Chr.6: 42890653-42890653 on Build GRCh38
rs973652857 Chr.6: 42890433-42890433 on Build GRCh38
rs201818343 Chr.6: 42890500-42890500 on Build GRCh38
rs755654789 Chr.6: 42890618-42890618 on Build GRCh38
rs375303872 Chr.6: 42890591-42890591 on Build GRCh38
rs905388338 Chr.6: 42890297-42890297 on Build GRCh38
rs1056654456 Chr.6: 42890352-42890352 on Build GRCh38
rs1009550364 Chr.6: 42890319-42890319 on Build GRCh38
rs766380245 Chr.6: 42890520-42890520 on Build GRCh38
rs368260314 Chr.6: 42890471-42890471 on Build GRCh38
rs200635833 Chr.6: 42890513-42890513 on Build GRCh38
rs779346295 Chr.6: 42890622-42890622 on Build GRCh38
rs777731353 Chr.6: 42890632-42890632 on Build GRCh38
rs781108369 Chr.6: 42890660-42890660 on Build GRCh38
rs901644588 Chr.6: 42890605-42890605 on Build GRCh38
rs371127703 Chr.6: 42890456-42890456 on Build GRCh38
rs931006952 Chr.6: 42890451-42890451 on Build GRCh38
rs946843411 Chr.6: 42890270-42890270 on Build GRCh38
rs796928817 Chr.6: 42890346-42890346 on Build GRCh38
rs563745232 Chr.6: 42890656-42890656 on Build GRCh38
rs749607827 Chr.6: 42890454-42890454 on Build GRCh38
rs758174016 Chr.6: 42890605-42890605 on Build GRCh38
rs776561622 Chr.6: 42890521-42890521 on Build GRCh38
rs770014759 Chr.6: 42890648-42890648 on Build GRCh38
rs755460663 Chr.6: 42890550-42890550 on Build GRCh38
rs779439746 Chr.6: 42890554-42890554 on Build GRCh38
rs754119240 Chr.6: 42890313-42890313 on Build GRCh38
rs777991435 Chr.6: 42890567-42890567 on Build GRCh38
rs1033812891 Chr.6: 42890395-42890395 on Build GRCh38
rs61747887 Chr.6: 42890489-42890489 on Build GRCh38
rs965111451 Chr.6: 42890602-42890602 on Build GRCh38
rs761711154 Chr.6: 42890654-42890654 on Build GRCh38
rs774490283 Chr.6: 42890458-42890458 on Build GRCh38
rs772876669 Chr.6: 42890657-42890657 on Build GRCh38
rs747197988 Chr.6: 42890570-42890570 on Build GRCh38
rs746558219 Chr.6: 42890442-42890442 on Build GRCh38
rs199636430 Chr.6: 42890519-42890519 on Build GRCh38
rs769218539 Chr.6: 42890507-42890507 on Build GRCh38
rs988158541 Chr.6: 42890494-42890494 on Build GRCh38
rs908336524 Chr.6: 42890460-42890460 on Build GRCh38
rs532386078 Chr.6: 42890405-42890405 on Build GRCh38
rs370082945 Chr.6: 42890650-42890650 on Build GRCh38
rs1021078806 Chr.6: 42890351-42890351 on Build GRCh38
rs765124953 Chr.6: 42890490-42890490 on Build GRCh38
rs756721562 Chr.6: 42890537-42890537 on Build GRCh38
rs767252429 Chr.6: 42890616-42890616 on Build GRCh38
rs758813682 Chr.6: 42890662-42890662 on Build GRCh38
rs533399484 Chr.6: 42890420-42890423 on Build GRCh38
rs187805394 Chr.6: 42890326-42890326 on Build GRCh38
rs767788989 Chr.6: 42890469-42890469 on Build GRCh38
rs776834890 Chr.6: 42890575-42890575 on Build GRCh38
rs982453317 Chr.6: 42890596-42890596 on Build GRCh38
rs897456480 Chr.6: 42890365-42890365 on Build GRCh38
rs774688452 Chr.6: 42890510-42890510 on Build GRCh38
rs754759710 Chr.6: 42890486-42890486 on Build GRCh38
rs201027556 Chr.6: 42890504-42890504 on Build GRCh38
rs756747626 Chr.6: 42890503-42890503 on Build GRCh38
rs145346317 Chr.6: 42890505-42890505 on Build GRCh38
rs550289275 Chr.6: 42890274-42890274 on Build GRCh38
rs749908014 Chr.6: 42890617-42890617 on Build GRCh38
rs939106791 Chr.6: 42890327-42890327 on Build GRCh38
rs976824765 Chr.6: 42890485-42890486 on Build GRCh38
rs775598089 Chr.6: 42890579-42890579 on Build GRCh38
rs754887149 Chr.6: 42890446-42890446 on Build GRCh38
rs546911997 Chr.6: 42890625-42890625 on Build GRCh38
rs552509832 Chr.6: 42890449-42890449 on Build GRCh38
rs377397211 Chr.6: 42890647-42890647 on Build GRCh38
rs765323184 Chr.6: 42890523-42890523 on Build GRCh38
rs530327692 Chr.6: 42890244-42890244 on Build GRCh38
rs747460808 Chr.6: 42890642-42890642 on Build GRCh38
rs1041282525 Chr.6: 42890485-42890485 on Build GRCh38
rs372343244 Chr.6: 42890571-42890571 on Build GRCh38
rs760711598 Chr.6: 42890472-42890472 on Build GRCh38
rs868080338 Chr.6: 42890261-42890261 on Build GRCh38
rs959899387 Chr.6: 42890410-42890410 on Build GRCh38
rs367919477 Chr.6: 42890498-42890498 on Build GRCh38
rs766303573 Chr.6: 42890477-42890477 on Build GRCh38
rs1032769064 Chr.6: 42890215-42890215 on Build GRCh38
rs915511510 Chr.6: 42890240-42890240 on Build GRCh38

View Full Product Details