Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42890578-42891075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf226
Entrez Gene ID
441150
Gene Name
chromosome 6 open reading frame 226
Gene Aliases
-
UniGene
Hs.571098
Gene Chromosome Location
Chr.6: 42890265-42890816 on Build GRCh38
Transcripts
NM_001008739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616467 c.186G>T p.E62D Chr.6: 42890603-42890603 on Build GRCh38
1546261 c.41C>G p.S14C Chr.6: 42890748-42890748 on Build GRCh38
6854872 c.210T>C p.L70L Chr.6: 42890579-42890579 on Build GRCh38
1193536 c.145C>T p.P49S Chr.6: 42890644-42890644 on Build GRCh38
6616471 c.48A>G p.S16S Chr.6: 42890741-42890741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375054273 Chr.6: 42890716-42890716 on Build GRCh38
rs751788337 Chr.6: 42890690-42890690 on Build GRCh38
rs757635908 Chr.6: 42890769-42890769 on Build GRCh38
rs780463736 Chr.6: 42890825-42890825 on Build GRCh38
rs372621973 Chr.6: 42890636-42890636 on Build GRCh38
rs753213869 Chr.6: 42890634-42890634 on Build GRCh38
rs374407918 Chr.6: 42890651-42890651 on Build GRCh38
rs575149900 Chr.6: 42890857-42890857 on Build GRCh38
rs762592431 Chr.6: 42890839-42890839 on Build GRCh38
rs766074741 Chr.6: 42890754-42890754 on Build GRCh38
rs895274309 Chr.6: 42890919-42890919 on Build GRCh38
rs772062049 Chr.6: 42890788-42890788 on Build GRCh38
rs377669808 Chr.6: 42890658-42890658 on Build GRCh38
rs552805023 Chr.6: 42890797-42890797 on Build GRCh38
rs756383097 Chr.6: 42890776-42890776 on Build GRCh38
rs777992267 Chr.6: 42890637-42890637 on Build GRCh38
rs368524074 Chr.6: 42890836-42890836 on Build GRCh38
rs771036494 Chr.6: 42890643-42890643 on Build GRCh38
rs771870261 Chr.6: 42890729-42890729 on Build GRCh38
rs761359843 Chr.6: 42890599-42890599 on Build GRCh38
rs777723743 Chr.6: 42890819-42890819 on Build GRCh38
rs201854814 Chr.6: 42890789-42890789 on Build GRCh38
rs528504704 Chr.6: 42890701-42890701 on Build GRCh38
rs1019182457 Chr.6: 42891009-42891009 on Build GRCh38
rs761481195 Chr.6: 42891032-42891032 on Build GRCh38
rs997900028 Chr.6: 42891023-42891023 on Build GRCh38
rs746766056 Chr.6: 42890821-42890821 on Build GRCh38
rs201521264 Chr.6: 42890779-42890779 on Build GRCh38
rs768190246 Chr.6: 42890806-42890806 on Build GRCh38
rs772131590 Chr.6: 42890653-42890653 on Build GRCh38
rs1015230206 Chr.6: 42890780-42890780 on Build GRCh38
rs939506285 Chr.6: 42890764-42890764 on Build GRCh38
rs781362607 Chr.6: 42890693-42890701 on Build GRCh38
rs755654789 Chr.6: 42890618-42890618 on Build GRCh38
rs908970004 Chr.6: 42891014-42891014 on Build GRCh38
rs731068 Chr.6: 42891018-42891018 on Build GRCh38
rs61732354 Chr.6: 42890786-42890786 on Build GRCh38
rs779346295 Chr.6: 42890622-42890622 on Build GRCh38
rs565965651 Chr.6: 42890846-42890846 on Build GRCh38
rs750877717 Chr.6: 42890774-42890774 on Build GRCh38
rs774380918 Chr.6: 42890712-42890712 on Build GRCh38
rs777731353 Chr.6: 42890632-42890632 on Build GRCh38
rs879754666 Chr.6: 42891006-42891006 on Build GRCh38
rs568039522 Chr.6: 42890772-42890772 on Build GRCh38
rs370729479 Chr.6: 42890765-42890765 on Build GRCh38
rs796988533 Chr.6: 42890963-42890963 on Build GRCh38
rs886584317 Chr.6: 42891000-42891000 on Build GRCh38
rs781108369 Chr.6: 42890660-42890660 on Build GRCh38
rs762398593 Chr.6: 42890805-42890805 on Build GRCh38
rs901644588 Chr.6: 42890605-42890605 on Build GRCh38
rs548206294 Chr.6: 42890975-42890975 on Build GRCh38
rs759125362 Chr.6: 42890760-42890760 on Build GRCh38
rs758305538 Chr.6: 42890818-42890818 on Build GRCh38
rs997466679 Chr.6: 42890863-42890863 on Build GRCh38
rs776377013 Chr.6: 42890793-42890793 on Build GRCh38
rs563745232 Chr.6: 42890656-42890656 on Build GRCh38
rs758174016 Chr.6: 42890605-42890605 on Build GRCh38
rs536834696 Chr.6: 42890782-42890782 on Build GRCh38
rs770014759 Chr.6: 42890648-42890648 on Build GRCh38
rs760314245 Chr.6: 42890753-42890753 on Build GRCh38
rs200737014 Chr.6: 42890790-42890790 on Build GRCh38
rs754106658 Chr.6: 42890810-42890810 on Build GRCh38
rs374573056 Chr.6: 42890799-42890799 on Build GRCh38
rs757553859 Chr.6: 42890692-42890692 on Build GRCh38
rs370290579 Chr.6: 42891046-42891046 on Build GRCh38
rs764707418 Chr.6: 42890663-42890663 on Build GRCh38
rs761711154 Chr.6: 42890654-42890654 on Build GRCh38
rs965111451 Chr.6: 42890602-42890602 on Build GRCh38
rs770733930 Chr.6: 42890823-42890823 on Build GRCh38
rs190292644 Chr.6: 42890948-42890948 on Build GRCh38
rs756473101 Chr.6: 42890683-42890683 on Build GRCh38
rs772876669 Chr.6: 42890657-42890657 on Build GRCh38
rs748124881 Chr.6: 42890787-42890787 on Build GRCh38
rs577373047 Chr.6: 42890983-42890983 on Build GRCh38
rs902083419 Chr.6: 42891019-42891019 on Build GRCh38
rs528332162 Chr.6: 42890889-42890889 on Build GRCh38
rs563855470 Chr.6: 42890941-42890941 on Build GRCh38
rs1004986807 Chr.6: 42891007-42891007 on Build GRCh38
rs1033912654 Chr.6: 42890942-42890942 on Build GRCh38
rs543779928 Chr.6: 42890933-42890933 on Build GRCh38
rs1022389337 Chr.6: 42890929-42890936 on Build GRCh38
rs370082945 Chr.6: 42890650-42890650 on Build GRCh38
rs747851573 Chr.6: 42890728-42890728 on Build GRCh38
rs770328145 Chr.6: 42890743-42890744 on Build GRCh38
rs767252429 Chr.6: 42890616-42890616 on Build GRCh38
rs745537384 Chr.6: 42890832-42890832 on Build GRCh38
rs758813682 Chr.6: 42890662-42890662 on Build GRCh38
rs546269160 Chr.6: 42891028-42891028 on Build GRCh38
rs749434680 Chr.6: 42890700-42890700 on Build GRCh38
rs750785935 Chr.6: 42890808-42890808 on Build GRCh38
rs149447705 Chr.6: 42890837-42890837 on Build GRCh38
rs746001838 Chr.6: 42890742-42890743 on Build GRCh38
rs1053065068 Chr.6: 42891041-42891041 on Build GRCh38
rs770537259 Chr.6: 42890792-42890792 on Build GRCh38
rs1002628128 Chr.6: 42890939-42890939 on Build GRCh38
rs780649544 Chr.6: 42890825-42890825 on Build GRCh38
rs372855560 Chr.6: 42890680-42890680 on Build GRCh38
rs767069817 Chr.6: 42891040-42891040 on Build GRCh38
rs552393834 Chr.6: 42890679-42890679 on Build GRCh38
rs780269155 Chr.6: 42890684-42890684 on Build GRCh38
rs1048830046 Chr.6: 42890803-42890803 on Build GRCh38
rs749908014 Chr.6: 42890617-42890617 on Build GRCh38
rs1052926974 Chr.6: 42890870-42890870 on Build GRCh38
rs752829784 Chr.6: 42890814-42890814 on Build GRCh38
rs532610402 Chr.6: 42890674-42890674 on Build GRCh38
rs992396793 Chr.6: 42890993-42890993 on Build GRCh38
rs546911997 Chr.6: 42890625-42890625 on Build GRCh38
rs1042419720 Chr.6: 42890676-42890676 on Build GRCh38
rs377397211 Chr.6: 42890647-42890647 on Build GRCh38
rs747460808 Chr.6: 42890642-42890642 on Build GRCh38
rs534602952 Chr.6: 42890809-42890809 on Build GRCh38
rs953827933 Chr.6: 42891035-42891035 on Build GRCh38
rs568058004 Chr.6: 42891051-42891051 on Build GRCh38
rs769391428 Chr.6: 42890835-42890835 on Build GRCh38
rs376513309 Chr.6: 42890740-42890740 on Build GRCh38
rs746099981 Chr.6: 42890682-42890682 on Build GRCh38
rs989330078 Chr.6: 42890791-42890791 on Build GRCh38
rs1004055453 Chr.6: 42890707-42890707 on Build GRCh38
rs779042260 Chr.6: 42890813-42890813 on Build GRCh38
rs775979355 Chr.6: 42890757-42890757 on Build GRCh38

View Full Product Details