Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43038764-43039275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL7
Entrez Gene ID
9820
Gene Name
cullin 7
Gene Aliases
3M1, KIAA0076, dJ20C7.5
UniGene
Hs.520136
Gene Chromosome Location
Chr.6: 43037617-43053950 on Build GRCh38
Transcripts
NM_001168370.1 NM_014780.4 XM_005249503.2 XM_006715285.1 XM_011515019.2 XM_011515020.2 XM_011515021.1 XM_017011533.1 XM_017011534.1 XM_017011535.1 XM_017011536.1 XM_017011537.1 XM_017011538.1 XM_017011539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1079407 c.4318C>T p.R1440* Chr.6: 43038964-43038964 on Build GRCh38
7317210 c.4689G>A p.L1563L Chr.6: 43038845-43038845 on Build GRCh38
1162562 c.4429A>T p.N1477Y Chr.6: 43038853-43038853 on Build GRCh38
4865410 c.4570C>T p.R1524* Chr.6: 43038964-43038964 on Build GRCh38
4647028 c.4608C>G p.G1536G Chr.6: 43038926-43038926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749958141 Chr.6: 43039027-43039027 on Build GRCh38
rs148472550 Chr.6: 43038959-43038959 on Build GRCh38
rs748555538 Chr.6: 43038964-43038964 on Build GRCh38
rs753530058 Chr.6: 43038824-43038824 on Build GRCh38
rs1056009860 Chr.6: 43039239-43039239 on Build GRCh38
rs772138262 Chr.6: 43038979-43038979 on Build GRCh38
rs773464504 Chr.6: 43038980-43038980 on Build GRCh38
rs781339095 Chr.6: 43039090-43039090 on Build GRCh38
rs867557529 Chr.6: 43039094-43039094 on Build GRCh38
rs1002776590 Chr.6: 43039067-43039067 on Build GRCh38
rs752498664 Chr.6: 43038910-43038910 on Build GRCh38
rs990698686 Chr.6: 43039207-43039207 on Build GRCh38
rs121918229 Chr.6: 43038891-43038891 on Build GRCh38
rs3805945 Chr.6: 43039244-43039244 on Build GRCh38
rs766373189 Chr.6: 43038821-43038821 on Build GRCh38
rs994593899 Chr.6: 43039024-43039024 on Build GRCh38
rs776085552 Chr.6: 43038850-43038850 on Build GRCh38
rs765006933 Chr.6: 43038901-43038901 on Build GRCh38
rs948407924 Chr.6: 43039022-43039022 on Build GRCh38
rs750529899 Chr.6: 43038802-43038802 on Build GRCh38
rs191382580 Chr.6: 43039068-43039068 on Build GRCh38
rs752447374 Chr.6: 43039004-43039004 on Build GRCh38
rs34563914 Chr.6: 43039046-43039047 on Build GRCh38
rs368643336 Chr.6: 43038839-43038839 on Build GRCh38
rs150057048 Chr.6: 43039069-43039070 on Build GRCh38
rs1018837222 Chr.6: 43039108-43039108 on Build GRCh38
rs959116264 Chr.6: 43038799-43038799 on Build GRCh38
rs749808640 Chr.6: 43038950-43038950 on Build GRCh38
rs537632525 Chr.6: 43039234-43039234 on Build GRCh38
rs1045968227 Chr.6: 43038958-43038958 on Build GRCh38
rs546151345 Chr.6: 43038997-43038997 on Build GRCh38
rs756768833 Chr.6: 43038948-43038948 on Build GRCh38
rs146472429 Chr.6: 43039168-43039168 on Build GRCh38
rs775516382 Chr.6: 43039023-43039023 on Build GRCh38
rs907135687 Chr.6: 43039047-43039047 on Build GRCh38
rs200803189 Chr.6: 43038881-43038881 on Build GRCh38
rs754748401 Chr.6: 43038828-43038828 on Build GRCh38
rs779355111 Chr.6: 43038963-43038963 on Build GRCh38
rs535028437 Chr.6: 43039196-43039196 on Build GRCh38
rs1001617999 Chr.6: 43038995-43038995 on Build GRCh38
rs746530839 Chr.6: 43038836-43038836 on Build GRCh38
rs149530175 Chr.6: 43038851-43038851 on Build GRCh38
rs546532902 Chr.6: 43039175-43039175 on Build GRCh38
rs201252688 Chr.6: 43039012-43039012 on Build GRCh38
rs753070951 Chr.6: 43039241-43039241 on Build GRCh38
rs777085030 Chr.6: 43038835-43038835 on Build GRCh38
rs767857304 Chr.6: 43038864-43038864 on Build GRCh38
rs913337678 Chr.6: 43039203-43039203 on Build GRCh38
rs755940230 Chr.6: 43038812-43038812 on Build GRCh38
rs879030495 Chr.6: 43039059-43039059 on Build GRCh38
rs1046167612 Chr.6: 43039223-43039223 on Build GRCh38
rs542050235 Chr.6: 43039018-43039018 on Build GRCh38
rs762069855 Chr.6: 43038862-43038862 on Build GRCh38
rs766421323 Chr.6: 43038880-43038880 on Build GRCh38
rs201386557 Chr.6: 43038865-43038865 on Build GRCh38
rs776636658 Chr.6: 43039000-43039000 on Build GRCh38
rs769592182 Chr.6: 43038996-43038996 on Build GRCh38
rs774922730 Chr.6: 43038855-43038855 on Build GRCh38
rs1043997680 Chr.6: 43039041-43039041 on Build GRCh38
rs965956457 Chr.6: 43039144-43039144 on Build GRCh38
rs763842022 Chr.6: 43039007-43039007 on Build GRCh38
rs757974664 Chr.6: 43038935-43038935 on Build GRCh38
rs58659748 Chr.6: 43039161-43039161 on Build GRCh38
rs568353076 Chr.6: 43039206-43039206 on Build GRCh38
rs550430647 Chr.6: 43039089-43039089 on Build GRCh38
rs758101201 Chr.6: 43038833-43038833 on Build GRCh38
rs946063204 Chr.6: 43038957-43038957 on Build GRCh38
rs760789869 Chr.6: 43039139-43039139 on Build GRCh38
rs144111004 Chr.6: 43038936-43038936 on Build GRCh38
rs144154816 Chr.6: 43039001-43039001 on Build GRCh38
rs928856441 Chr.6: 43039228-43039228 on Build GRCh38
rs936080339 Chr.6: 43038886-43038886 on Build GRCh38
rs774696952 Chr.6: 43038830-43038830 on Build GRCh38
rs767699502 Chr.6: 43038800-43038800 on Build GRCh38
rs773406019 Chr.6: 43039083-43039083 on Build GRCh38
rs121918228 Chr.6: 43038949-43038949 on Build GRCh38
rs186188012 Chr.6: 43039036-43039036 on Build GRCh38
rs747798028 Chr.6: 43038832-43038832 on Build GRCh38
rs986108755 Chr.6: 43038890-43038890 on Build GRCh38
rs914276279 Chr.6: 43039016-43039016 on Build GRCh38
rs944492565 Chr.6: 43038859-43038859 on Build GRCh38
rs769111630 Chr.6: 43038852-43038852 on Build GRCh38
rs757900162 Chr.6: 43039234-43039234 on Build GRCh38
rs144245050 Chr.6: 43038829-43038829 on Build GRCh38
rs557198417 Chr.6: 43039238-43039238 on Build GRCh38
rs866647453 Chr.6: 43039172-43039172 on Build GRCh38
rs957230547 Chr.6: 43039171-43039171 on Build GRCh38
rs554836006 Chr.6: 43039204-43039204 on Build GRCh38
rs181427106 Chr.6: 43039177-43039177 on Build GRCh38
rs937585371 Chr.6: 43039232-43039232 on Build GRCh38
rs780739890 Chr.6: 43039021-43039021 on Build GRCh38
rs377169342 Chr.6: 43038985-43038985 on Build GRCh38

View Full Product Details