Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43427463-43427967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC10
Entrez Gene ID
89845
Gene Name
ATP binding cassette subfamily C member 10
Gene Aliases
EST182763, MRP7, SIMRP7
UniGene
Hs.55879
Gene Chromosome Location
Chr.6: 43427538-43451994 on Build GRCh38
Transcripts
NM_001198934.1 XM_011514974.2 XM_011514983.2 XM_017011445.1 XM_017011446.1 XM_017011447.1 XM_017011448.1 XM_017011449.1 XM_017011450.1 XM_017011451.1 XM_017011452.1 XM_017011453.1 XM_017011454.1

SNP Detail

SNP ID Chromosome Location
rs766631399 Chr.6: 43427894-43427894 on Build GRCh38
rs904770183 Chr.6: 43427617-43427617 on Build GRCh38
rs185518884 Chr.6: 43427796-43427796 on Build GRCh38
rs760350298 Chr.6: 43427919-43427919 on Build GRCh38
rs772414760 Chr.6: 43427944-43427944 on Build GRCh38
rs573526685 Chr.6: 43427526-43427526 on Build GRCh38
rs17287914 Chr.6: 43427658-43427658 on Build GRCh38
rs772201777 Chr.6: 43427880-43427880 on Build GRCh38
rs758154128 Chr.6: 43427916-43427916 on Build GRCh38
rs769474089 Chr.6: 43427933-43427933 on Build GRCh38
rs1034789987 Chr.6: 43427566-43427566 on Build GRCh38
rs869161532 Chr.6: 43427646-43427647 on Build GRCh38
rs922428656 Chr.6: 43427507-43427507 on Build GRCh38
rs780732377 Chr.6: 43427927-43427927 on Build GRCh38
rs759433166 Chr.6: 43427900-43427900 on Build GRCh38
rs758018580 Chr.6: 43427831-43427831 on Build GRCh38
rs762194626 Chr.6: 43427869-43427869 on Build GRCh38
rs568711910 Chr.6: 43427556-43427556 on Build GRCh38
rs902213698 Chr.6: 43427548-43427548 on Build GRCh38
rs1012428033 Chr.6: 43427586-43427586 on Build GRCh38
rs753951985 Chr.6: 43427899-43427899 on Build GRCh38
rs575147150 Chr.6: 43427933-43427933 on Build GRCh38
rs952330331 Chr.6: 43427700-43427700 on Build GRCh38
rs773510341 Chr.6: 43427891-43427891 on Build GRCh38
rs372777118 Chr.6: 43427937-43427937 on Build GRCh38
rs745495146 Chr.6: 43427913-43427914 on Build GRCh38
rs756893160 Chr.6: 43427923-43427923 on Build GRCh38
rs539893819 Chr.6: 43427747-43427747 on Build GRCh38
rs1029964484 Chr.6: 43427527-43427527 on Build GRCh38
rs900341984 Chr.6: 43427763-43427763 on Build GRCh38
rs917900373 Chr.6: 43427672-43427672 on Build GRCh38
rs752989099 Chr.6: 43427512-43427512 on Build GRCh38
rs780679266 Chr.6: 43427859-43427859 on Build GRCh38
rs556076955 Chr.6: 43427762-43427762 on Build GRCh38
rs772875961 Chr.6: 43427863-43427863 on Build GRCh38
rs985482843 Chr.6: 43427707-43427707 on Build GRCh38
rs566841132 Chr.6: 43427781-43427781 on Build GRCh38
rs889723690 Chr.6: 43427534-43427534 on Build GRCh38
rs896622274 Chr.6: 43427583-43427583 on Build GRCh38
rs182453024 Chr.6: 43427541-43427541 on Build GRCh38
rs570967327 Chr.6: 43427733-43427733 on Build GRCh38
rs933865166 Chr.6: 43427510-43427510 on Build GRCh38
rs779424095 Chr.6: 43427941-43427941 on Build GRCh38
rs770628225 Chr.6: 43427935-43427935 on Build GRCh38
rs949437847 Chr.6: 43427686-43427686 on Build GRCh38
rs933791200 Chr.6: 43427789-43427789 on Build GRCh38
rs528812923 Chr.6: 43427494-43427494 on Build GRCh38
rs548850526 Chr.6: 43427543-43427543 on Build GRCh38
rs1024272138 Chr.6: 43427619-43427619 on Build GRCh38
rs555237268 Chr.6: 43427901-43427901 on Build GRCh38
rs79196551 Chr.6: 43427635-43427635 on Build GRCh38
rs748471384 Chr.6: 43427943-43427943 on Build GRCh38
rs752458700 Chr.6: 43427903-43427903 on Build GRCh38
rs777435275 Chr.6: 43427836-43427836 on Build GRCh38
rs751283491 Chr.6: 43427920-43427920 on Build GRCh38
rs999262764 Chr.6: 43427557-43427557 on Build GRCh38
rs770365542 Chr.6: 43427853-43427853 on Build GRCh38
rs746539483 Chr.6: 43427845-43427845 on Build GRCh38
rs1010955870 Chr.6: 43427537-43427537 on Build GRCh38
rs1040472959 Chr.6: 43427492-43427492 on Build GRCh38
rs536021138 Chr.6: 43427616-43427616 on Build GRCh38
rs1006007883 Chr.6: 43427719-43427719 on Build GRCh38
rs531125172 Chr.6: 43427643-43427643 on Build GRCh38
rs749678764 Chr.6: 43427582-43427582 on Build GRCh38
rs773706758 Chr.6: 43427945-43427945 on Build GRCh38
rs938241194 Chr.6: 43427578-43427578 on Build GRCh38
rs1014873256 Chr.6: 43427726-43427726 on Build GRCh38
rs768966379 Chr.6: 43427692-43427692 on Build GRCh38
rs760840739 Chr.6: 43427892-43427892 on Build GRCh38

View Full Product Details