Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44408241-44408763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC5L
Entrez Gene ID
988
Gene Name
cell division cycle 5 like
Gene Aliases
CDC5, CDC5-LIKE, CEF1, PCDC5RP, dJ319D22.1
UniGene
Hs.485471
Gene Chromosome Location
Chr.6: 44387514-44450424 on Build GRCh38
Transcripts
NM_001253.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670737 c.961C>T p.R321C Chr.6: 44408501-44408501 on Build GRCh38
6253843 c.1045G>T p.A349S Chr.6: 44408585-44408585 on Build GRCh38
1079789 c.1061G>A p.R354Q Chr.6: 44408601-44408601 on Build GRCh38
1079788 c.991A>G p.N331D Chr.6: 44408531-44408531 on Build GRCh38
5729648 c.1057C>T p.P353S Chr.6: 44408597-44408597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941493029 Chr.6: 44408502-44408502 on Build GRCh38
rs773251690 Chr.6: 44408620-44408620 on Build GRCh38
rs536765287 Chr.6: 44408520-44408520 on Build GRCh38
rs11552843 Chr.6: 44408498-44408498 on Build GRCh38
rs370507115 Chr.6: 44408647-44408647 on Build GRCh38
rs745430137 Chr.6: 44408538-44408538 on Build GRCh38
rs769584179 Chr.6: 44408354-44408354 on Build GRCh38
rs770590185 Chr.6: 44408421-44408421 on Build GRCh38
rs376004864 Chr.6: 44408565-44408565 on Build GRCh38
rs755405339 Chr.6: 44408506-44408506 on Build GRCh38
rs571565932 Chr.6: 44408401-44408401 on Build GRCh38
rs9381320 Chr.6: 44408535-44408535 on Build GRCh38
rs748815520 Chr.6: 44408637-44408637 on Build GRCh38
rs1022052815 Chr.6: 44408491-44408491 on Build GRCh38
rs763145775 Chr.6: 44408500-44408500 on Build GRCh38
rs375757131 Chr.6: 44408267-44408268 on Build GRCh38
rs757947517 Chr.6: 44408658-44408658 on Build GRCh38
rs992067727 Chr.6: 44408404-44408404 on Build GRCh38
rs934944592 Chr.6: 44408702-44408702 on Build GRCh38
rs771826078 Chr.6: 44408441-44408441 on Build GRCh38
rs375896127 Chr.6: 44408644-44408644 on Build GRCh38
rs780275529 Chr.6: 44408395-44408395 on Build GRCh38
rs930670024 Chr.6: 44408617-44408617 on Build GRCh38
rs199688549 Chr.6: 44408629-44408629 on Build GRCh38
rs576571561 Chr.6: 44408382-44408382 on Build GRCh38
rs528119693 Chr.6: 44408324-44408324 on Build GRCh38
rs763840295 Chr.6: 44408341-44408341 on Build GRCh38
rs892126229 Chr.6: 44408471-44408471 on Build GRCh38
rs778564165 Chr.6: 44408525-44408525 on Build GRCh38
rs972497663 Chr.6: 44408322-44408322 on Build GRCh38
rs1024863105 Chr.6: 44408708-44408708 on Build GRCh38
rs750565964 Chr.6: 44408496-44408496 on Build GRCh38
rs567164340 Chr.6: 44408321-44408321 on Build GRCh38
rs1038853577 Chr.6: 44408564-44408564 on Build GRCh38
rs756763383 Chr.6: 44408523-44408523 on Build GRCh38
rs779796790 Chr.6: 44408563-44408563 on Build GRCh38
rs76056385 Chr.6: 44408281-44408281 on Build GRCh38
rs144083420 Chr.6: 44408501-44408501 on Build GRCh38
rs776484259 Chr.6: 44408581-44408581 on Build GRCh38
rs751834925 Chr.6: 44408505-44408505 on Build GRCh38
rs771602856 Chr.6: 44408552-44408552 on Build GRCh38
rs200986556 Chr.6: 44408572-44408572 on Build GRCh38
rs748772236 Chr.6: 44408405-44408405 on Build GRCh38
rs148664123 Chr.6: 44408600-44408600 on Build GRCh38
rs753603218 Chr.6: 44408453-44408453 on Build GRCh38
rs778897032 Chr.6: 44408268-44408268 on Build GRCh38
rs546558952 Chr.6: 44408379-44408379 on Build GRCh38
rs372318460 Chr.6: 44408397-44408397 on Build GRCh38
rs972004000 Chr.6: 44408579-44408579 on Build GRCh38
rs141567880 Chr.6: 44408455-44408455 on Build GRCh38
rs200890159 Chr.6: 44408547-44408547 on Build GRCh38
rs558975912 Chr.6: 44408707-44408707 on Build GRCh38
rs369777781 Chr.6: 44408482-44408482 on Build GRCh38
rs763130791 Chr.6: 44408623-44408623 on Build GRCh38
rs755414360 Chr.6: 44408398-44408398 on Build GRCh38
rs969244180 Chr.6: 44408738-44408738 on Build GRCh38
rs781506877 Chr.6: 44408512-44408512 on Build GRCh38
rs193041111 Chr.6: 44408282-44408282 on Build GRCh38
rs200087867 Chr.6: 44408469-44408469 on Build GRCh38
rs370106997 Chr.6: 44408678-44408678 on Build GRCh38
rs760568173 Chr.6: 44408461-44408461 on Build GRCh38
rs149525934 Chr.6: 44408475-44408475 on Build GRCh38
rs539044223 Chr.6: 44408614-44408614 on Build GRCh38
rs1006568146 Chr.6: 44408582-44408582 on Build GRCh38
rs373916286 Chr.6: 44408487-44408487 on Build GRCh38
rs372684842 Chr.6: 44408640-44408640 on Build GRCh38
rs1051362293 Chr.6: 44408575-44408575 on Build GRCh38
rs891353395 Chr.6: 44408580-44408580 on Build GRCh38
rs577127953 Chr.6: 44408717-44408717 on Build GRCh38
rs889166361 Chr.6: 44408364-44408364 on Build GRCh38
rs748222402 Chr.6: 44408513-44408513 on Build GRCh38
rs13213637 Chr.6: 44408651-44408651 on Build GRCh38
rs372513973 Chr.6: 44408423-44408423 on Build GRCh38
rs756677489 Chr.6: 44408645-44408645 on Build GRCh38
rs930762841 Chr.6: 44408347-44408347 on Build GRCh38
rs967788437 Chr.6: 44408271-44408271 on Build GRCh38
rs377117589 Chr.6: 44408434-44408434 on Build GRCh38
rs749923178 Chr.6: 44408649-44408649 on Build GRCh38
rs1049149515 Chr.6: 44408353-44408353 on Build GRCh38
rs768509617 Chr.6: 44408568-44408568 on Build GRCh38

View Full Product Details