Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46144852-46145353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP4
Entrez Gene ID
22875
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative)
Gene Aliases
NPP4
UniGene
Hs.643497
Gene Chromosome Location
Chr.6: 46129964-46146699 on Build GRCh38
Transcripts
NM_014936.4

SNP Detail

SNP ID Chromosome Location
rs995025932 Chr.6: 46145136-46145136 on Build GRCh38
rs899366376 Chr.6: 46145063-46145063 on Build GRCh38
rs896120328 Chr.6: 46145262-46145262 on Build GRCh38
rs947928343 Chr.6: 46144999-46144999 on Build GRCh38
rs1030757527 Chr.6: 46145323-46145323 on Build GRCh38
rs879031761 Chr.6: 46145107-46145107 on Build GRCh38
rs1053096932 Chr.6: 46145167-46145167 on Build GRCh38
rs200153618 Chr.6: 46145296-46145296 on Build GRCh38
rs755496567 Chr.6: 46145278-46145278 on Build GRCh38
rs779748665 Chr.6: 46145117-46145117 on Build GRCh38
rs370684103 Chr.6: 46144909-46144909 on Build GRCh38
rs376992511 Chr.6: 46145264-46145264 on Build GRCh38
rs184844576 Chr.6: 46145242-46145242 on Build GRCh38
rs566731876 Chr.6: 46145147-46145149 on Build GRCh38
rs548148268 Chr.6: 46145255-46145255 on Build GRCh38
rs936509195 Chr.6: 46144940-46144940 on Build GRCh38
rs572916314 Chr.6: 46145034-46145034 on Build GRCh38
rs1009089615 Chr.6: 46145206-46145206 on Build GRCh38
rs889386344 Chr.6: 46145201-46145201 on Build GRCh38
rs955362962 Chr.6: 46145105-46145105 on Build GRCh38
rs377707148 Chr.6: 46145190-46145190 on Build GRCh38
rs192082472 Chr.6: 46145030-46145030 on Build GRCh38
rs988361049 Chr.6: 46145130-46145130 on Build GRCh38
rs1029710141 Chr.6: 46145074-46145074 on Build GRCh38
rs993336096 Chr.6: 46144949-46144949 on Build GRCh38
rs1036447006 Chr.6: 46145049-46145049 on Build GRCh38
rs940779970 Chr.6: 46145048-46145048 on Build GRCh38
rs1056143616 Chr.6: 46145241-46145241 on Build GRCh38
rs887588550 Chr.6: 46144971-46144971 on Build GRCh38
rs772364892 Chr.6: 46144999-46145002 on Build GRCh38
rs1047616370 Chr.6: 46145318-46145318 on Build GRCh38
rs896399744 Chr.6: 46144891-46144891 on Build GRCh38
rs150341420 Chr.6: 46145292-46145292 on Build GRCh38
rs776173330 Chr.6: 46145215-46145215 on Build GRCh38
rs937796739 Chr.6: 46145231-46145231 on Build GRCh38
rs555583885 Chr.6: 46145230-46145230 on Build GRCh38
rs3777632 Chr.6: 46145150-46145150 on Build GRCh38
rs975401896 Chr.6: 46145073-46145073 on Build GRCh38
rs754430791 Chr.6: 46145189-46145189 on Build GRCh38
rs114300306 Chr.6: 46144982-46144982 on Build GRCh38
rs569041471 Chr.6: 46144963-46144963 on Build GRCh38

View Full Product Details