Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46145974-46146483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP4
Entrez Gene ID
22875
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative)
Gene Aliases
NPP4
UniGene
Hs.643497
Gene Chromosome Location
Chr.6: 46129964-46146699 on Build GRCh38
Transcripts
NM_014936.4

SNP Detail

SNP ID Chromosome Location
rs1042480517 Chr.6: 46146296-46146296 on Build GRCh38
rs550877643 Chr.6: 46146173-46146173 on Build GRCh38
rs775423114 Chr.6: 46146089-46146089 on Build GRCh38
rs760931327 Chr.6: 46146343-46146343 on Build GRCh38
rs983134564 Chr.6: 46146028-46146028 on Build GRCh38
rs554189531 Chr.6: 46146293-46146293 on Build GRCh38
rs904964384 Chr.6: 46146299-46146299 on Build GRCh38
rs933124843 Chr.6: 46146129-46146129 on Build GRCh38
rs149477655 Chr.6: 46146032-46146032 on Build GRCh38
rs1034872075 Chr.6: 46146454-46146454 on Build GRCh38
rs566893136 Chr.6: 46146355-46146355 on Build GRCh38
rs1055114708 Chr.6: 46146265-46146265 on Build GRCh38
rs893507335 Chr.6: 46146267-46146267 on Build GRCh38
rs548273280 Chr.6: 46146300-46146300 on Build GRCh38
rs1033874445 Chr.6: 46146311-46146311 on Build GRCh38
rs1051626332 Chr.6: 46146280-46146280 on Build GRCh38
rs1801905 Chr.6: 46146350-46146350 on Build GRCh38
rs1000891218 Chr.6: 46146302-46146302 on Build GRCh38
rs539879527 Chr.6: 46146120-46146120 on Build GRCh38
rs946384679 Chr.6: 46146281-46146281 on Build GRCh38
rs976235117 Chr.6: 46146254-46146254 on Build GRCh38
rs934953600 Chr.6: 46146264-46146264 on Build GRCh38
rs908829273 Chr.6: 46146043-46146043 on Build GRCh38
rs921867486 Chr.6: 46146121-46146121 on Build GRCh38
rs9763 Chr.6: 46146118-46146118 on Build GRCh38
rs562701474 Chr.6: 46146179-46146179 on Build GRCh38
rs895430694 Chr.6: 46146335-46146335 on Build GRCh38
rs565549222 Chr.6: 46146164-46146164 on Build GRCh38
rs972371074 Chr.6: 46146042-46146042 on Build GRCh38
rs1025135313 Chr.6: 46146353-46146353 on Build GRCh38
rs986114534 Chr.6: 46146137-46146137 on Build GRCh38
rs950885981 Chr.6: 46146361-46146363 on Build GRCh38
rs924769916 Chr.6: 46146256-46146256 on Build GRCh38
rs983570957 Chr.6: 46146386-46146386 on Build GRCh38
rs768002960 Chr.6: 46146444-46146444 on Build GRCh38
rs532664880 Chr.6: 46146172-46146172 on Build GRCh38
rs534183827 Chr.6: 46146381-46146381 on Build GRCh38
rs1034829542 Chr.6: 46146003-46146003 on Build GRCh38

View Full Product Details