Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49517336-49517871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYATL3
Entrez Gene ID
389396
Gene Name
glycine-N-acyltransferase like 3
Gene Aliases
C6orf140, bA28H17.2
UniGene
Hs.585172
Gene Chromosome Location
Chr.6: 49499958-49528064 on Build GRCh38
Transcripts
NM_001010904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1445000 c.257G>A p.R86Q Chr.6: 49517500-49517500 on Build GRCh38
320614 c.207T>A p.D69E Chr.6: 49517450-49517450 on Build GRCh38
1444999 c.221C>T p.A74V Chr.6: 49517464-49517464 on Build GRCh38
6193309 c.205G>C p.D69H Chr.6: 49517448-49517448 on Build GRCh38
150100 c.308T>C p.I103T Chr.6: 49517551-49517551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013892904 Chr.6: 49517744-49517744 on Build GRCh38
rs933972627 Chr.6: 49517635-49517635 on Build GRCh38
rs571013833 Chr.6: 49517784-49517784 on Build GRCh38
rs902671160 Chr.6: 49517576-49517580 on Build GRCh38
rs1029651077 Chr.6: 49517376-49517376 on Build GRCh38
rs562359963 Chr.6: 49517480-49517480 on Build GRCh38
rs962527014 Chr.6: 49517401-49517401 on Build GRCh38
rs1025982881 Chr.6: 49517753-49517753 on Build GRCh38
rs557005566 Chr.6: 49517762-49517762 on Build GRCh38
rs567199488 Chr.6: 49517688-49517688 on Build GRCh38
rs909995533 Chr.6: 49517584-49517584 on Build GRCh38
rs909704819 Chr.6: 49517769-49517769 on Build GRCh38
rs904298824 Chr.6: 49517701-49517701 on Build GRCh38
rs959705332 Chr.6: 49517727-49517727 on Build GRCh38
rs971238333 Chr.6: 49517673-49517673 on Build GRCh38
rs534547893 Chr.6: 49517693-49517693 on Build GRCh38
rs267601060 Chr.6: 49517448-49517448 on Build GRCh38
rs771386137 Chr.6: 49517562-49517562 on Build GRCh38
rs553096176 Chr.6: 49517694-49517694 on Build GRCh38
rs375109506 Chr.6: 49517509-49517509 on Build GRCh38
rs563265637 Chr.6: 49517370-49517370 on Build GRCh38
rs968459231 Chr.6: 49517517-49517517 on Build GRCh38
rs889997094 Chr.6: 49517809-49517809 on Build GRCh38
rs1054082835 Chr.6: 49517707-49517707 on Build GRCh38
rs776295863 Chr.6: 49517395-49517395 on Build GRCh38
rs35128069 Chr.6: 49517575-49517576 on Build GRCh38
rs547172123 Chr.6: 49517661-49517661 on Build GRCh38
rs918425862 Chr.6: 49517436-49517436 on Build GRCh38
rs538679778 Chr.6: 49517760-49517760 on Build GRCh38
rs950044046 Chr.6: 49517472-49517472 on Build GRCh38
rs181676142 Chr.6: 49517378-49517378 on Build GRCh38
rs983929451 Chr.6: 49517579-49517579 on Build GRCh38
rs1011046153 Chr.6: 49517486-49517486 on Build GRCh38
rs544088659 Chr.6: 49517430-49517430 on Build GRCh38
rs936969330 Chr.6: 49517521-49517521 on Build GRCh38
rs769987419 Chr.6: 49517572-49517572 on Build GRCh38
rs2501969 Chr.6: 49517671-49517671 on Build GRCh38
rs866848413 Chr.6: 49517538-49517538 on Build GRCh38
rs34020948 Chr.6: 49517532-49517533 on Build GRCh38
rs1011496131 Chr.6: 49517687-49517687 on Build GRCh38
rs151146571 Chr.6: 49517764-49517764 on Build GRCh38
rs755043562 Chr.6: 49517507-49517507 on Build GRCh38
rs1001266110 Chr.6: 49517710-49517710 on Build GRCh38
rs570894431 Chr.6: 49517547-49517547 on Build GRCh38
rs190724509 Chr.6: 49517611-49517611 on Build GRCh38
rs756401643 Chr.6: 49517548-49517548 on Build GRCh38
rs186838302 Chr.6: 49517514-49517514 on Build GRCh38
rs1016926030 Chr.6: 49517845-49517845 on Build GRCh38
rs1034002267 Chr.6: 49517546-49517546 on Build GRCh38
rs954063155 Chr.6: 49517617-49517617 on Build GRCh38
rs1007093947 Chr.6: 49517825-49517825 on Build GRCh38
rs371973497 Chr.6: 49517499-49517499 on Build GRCh38
rs750210750 Chr.6: 49517456-49517456 on Build GRCh38
rs536961591 Chr.6: 49517619-49517619 on Build GRCh38
rs763367262 Chr.6: 49517626-49517626 on Build GRCh38
rs942696507 Chr.6: 49517397-49517397 on Build GRCh38
rs571275293 Chr.6: 49517712-49517712 on Build GRCh38
rs888502873 Chr.6: 49517396-49517396 on Build GRCh38
rs979797535 Chr.6: 49517523-49517523 on Build GRCh38
rs892646871 Chr.6: 49517484-49517484 on Build GRCh38
rs9367358 Chr.6: 49517553-49517553 on Build GRCh38
rs368494673 Chr.6: 49517429-49517429 on Build GRCh38
rs1045639354 Chr.6: 49517489-49517489 on Build GRCh38
rs547949519 Chr.6: 49517500-49517500 on Build GRCh38
rs79034532 Chr.6: 49517551-49517551 on Build GRCh38
rs1042987446 Chr.6: 49517554-49517554 on Build GRCh38

View Full Product Details