Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49692221-49692744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISP2
Entrez Gene ID
7180
Gene Name
cysteine rich secretory protein 2
Gene Aliases
CRISP-2, CT36, GAPDL5, TPX1, TSP1
UniGene
Hs.2042
Gene Chromosome Location
Chr.6: 49676665-49714198 on Build GRCh38
Transcripts
NM_001142407.2 NM_001142408.2 NM_001142417.2 NM_001142435.2 NM_001261822.1 NM_003296.3 XM_005249349.1 XM_005249350.1 XM_005249351.3 XM_005249352.3 XM_005249353.3 XM_005249356.1 XM_011514841.1 XM_011514842.1 XM_011514843.2

SNP Detail

SNP ID Chromosome Location
rs558085936 Chr.6: 49692434-49692434 on Build GRCh38
rs980527724 Chr.6: 49692606-49692606 on Build GRCh38
rs998004932 Chr.6: 49692599-49692599 on Build GRCh38
rs113856808 Chr.6: 49692358-49692358 on Build GRCh38
rs962303513 Chr.6: 49692581-49692581 on Build GRCh38
rs989186107 Chr.6: 49692363-49692363 on Build GRCh38
rs189818348 Chr.6: 49692391-49692391 on Build GRCh38
rs1025250186 Chr.6: 49692590-49692590 on Build GRCh38
rs937545186 Chr.6: 49692406-49692406 on Build GRCh38
rs946230492 Chr.6: 49692433-49692435 on Build GRCh38
rs901909852 Chr.6: 49692437-49692437 on Build GRCh38
rs142795989 Chr.6: 49692588-49692588 on Build GRCh38
rs1011254004 Chr.6: 49692646-49692646 on Build GRCh38
rs552074407 Chr.6: 49692645-49692645 on Build GRCh38
rs536401902 Chr.6: 49692561-49692561 on Build GRCh38
rs1028922270 Chr.6: 49692508-49692508 on Build GRCh38
rs928080593 Chr.6: 49692422-49692422 on Build GRCh38
rs898104598 Chr.6: 49692463-49692463 on Build GRCh38
rs1007563898 Chr.6: 49692539-49692539 on Build GRCh38
rs754228213 Chr.6: 49692443-49692443 on Build GRCh38
rs960850006 Chr.6: 49692718-49692718 on Build GRCh38
rs976598376 Chr.6: 49692338-49692338 on Build GRCh38
rs751628883 Chr.6: 49692493-49692493 on Build GRCh38
rs374178574 Chr.6: 49692598-49692598 on Build GRCh38
rs1018033813 Chr.6: 49692286-49692286 on Build GRCh38
rs762436093 Chr.6: 49692612-49692612 on Build GRCh38
rs138757872 Chr.6: 49692533-49692533 on Build GRCh38
rs869264731 Chr.6: 49692264-49692265 on Build GRCh38
rs9369914 Chr.6: 49692592-49692592 on Build GRCh38
rs560449033 Chr.6: 49692448-49692448 on Build GRCh38
rs572006305 Chr.6: 49692462-49692462 on Build GRCh38
rs758402670 Chr.6: 49692326-49692326 on Build GRCh38
rs539358850 Chr.6: 49692489-49692489 on Build GRCh38
rs569440907 Chr.6: 49692547-49692547 on Build GRCh38
rs750275522 Chr.6: 49692308-49692308 on Build GRCh38
rs865899371 Chr.6: 49692622-49692622 on Build GRCh38
rs12624 Chr.6: 49692705-49692705 on Build GRCh38
rs1021930647 Chr.6: 49692647-49692647 on Build GRCh38
rs1016179701 Chr.6: 49692564-49692564 on Build GRCh38
rs1007616308 Chr.6: 49692249-49692249 on Build GRCh38
rs527329721 Chr.6: 49692410-49692410 on Build GRCh38
rs397723888 Chr.6: 49692263-49692264 on Build GRCh38
rs980413309 Chr.6: 49692323-49692323 on Build GRCh38
rs936322053 Chr.6: 49692613-49692613 on Build GRCh38
rs529822179 Chr.6: 49692357-49692357 on Build GRCh38
rs3839534 Chr.6: 49692260-49692261 on Build GRCh38
rs989587469 Chr.6: 49692663-49692663 on Build GRCh38
rs965190754 Chr.6: 49692320-49692320 on Build GRCh38
rs185890423 Chr.6: 49692367-49692367 on Build GRCh38
rs550967205 Chr.6: 49692506-49692506 on Build GRCh38
rs373065610 Chr.6: 49692716-49692716 on Build GRCh38
rs781434350 Chr.6: 49692557-49692557 on Build GRCh38
rs755110457 Chr.6: 49692507-49692507 on Build GRCh38
rs980527499 Chr.6: 49692419-49692419 on Build GRCh38
rs969047837 Chr.6: 49692669-49692669 on Build GRCh38

View Full Product Details