Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49711217-49711755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISP2
Entrez Gene ID
7180
Gene Name
cysteine rich secretory protein 2
Gene Aliases
CRISP-2, CT36, GAPDL5, TPX1, TSP1
UniGene
Hs.2042
Gene Chromosome Location
Chr.6: 49676665-49714198 on Build GRCh38
Transcripts
NM_001142407.2 NM_001142408.2 NM_001142417.2 NM_001142435.2 NM_001261822.1 NM_003296.3 XM_005249349.1 XM_005249350.1 XM_005249351.3 XM_005249352.3 XM_005249353.3 XM_005249356.1 XM_011514841.1 XM_011514842.1 XM_011514843.2 XM_011514844.1

SNP Detail

SNP ID Chromosome Location
rs754306851 Chr.6: 49711507-49711507 on Build GRCh38
rs1027138368 Chr.6: 49711387-49711387 on Build GRCh38
rs1050219336 Chr.6: 49711597-49711597 on Build GRCh38
rs896901362 Chr.6: 49711351-49711351 on Build GRCh38
rs774798009 Chr.6: 49711281-49711281 on Build GRCh38
rs888215309 Chr.6: 49711625-49711625 on Build GRCh38
rs888827267 Chr.6: 49711267-49711267 on Build GRCh38
rs183655470 Chr.6: 49711354-49711354 on Build GRCh38
rs980347305 Chr.6: 49711557-49711557 on Build GRCh38
rs904741489 Chr.6: 49711719-49711743 on Build GRCh38
rs76783602 Chr.6: 49711389-49711389 on Build GRCh38
rs896849768 Chr.6: 49711581-49711581 on Build GRCh38
rs932965519 Chr.6: 49711252-49711252 on Build GRCh38
rs1038618004 Chr.6: 49711290-49711290 on Build GRCh38
rs137947482 Chr.6: 49711554-49711554 on Build GRCh38
rs549019558 Chr.6: 49711577-49711577 on Build GRCh38
rs958770810 Chr.6: 49711503-49711503 on Build GRCh38
rs927200520 Chr.6: 49711565-49711565 on Build GRCh38
rs1057210998 Chr.6: 49711312-49711312 on Build GRCh38
rs773551554 Chr.6: 49711279-49711282 on Build GRCh38
rs910196100 Chr.6: 49711655-49711655 on Build GRCh38
rs1040283064 Chr.6: 49711654-49711654 on Build GRCh38
rs1044346604 Chr.6: 49711246-49711246 on Build GRCh38
rs938323123 Chr.6: 49711287-49711287 on Build GRCh38
rs536485718 Chr.6: 49711357-49711357 on Build GRCh38
rs764617207 Chr.6: 49711531-49711531 on Build GRCh38
rs189155925 Chr.6: 49711368-49711368 on Build GRCh38
rs772843562 Chr.6: 49711555-49711555 on Build GRCh38
rs1002836933 Chr.6: 49711484-49711484 on Build GRCh38
rs772667234 Chr.6: 49711377-49711377 on Build GRCh38
rs1034763610 Chr.6: 49711501-49711501 on Build GRCh38
rs1041462646 Chr.6: 49711243-49711243 on Build GRCh38
rs901415711 Chr.6: 49711724-49711724 on Build GRCh38
rs1019759959 Chr.6: 49711498-49711498 on Build GRCh38
rs905796620 Chr.6: 49711270-49711270 on Build GRCh38
rs762578528 Chr.6: 49711618-49711618 on Build GRCh38
rs766036258 Chr.6: 49711706-49711706 on Build GRCh38
rs938633392 Chr.6: 49711578-49711578 on Build GRCh38
rs1018504420 Chr.6: 49711432-49711432 on Build GRCh38
rs1007019088 Chr.6: 49711644-49711644 on Build GRCh38
rs956782917 Chr.6: 49711538-49711538 on Build GRCh38
rs1050514590 Chr.6: 49711263-49711263 on Build GRCh38
rs929698850 Chr.6: 49711605-49711605 on Build GRCh38
rs966016291 Chr.6: 49711454-49711454 on Build GRCh38
rs898649417 Chr.6: 49711345-49711345 on Build GRCh38
rs182212249 Chr.6: 49711580-49711580 on Build GRCh38
rs528872003 Chr.6: 49711395-49711395 on Build GRCh38
rs374690674 Chr.6: 49711729-49711729 on Build GRCh38
rs989545393 Chr.6: 49711544-49711544 on Build GRCh38

View Full Product Details