Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49733611-49734111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISP3
Entrez Gene ID
10321
Gene Name
cysteine rich secretory protein 3
Gene Aliases
Aeg2, CRISP-3, CRS3, SGP28, dJ442L6.3
UniGene
Hs.404466
Gene Chromosome Location
Chr.6: 49727376-49744455 on Build GRCh38
Transcripts
NM_001190986.1 NM_006061.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7233272 c.426G>C p.E142D Chr.6: 49733769-49733769 on Build GRCh38
5935102 c.295A>G p.N99D Chr.6: 49733831-49733831 on Build GRCh38
5935101 c.364A>G p.N122D Chr.6: 49733831-49733831 on Build GRCh38
4674720 c.478G>A p.G160R Chr.6: 49733717-49733717 on Build GRCh38
7233273 c.357G>C p.E119D Chr.6: 49733769-49733769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779170235 Chr.6: 49733876-49733876 on Build GRCh38
rs750037143 Chr.6: 49733692-49733692 on Build GRCh38
rs138539923 Chr.6: 49733816-49733816 on Build GRCh38
rs186676018 Chr.6: 49733728-49733728 on Build GRCh38
rs756766734 Chr.6: 49733911-49733912 on Build GRCh38
rs552959054 Chr.6: 49733672-49733672 on Build GRCh38
rs115036483 Chr.6: 49733864-49733864 on Build GRCh38
rs372524501 Chr.6: 49733662-49733662 on Build GRCh38
rs779983581 Chr.6: 49733840-49733840 on Build GRCh38
rs762689400 Chr.6: 49733826-49733826 on Build GRCh38
rs753593361 Chr.6: 49733670-49733670 on Build GRCh38
rs1022271471 Chr.6: 49733912-49733913 on Build GRCh38
rs772417674 Chr.6: 49733814-49733814 on Build GRCh38
rs61735632 Chr.6: 49733756-49733756 on Build GRCh38
rs754926235 Chr.6: 49733711-49733711 on Build GRCh38
rs769384663 Chr.6: 49733822-49733822 on Build GRCh38
rs534403100 Chr.6: 49733661-49733661 on Build GRCh38
rs1037959888 Chr.6: 49733801-49733801 on Build GRCh38
rs776121414 Chr.6: 49733739-49733739 on Build GRCh38
rs544050502 Chr.6: 49733734-49733734 on Build GRCh38
rs534759956 Chr.6: 49733881-49733881 on Build GRCh38
rs533184595 Chr.6: 49733773-49733773 on Build GRCh38
rs199808242 Chr.6: 49733824-49733824 on Build GRCh38
rs765889916 Chr.6: 49733834-49733834 on Build GRCh38
rs550597593 Chr.6: 49733651-49733651 on Build GRCh38
rs149267211 Chr.6: 49733839-49733839 on Build GRCh38
rs780364431 Chr.6: 49733896-49733896 on Build GRCh38
rs1008077421 Chr.6: 49734006-49734006 on Build GRCh38
rs779258242 Chr.6: 49733803-49733803 on Build GRCh38
rs758123656 Chr.6: 49733810-49733810 on Build GRCh38
rs367914160 Chr.6: 49733860-49733860 on Build GRCh38
rs749567736 Chr.6: 49733791-49733791 on Build GRCh38
rs758268585 Chr.6: 49733701-49733701 on Build GRCh38
rs73441834 Chr.6: 49733865-49733865 on Build GRCh38
rs978010771 Chr.6: 49733720-49733720 on Build GRCh38
rs751350628 Chr.6: 49733767-49733767 on Build GRCh38
rs192075352 Chr.6: 49733926-49733926 on Build GRCh38
rs184193221 Chr.6: 49733681-49733681 on Build GRCh38
rs985341465 Chr.6: 49733677-49733677 on Build GRCh38
rs757501536 Chr.6: 49733870-49733870 on Build GRCh38
rs749461707 Chr.6: 49733727-49733727 on Build GRCh38
rs748181777 Chr.6: 49733721-49733721 on Build GRCh38
rs947818445 Chr.6: 49733765-49733765 on Build GRCh38
rs138179646 Chr.6: 49733705-49733705 on Build GRCh38
rs368363779 Chr.6: 49733868-49733868 on Build GRCh38
rs138993320 Chr.6: 49733777-49733777 on Build GRCh38
rs976690564 Chr.6: 49734036-49734036 on Build GRCh38
rs752734188 Chr.6: 49733856-49733856 on Build GRCh38
rs772472075 Chr.6: 49733891-49733891 on Build GRCh38
rs142187249 Chr.6: 49733788-49733788 on Build GRCh38
rs978128556 Chr.6: 49733946-49733946 on Build GRCh38
rs781500082 Chr.6: 49734034-49734034 on Build GRCh38
rs556967536 Chr.6: 49734000-49734000 on Build GRCh38
rs1864312 Chr.6: 49733726-49733726 on Build GRCh38
rs761063266 Chr.6: 49733820-49733820 on Build GRCh38
rs781444205 Chr.6: 49733716-49733716 on Build GRCh38
rs977078330 Chr.6: 49733717-49733717 on Build GRCh38
rs552780455 Chr.6: 49733920-49733920 on Build GRCh38
rs138786566 Chr.6: 49734025-49734025 on Build GRCh38
rs559738360 Chr.6: 49733764-49733764 on Build GRCh38
rs764874496 Chr.6: 49733687-49733687 on Build GRCh38
rs495335 Chr.6: 49733810-49733810 on Build GRCh38
rs952505944 Chr.6: 49733653-49733653 on Build GRCh38
rs942461836 Chr.6: 49733798-49733798 on Build GRCh38
rs747800445 Chr.6: 49734075-49734075 on Build GRCh38
rs746981995 Chr.6: 49733709-49733709 on Build GRCh38
rs968034904 Chr.6: 49733925-49733925 on Build GRCh38
rs1027781354 Chr.6: 49733637-49733637 on Build GRCh38
rs771022504 Chr.6: 49733792-49733792 on Build GRCh38
rs868076293 Chr.6: 49733762-49733762 on Build GRCh38
rs956358508 Chr.6: 49734074-49734074 on Build GRCh38
rs551317904 Chr.6: 49733780-49733780 on Build GRCh38
rs772506834 Chr.6: 49733736-49733736 on Build GRCh38
rs529418068 Chr.6: 49733748-49733748 on Build GRCh38
rs568259097 Chr.6: 49733812-49733812 on Build GRCh38
rs756277555 Chr.6: 49733787-49733787 on Build GRCh38
rs75675396 Chr.6: 49733743-49733743 on Build GRCh38
rs73441836 Chr.6: 49733972-49733972 on Build GRCh38
rs911047418 Chr.6: 49734048-49734048 on Build GRCh38
rs373635344 Chr.6: 49733832-49733832 on Build GRCh38
rs190904139 Chr.6: 49733658-49733658 on Build GRCh38
rs779938363 Chr.6: 49733851-49733851 on Build GRCh38
rs923880743 Chr.6: 49734056-49734056 on Build GRCh38

View Full Product Details