Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49857025-49857543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISP1
Entrez Gene ID
167
Gene Name
cysteine rich secretory protein 1
Gene Aliases
AEGL1, ARP, CRISP-1, HEL-S-57, HSCRISP1D, HSCRISP1G, HUMARP
UniGene
Hs.109620
Gene Chromosome Location
Chr.6: 49834257-49877096 on Build GRCh38
Transcripts
NM_001131.2 NM_001205220.1 NM_170609.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1445022 c.37G>A p.A13T Chr.6: 49857364-49857364 on Build GRCh38
7362690 c.49C>T p.P17S Chr.6: 49857352-49857352 on Build GRCh38
387666 c.19T>A p.L7M Chr.6: 49857382-49857382 on Build GRCh38
3874648 c.61A>G p.M21V Chr.6: 49857340-49857340 on Build GRCh38
6514024 c.66A>T p.K22N Chr.6: 49857335-49857335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570181147 Chr.6: 49857165-49857165 on Build GRCh38
rs576090122 Chr.6: 49857156-49857156 on Build GRCh38
rs762440567 Chr.6: 49857287-49857287 on Build GRCh38
rs937338359 Chr.6: 49857390-49857390 on Build GRCh38
rs149138271 Chr.6: 49857400-49857400 on Build GRCh38
rs566222962 Chr.6: 49857263-49857263 on Build GRCh38
rs539092809 Chr.6: 49857430-49857430 on Build GRCh38
rs921501046 Chr.6: 49857087-49857087 on Build GRCh38
rs762032785 Chr.6: 49857415-49857415 on Build GRCh38
rs758721480 Chr.6: 49857340-49857340 on Build GRCh38
rs1001518404 Chr.6: 49857070-49857070 on Build GRCh38
rs775294325 Chr.6: 49857386-49857386 on Build GRCh38
rs921553980 Chr.6: 49857493-49857493 on Build GRCh38
rs549590933 Chr.6: 49857199-49857199 on Build GRCh38
rs757406391 Chr.6: 49857331-49857331 on Build GRCh38
rs112657420 Chr.6: 49857308-49857308 on Build GRCh38
rs748840852 Chr.6: 49857367-49857367 on Build GRCh38
rs143467881 Chr.6: 49857058-49857058 on Build GRCh38
rs146555761 Chr.6: 49857068-49857068 on Build GRCh38
rs371226413 Chr.6: 49857081-49857081 on Build GRCh38
rs1009616779 Chr.6: 49857149-49857149 on Build GRCh38
rs868623163 Chr.6: 49857402-49857403 on Build GRCh38
rs199731457 Chr.6: 49857309-49857309 on Build GRCh38
rs759302181 Chr.6: 49857303-49857303 on Build GRCh38
rs769212429 Chr.6: 49857346-49857346 on Build GRCh38
rs188741658 Chr.6: 49857488-49857488 on Build GRCh38
rs148067663 Chr.6: 49857358-49857358 on Build GRCh38
rs772652189 Chr.6: 49857286-49857286 on Build GRCh38
rs764278973 Chr.6: 49857323-49857323 on Build GRCh38
rs368103719 Chr.6: 49857074-49857074 on Build GRCh38
rs771766264 Chr.6: 49857379-49857379 on Build GRCh38
rs184727202 Chr.6: 49857302-49857302 on Build GRCh38
rs753795933 Chr.6: 49857403-49857403 on Build GRCh38
rs766089358 Chr.6: 49857292-49857292 on Build GRCh38
rs1028777525 Chr.6: 49857228-49857228 on Build GRCh38
rs1051096089 Chr.6: 49857123-49857123 on Build GRCh38
rs755752840 Chr.6: 49857334-49857334 on Build GRCh38
rs759184426 Chr.6: 49857378-49857378 on Build GRCh38
rs971161600 Chr.6: 49857048-49857048 on Build GRCh38
rs1023970420 Chr.6: 49857365-49857365 on Build GRCh38
rs533425556 Chr.6: 49857146-49857146 on Build GRCh38
rs372523054 Chr.6: 49857312-49857312 on Build GRCh38
rs917014819 Chr.6: 49857047-49857047 on Build GRCh38
rs932686369 Chr.6: 49857105-49857105 on Build GRCh38
rs771034381 Chr.6: 49857458-49857458 on Build GRCh38
rs773221352 Chr.6: 49857361-49857361 on Build GRCh38
rs1020080160 Chr.6: 49857153-49857153 on Build GRCh38
rs953019740 Chr.6: 49857272-49857272 on Build GRCh38
rs975813767 Chr.6: 49857213-49857213 on Build GRCh38
rs3204891 Chr.6: 49857337-49857337 on Build GRCh38
rs758590335 Chr.6: 49857439-49857439 on Build GRCh38
rs1033332458 Chr.6: 49857071-49857071 on Build GRCh38
rs765320798 Chr.6: 49857429-49857429 on Build GRCh38
rs1015902959 Chr.6: 49857515-49857515 on Build GRCh38
rs1028744599 Chr.6: 49857504-49857504 on Build GRCh38
rs780127446 Chr.6: 49857445-49857445 on Build GRCh38
rs764192737 Chr.6: 49857402-49857402 on Build GRCh38
rs1045779797 Chr.6: 49857375-49857375 on Build GRCh38
rs191321179 Chr.6: 49857511-49857511 on Build GRCh38
rs143443747 Chr.6: 49857338-49857338 on Build GRCh38
rs779043634 Chr.6: 49857335-49857335 on Build GRCh38
rs983393247 Chr.6: 49857052-49857052 on Build GRCh38
rs760620118 Chr.6: 49857317-49857317 on Build GRCh38
rs754028118 Chr.6: 49857324-49857324 on Build GRCh38
rs561817950 Chr.6: 49857056-49857056 on Build GRCh38
rs780400857 Chr.6: 49857341-49857341 on Build GRCh38
rs374875765 Chr.6: 49857285-49857285 on Build GRCh38
rs1000133029 Chr.6: 49857212-49857212 on Build GRCh38
rs941623023 Chr.6: 49857064-49857064 on Build GRCh38
rs770408802 Chr.6: 49857373-49857373 on Build GRCh38
rs751989422 Chr.6: 49857448-49857448 on Build GRCh38
rs953069056 Chr.6: 49857508-49857508 on Build GRCh38
rs997386388 Chr.6: 49857501-49857501 on Build GRCh38
rs192912585 Chr.6: 49857176-49857176 on Build GRCh38
rs267601066 Chr.6: 49857398-49857398 on Build GRCh38
rs973409449 Chr.6: 49857349-49857349 on Build GRCh38
rs779752626 Chr.6: 49857403-49857404 on Build GRCh38

View Full Product Details