Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49963723-49964263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB114
Entrez Gene ID
245928
Gene Name
defensin beta 114
Gene Aliases
DEFB-14, DEFB14
UniGene
Hs.381372
Gene Chromosome Location
Chr.6: 49960292-49964105 on Build GRCh38
Transcripts
NM_001037499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5466878 c.9C>A p.I3I Chr.6: 49964097-49964097 on Build GRCh38
7308197 c.36T>C p.Y12Y Chr.6: 49964070-49964070 on Build GRCh38
3995037 c.49C>T p.L17L Chr.6: 49964057-49964057 on Build GRCh38
6471277 c.42C>T p.T14T Chr.6: 49964064-49964064 on Build GRCh38
3354172 c.15C>A p.Y5* Chr.6: 49964091-49964091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375731886 Chr.6: 49964152-49964152 on Build GRCh38
rs771412617 Chr.6: 49964150-49964150 on Build GRCh38
rs773884354 Chr.6: 49964063-49964063 on Build GRCh38
rs566064050 Chr.6: 49963793-49963793 on Build GRCh38
rs761389572 Chr.6: 49964157-49964157 on Build GRCh38
rs778869229 Chr.6: 49964028-49964028 on Build GRCh38
rs927613552 Chr.6: 49964179-49964179 on Build GRCh38
rs1053704863 Chr.6: 49963857-49963857 on Build GRCh38
rs199591078 Chr.6: 49964105-49964105 on Build GRCh38
rs768828999 Chr.6: 49964050-49964050 on Build GRCh38
rs891161909 Chr.6: 49964189-49964189 on Build GRCh38
rs770283612 Chr.6: 49964136-49964136 on Build GRCh38
rs774924564 Chr.6: 49964069-49964069 on Build GRCh38
rs939143917 Chr.6: 49964201-49964201 on Build GRCh38
rs895336213 Chr.6: 49963818-49963818 on Build GRCh38
rs534750447 Chr.6: 49963856-49963856 on Build GRCh38
rs759690511 Chr.6: 49964133-49964134 on Build GRCh38
rs571656149 Chr.6: 49963884-49963884 on Build GRCh38
rs966506571 Chr.6: 49963819-49963819 on Build GRCh38
rs773649526 Chr.6: 49964138-49964138 on Build GRCh38
rs181557019 Chr.6: 49964216-49964216 on Build GRCh38
rs750366094 Chr.6: 49964102-49964102 on Build GRCh38
rs986506225 Chr.6: 49963754-49963754 on Build GRCh38
rs772168815 Chr.6: 49964037-49964037 on Build GRCh38
rs1052514619 Chr.6: 49964180-49964180 on Build GRCh38
rs563452444 Chr.6: 49964209-49964209 on Build GRCh38
rs533779730 Chr.6: 49964145-49964145 on Build GRCh38
rs184825813 Chr.6: 49964021-49964021 on Build GRCh38
rs375512510 Chr.6: 49963875-49963875 on Build GRCh38
rs557113803 Chr.6: 49963957-49963957 on Build GRCh38
rs753469860 Chr.6: 49964080-49964080 on Build GRCh38
rs537506891 Chr.6: 49963888-49963888 on Build GRCh38
rs982456386 Chr.6: 49964026-49964026 on Build GRCh38
rs748570141 Chr.6: 49964056-49964056 on Build GRCh38
rs955357014 Chr.6: 49963892-49963892 on Build GRCh38
rs776844812 Chr.6: 49964101-49964102 on Build GRCh38
rs752402253 Chr.6: 49964001-49964001 on Build GRCh38
rs994927777 Chr.6: 49963952-49963952 on Build GRCh38
rs549037101 Chr.6: 49963771-49963771 on Build GRCh38
rs928269221 Chr.6: 49963784-49963784 on Build GRCh38
rs757068824 Chr.6: 49964089-49964089 on Build GRCh38
rs748337284 Chr.6: 49964129-49964129 on Build GRCh38
rs939091504 Chr.6: 49963910-49963910 on Build GRCh38
rs773585958 Chr.6: 49963941-49963941 on Build GRCh38
rs776298457 Chr.6: 49964155-49964155 on Build GRCh38
rs763483599 Chr.6: 49964067-49964067 on Build GRCh38
rs763612924 Chr.6: 49964153-49964153 on Build GRCh38
rs763696432 Chr.6: 49964074-49964074 on Build GRCh38
rs557420975 Chr.6: 49963906-49963906 on Build GRCh38
rs1056713410 Chr.6: 49963812-49963812 on Build GRCh38
rs781290339 Chr.6: 49964128-49964128 on Build GRCh38
rs776883423 Chr.6: 49964051-49964051 on Build GRCh38
rs201539390 Chr.6: 49964003-49964003 on Build GRCh38
rs1027871999 Chr.6: 49963954-49963954 on Build GRCh38
rs112237322 Chr.6: 49964032-49964032 on Build GRCh38
rs201002944 Chr.6: 49964016-49964016 on Build GRCh38
rs923760736 Chr.6: 49963832-49963832 on Build GRCh38
rs999578472 Chr.6: 49963872-49963872 on Build GRCh38
rs976608845 Chr.6: 49963826-49963826 on Build GRCh38
rs770232340 Chr.6: 49964058-49964058 on Build GRCh38
rs764978252 Chr.6: 49964090-49964090 on Build GRCh38
rs753640533 Chr.6: 49964017-49964017 on Build GRCh38
rs755061804 Chr.6: 49964126-49964126 on Build GRCh38
rs897546238 Chr.6: 49964210-49964210 on Build GRCh38
rs765320655 Chr.6: 49964134-49964135 on Build GRCh38
rs1049674278 Chr.6: 49964226-49964226 on Build GRCh38
rs1008210918 Chr.6: 49964199-49964199 on Build GRCh38
rs758274794 Chr.6: 49964104-49964104 on Build GRCh38
rs767063995 Chr.6: 49964068-49964068 on Build GRCh38
rs1007579820 Chr.6: 49964025-49964025 on Build GRCh38
rs1044220033 Chr.6: 49963829-49963829 on Build GRCh38
rs139140570 Chr.6: 49964122-49964122 on Build GRCh38
rs747099295 Chr.6: 49964047-49964047 on Build GRCh38
rs749661598 Chr.6: 49964141-49964141 on Build GRCh38
rs903953636 Chr.6: 49963844-49963844 on Build GRCh38
rs1036524875 Chr.6: 49963927-49963927 on Build GRCh38
rs189596822 Chr.6: 49964109-49964109 on Build GRCh38
rs889401341 Chr.6: 49964232-49964232 on Build GRCh38
rs1045101882 Chr.6: 49963901-49963901 on Build GRCh38
rs371971434 Chr.6: 49964218-49964218 on Build GRCh38
rs779930378 Chr.6: 49964041-49964041 on Build GRCh38
rs939607215 Chr.6: 49963811-49963811 on Build GRCh38
rs573921159 Chr.6: 49963986-49963986 on Build GRCh38
rs73737276 Chr.6: 49964070-49964070 on Build GRCh38
rs911996274 Chr.6: 49963804-49963804 on Build GRCh38
rs935273723 Chr.6: 49963836-49963836 on Build GRCh38

View Full Product Details