Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53128532-53129027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCM1
Entrez Gene ID
8521
Gene Name
glial cells missing homolog 1
Gene Aliases
GCMA, hGCMa
UniGene
Hs.23589
Gene Chromosome Location
Chr.6: 53125644-53262803 on Build GRCh38
Transcripts
NM_003643.3 XM_017011390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3411200 c.735A>G p.G245G Chr.6: 53128782-53128782 on Build GRCh38
6107423 c.648T>A p.G216G Chr.6: 53128869-53128869 on Build GRCh38
4686647 c.623_624insT p.Q209fs*9 Chr.6: 53128893-53128894 on Build GRCh38
1080314 c.925G>T p.A309S Chr.6: 53128592-53128592 on Build GRCh38
6031286 c.866C>A p.S289Y Chr.6: 53128651-53128651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112673732 Chr.6: 53128787-53128787 on Build GRCh38
rs748358524 Chr.6: 53128695-53128695 on Build GRCh38
rs749561815 Chr.6: 53128707-53128707 on Build GRCh38
rs766178059 Chr.6: 53128656-53128656 on Build GRCh38
rs772628092 Chr.6: 53128881-53128881 on Build GRCh38
rs763875998 Chr.6: 53128629-53128629 on Build GRCh38
rs903615600 Chr.6: 53128972-53128972 on Build GRCh38
rs35934676 Chr.6: 53128811-53128811 on Build GRCh38
rs150106558 Chr.6: 53128838-53128838 on Build GRCh38
rs770538680 Chr.6: 53128977-53128977 on Build GRCh38
rs143634385 Chr.6: 53128943-53128943 on Build GRCh38
rs747852377 Chr.6: 53128862-53128862 on Build GRCh38
rs753340260 Chr.6: 53128878-53128878 on Build GRCh38
rs1023399945 Chr.6: 53128689-53128689 on Build GRCh38
rs781507485 Chr.6: 53128884-53128884 on Build GRCh38
rs762616288 Chr.6: 53128771-53128771 on Build GRCh38
rs779528606 Chr.6: 53128846-53128846 on Build GRCh38
rs752709375 Chr.6: 53128826-53128826 on Build GRCh38
rs769317078 Chr.6: 53128869-53128869 on Build GRCh38
rs146630061 Chr.6: 53128734-53128734 on Build GRCh38
rs773767712 Chr.6: 53128594-53128594 on Build GRCh38
rs3799267 Chr.6: 53128722-53128722 on Build GRCh38
rs780606225 Chr.6: 53128955-53128955 on Build GRCh38
rs1000959716 Chr.6: 53128718-53128718 on Build GRCh38
rs376111326 Chr.6: 53128597-53128597 on Build GRCh38
rs764189267 Chr.6: 53128918-53128918 on Build GRCh38
rs771495205 Chr.6: 53128710-53128710 on Build GRCh38
rs145102927 Chr.6: 53128806-53128806 on Build GRCh38
rs761560575 Chr.6: 53128742-53128742 on Build GRCh38
rs760060174 Chr.6: 53128612-53128612 on Build GRCh38
rs3823112 Chr.6: 53128827-53128827 on Build GRCh38
rs945081289 Chr.6: 53128989-53128989 on Build GRCh38
rs778207915 Chr.6: 53128700-53128700 on Build GRCh38
rs369083207 Chr.6: 53128970-53128970 on Build GRCh38
rs367761617 Chr.6: 53128720-53128720 on Build GRCh38
rs761544740 Chr.6: 53128751-53128781 on Build GRCh38
rs945600001 Chr.6: 53128688-53128688 on Build GRCh38
rs190371776 Chr.6: 53128662-53128662 on Build GRCh38
rs759319118 Chr.6: 53128885-53128885 on Build GRCh38
rs961931855 Chr.6: 53128552-53128552 on Build GRCh38
rs768265831 Chr.6: 53128729-53128729 on Build GRCh38
rs1044899354 Chr.6: 53128971-53128971 on Build GRCh38
rs3799268 Chr.6: 53128685-53128685 on Build GRCh38
rs762898529 Chr.6: 53128883-53128883 on Build GRCh38
rs746241994 Chr.6: 53128856-53128856 on Build GRCh38
rs372973340 Chr.6: 53128962-53128962 on Build GRCh38
rs758571009 Chr.6: 53128598-53128598 on Build GRCh38
rs761626331 Chr.6: 53128985-53128985 on Build GRCh38
rs758368502 Chr.6: 53128641-53128641 on Build GRCh38
rs776398452 Chr.6: 53128630-53128630 on Build GRCh38
rs775083311 Chr.6: 53128601-53128601 on Build GRCh38
rs148920545 Chr.6: 53128626-53128626 on Build GRCh38
rs767330728 Chr.6: 53128949-53128953 on Build GRCh38
rs182942996 Chr.6: 53128940-53128940 on Build GRCh38
rs774283897 Chr.6: 53128668-53128670 on Build GRCh38
rs373759594 Chr.6: 53128690-53128690 on Build GRCh38
rs753881228 Chr.6: 53128920-53128920 on Build GRCh38
rs966216322 Chr.6: 53128769-53128769 on Build GRCh38
rs773081660 Chr.6: 53128767-53128767 on Build GRCh38
rs1005542010 Chr.6: 53128823-53128823 on Build GRCh38
rs767355404 Chr.6: 53128889-53128889 on Build GRCh38
rs1012081640 Chr.6: 53128671-53128671 on Build GRCh38
rs140778484 Chr.6: 53128831-53128831 on Build GRCh38
rs772634447 Chr.6: 53128858-53128858 on Build GRCh38
rs780980247 Chr.6: 53128861-53128861 on Build GRCh38
rs750116594 Chr.6: 53128640-53128640 on Build GRCh38
rs752890981 Chr.6: 53128911-53128911 on Build GRCh38
rs760923364 Chr.6: 53128915-53128915 on Build GRCh38
rs888470872 Chr.6: 53128821-53128821 on Build GRCh38
rs200430389 Chr.6: 53128998-53128998 on Build GRCh38
rs374967752 Chr.6: 53128750-53128750 on Build GRCh38
rs141372786 Chr.6: 53128802-53128802 on Build GRCh38
rs767582312 Chr.6: 53128816-53128816 on Build GRCh38
rs756540220 Chr.6: 53128697-53128697 on Build GRCh38
rs748552826 Chr.6: 53128589-53128589 on Build GRCh38
rs930646909 Chr.6: 53128617-53128617 on Build GRCh38
rs138391063 Chr.6: 53128932-53128932 on Build GRCh38
rs374961417 Chr.6: 53128969-53128969 on Build GRCh38
rs745851219 Chr.6: 53128986-53128986 on Build GRCh38
rs371602759 Chr.6: 53128721-53128721 on Build GRCh38
rs368633414 Chr.6: 53128631-53128631 on Build GRCh38
rs779285520 Chr.6: 53128939-53128939 on Build GRCh38
rs769512344 Chr.6: 53128743-53128743 on Build GRCh38
rs757536089 Chr.6: 53128844-53128844 on Build GRCh38
rs972299340 Chr.6: 53128576-53128576 on Build GRCh38
rs770122746 Chr.6: 53128590-53128590 on Build GRCh38
rs748751354 Chr.6: 53128976-53128976 on Build GRCh38
rs980415104 Chr.6: 53128628-53128628 on Build GRCh38
rs781443217 Chr.6: 53128693-53128693 on Build GRCh38
rs577227157 Chr.6: 53128749-53128749 on Build GRCh38
rs976960152 Chr.6: 53128999-53128999 on Build GRCh38

View Full Product Details