Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53145478-53145978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCM1
Entrez Gene ID
8521
Gene Name
glial cells missing homolog 1
Gene Aliases
GCMA, hGCMa
UniGene
Hs.23589
Gene Chromosome Location
Chr.6: 53125644-53262803 on Build GRCh38
Transcripts
NM_003643.3 XM_017011390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4553404 c.58G>A p.D20N Chr.6: 53145575-53145575 on Build GRCh38
1471984 c.57T>C p.N19N Chr.6: 53145576-53145576 on Build GRCh38
3921972 c.63G>A p.V21V Chr.6: 53145570-53145570 on Build GRCh38
6670029 c.12C>T p.D4D Chr.6: 53145621-53145621 on Build GRCh38
5976060 c.60T>C p.D20D Chr.6: 53145573-53145573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372689809 Chr.6: 53145785-53145785 on Build GRCh38
rs45476600 Chr.6: 53145665-53145665 on Build GRCh38
rs368234030 Chr.6: 53145512-53145512 on Build GRCh38
rs867043933 Chr.6: 53145573-53145573 on Build GRCh38
rs868470250 Chr.6: 53145742-53145742 on Build GRCh38
rs541918829 Chr.6: 53145950-53145950 on Build GRCh38
rs1056864230 Chr.6: 53145813-53145813 on Build GRCh38
rs533348953 Chr.6: 53145707-53145707 on Build GRCh38
rs1129070 Chr.6: 53145616-53145616 on Build GRCh38
rs966654946 Chr.6: 53145653-53145653 on Build GRCh38
rs371574245 Chr.6: 53145658-53145658 on Build GRCh38
rs1022338494 Chr.6: 53145866-53145866 on Build GRCh38
rs779224988 Chr.6: 53145621-53145621 on Build GRCh38
rs966666124 Chr.6: 53145882-53145882 on Build GRCh38
rs756598779 Chr.6: 53145566-53145566 on Build GRCh38
rs376763700 Chr.6: 53145692-53145692 on Build GRCh38
rs1032100480 Chr.6: 53145886-53145886 on Build GRCh38
rs185103416 Chr.6: 53145517-53145517 on Build GRCh38
rs1008937685 Chr.6: 53145905-53145905 on Build GRCh38
rs867536401 Chr.6: 53145690-53145690 on Build GRCh38
rs766469178 Chr.6: 53145537-53145538 on Build GRCh38
rs768506971 Chr.6: 53145685-53145685 on Build GRCh38
rs933772413 Chr.6: 53145714-53145714 on Build GRCh38
rs552244358 Chr.6: 53145503-53145503 on Build GRCh38
rs543564896 Chr.6: 53145717-53145717 on Build GRCh38
rs201962661 Chr.6: 53145643-53145643 on Build GRCh38
rs1035873729 Chr.6: 53145524-53145524 on Build GRCh38
rs1038061002 Chr.6: 53145727-53145727 on Build GRCh38
rs773268519 Chr.6: 53145654-53145654 on Build GRCh38
rs781288161 Chr.6: 53145547-53145547 on Build GRCh38
rs549380902 Chr.6: 53145793-53145793 on Build GRCh38
rs955395135 Chr.6: 53145911-53145911 on Build GRCh38
rs886748620 Chr.6: 53145799-53145799 on Build GRCh38
rs372405902 Chr.6: 53145726-53145726 on Build GRCh38
rs778138761 Chr.6: 53145595-53145595 on Build GRCh38
rs962529920 Chr.6: 53145952-53145952 on Build GRCh38
rs149679322 Chr.6: 53145949-53145949 on Build GRCh38
rs368938078 Chr.6: 53145527-53145527 on Build GRCh38
rs1000426337 Chr.6: 53145873-53145873 on Build GRCh38
rs59049517 Chr.6: 53145812-53145812 on Build GRCh38
rs35646561 Chr.6: 53145620-53145620 on Build GRCh38
rs1013178580 Chr.6: 53145533-53145533 on Build GRCh38
rs1047935034 Chr.6: 53145811-53145812 on Build GRCh38
rs1011391480 Chr.6: 53145829-53145829 on Build GRCh38
rs547704889 Chr.6: 53145821-53145821 on Build GRCh38
rs200131014 Chr.6: 53145598-53145598 on Build GRCh38
rs766210321 Chr.6: 53145667-53145667 on Build GRCh38
rs761753901 Chr.6: 53145626-53145626 on Build GRCh38
rs9370179 Chr.6: 53145902-53145902 on Build GRCh38
rs931019906 Chr.6: 53145769-53145769 on Build GRCh38
rs35697624 Chr.6: 53145860-53145861 on Build GRCh38
rs779834876 Chr.6: 53145602-53145602 on Build GRCh38
rs774524908 Chr.6: 53145672-53145672 on Build GRCh38
rs759783077 Chr.6: 53145677-53145677 on Build GRCh38
rs112412851 Chr.6: 53145812-53145812 on Build GRCh38
rs985630058 Chr.6: 53145719-53145719 on Build GRCh38
rs374427292 Chr.6: 53145561-53145561 on Build GRCh38
rs904754958 Chr.6: 53145830-53145830 on Build GRCh38
rs796312941 Chr.6: 53145820-53145820 on Build GRCh38
rs572717204 Chr.6: 53145618-53145618 on Build GRCh38
rs771485379 Chr.6: 53145599-53145599 on Build GRCh38
rs757504303 Chr.6: 53145528-53145528 on Build GRCh38
rs541816809 Chr.6: 53145544-53145544 on Build GRCh38
rs751835986 Chr.6: 53145542-53145542 on Build GRCh38
rs527617768 Chr.6: 53145815-53145815 on Build GRCh38
rs921169403 Chr.6: 53145764-53145764 on Build GRCh38
rs1023244204 Chr.6: 53145827-53145827 on Build GRCh38
rs895261564 Chr.6: 53145824-53145824 on Build GRCh38
rs60944691 Chr.6: 53145920-53145920 on Build GRCh38
rs911026691 Chr.6: 53145939-53145939 on Build GRCh38
rs879057770 Chr.6: 53145924-53145924 on Build GRCh38
rs776030675 Chr.6: 53145623-53145623 on Build GRCh38
rs758194249 Chr.6: 53145900-53145902 on Build GRCh38

View Full Product Details