Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 54870062-54870577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83B
Entrez Gene ID
222584
Gene Name
family with sequence similarity 83 member B
Gene Aliases
C6orf143
UniGene
Hs.152423
Gene Chromosome Location
Chr.6: 54846323-54945099 on Build GRCh38
Transcripts
NM_001010872.2 XM_006715022.3 XM_011514394.2 XM_011514395.2 XM_017010478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6107392 c.153A>T p.R51R Chr.6: 54870399-54870399 on Build GRCh38
6514062 c.23C>A p.S8Y Chr.6: 54870269-54870269 on Build GRCh38
3628648 c.195G>A p.L65L Chr.6: 54870441-54870441 on Build GRCh38
4004082 c.78A>G p.E26E Chr.6: 54870324-54870324 on Build GRCh38
6107394 c.109G>A p.E37K Chr.6: 54870355-54870355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765150070 Chr.6: 54870398-54870398 on Build GRCh38
rs867436056 Chr.6: 54870490-54870491 on Build GRCh38
rs555330987 Chr.6: 54870420-54870420 on Build GRCh38
rs774936889 Chr.6: 54870379-54870379 on Build GRCh38
rs760403012 Chr.6: 54870381-54870381 on Build GRCh38
rs750389951 Chr.6: 54870402-54870402 on Build GRCh38
rs368238775 Chr.6: 54870338-54870338 on Build GRCh38
rs769300499 Chr.6: 54870541-54870541 on Build GRCh38
rs771157243 Chr.6: 54870472-54870472 on Build GRCh38
rs764003806 Chr.6: 54870227-54870227 on Build GRCh38
rs377607948 Chr.6: 54870370-54870370 on Build GRCh38
rs778707712 Chr.6: 54870238-54870238 on Build GRCh38
rs554307217 Chr.6: 54870471-54870471 on Build GRCh38
rs754974713 Chr.6: 54870349-54870349 on Build GRCh38
rs757204057 Chr.6: 54870292-54870292 on Build GRCh38
rs748698722 Chr.6: 54870254-54870254 on Build GRCh38
rs761383368 Chr.6: 54870397-54870397 on Build GRCh38
rs369748971 Chr.6: 54870378-54870378 on Build GRCh38
rs144474594 Chr.6: 54870277-54870277 on Build GRCh38
rs781446217 Chr.6: 54870351-54870351 on Build GRCh38
rs971514482 Chr.6: 54870327-54870327 on Build GRCh38
rs753733022 Chr.6: 54870285-54870285 on Build GRCh38
rs933146096 Chr.6: 54870110-54870110 on Build GRCh38
rs373799741 Chr.6: 54870228-54870228 on Build GRCh38
rs747262889 Chr.6: 54870339-54870339 on Build GRCh38
rs772987010 Chr.6: 54870543-54870543 on Build GRCh38
rs763596819 Chr.6: 54870284-54870284 on Build GRCh38
rs747048951 Chr.6: 54870245-54870245 on Build GRCh38
rs139758773 Chr.6: 54870253-54870253 on Build GRCh38
rs755716380 Chr.6: 54870214-54870214 on Build GRCh38
rs914547382 Chr.6: 54870475-54870475 on Build GRCh38
rs926507610 Chr.6: 54870108-54870108 on Build GRCh38
rs917248640 Chr.6: 54870192-54870192 on Build GRCh38
rs773921453 Chr.6: 54870361-54870361 on Build GRCh38
rs981588059 Chr.6: 54870332-54870332 on Build GRCh38
rs750183894 Chr.6: 54870319-54870319 on Build GRCh38
rs7765721 Chr.6: 54870143-54870143 on Build GRCh38
rs139994975 Chr.6: 54870518-54870518 on Build GRCh38
rs770234600 Chr.6: 54870261-54870261 on Build GRCh38
rs769047828 Chr.6: 54870452-54870452 on Build GRCh38
rs200423933 Chr.6: 54870271-54870271 on Build GRCh38
rs868719938 Chr.6: 54870283-54870283 on Build GRCh38
rs776327181 Chr.6: 54870509-54870509 on Build GRCh38
rs752819836 Chr.6: 54870448-54870448 on Build GRCh38
rs796718901 Chr.6: 54870545-54870545 on Build GRCh38
rs992510499 Chr.6: 54870144-54870144 on Build GRCh38
rs147188411 Chr.6: 54870280-54870280 on Build GRCh38
rs565758775 Chr.6: 54870353-54870353 on Build GRCh38
rs779355597 Chr.6: 54870485-54870485 on Build GRCh38
rs752115650 Chr.6: 54870201-54870201 on Build GRCh38
rs766230067 Chr.6: 54870417-54870417 on Build GRCh38
rs1472679 Chr.6: 54870324-54870324 on Build GRCh38
rs771673694 Chr.6: 54870154-54870154 on Build GRCh38
rs776093156 Chr.6: 54870395-54870395 on Build GRCh38
rs762591829 Chr.6: 54870416-54870416 on Build GRCh38
rs755228965 Chr.6: 54870422-54870422 on Build GRCh38
rs372938760 Chr.6: 54870426-54870426 on Build GRCh38
rs534742689 Chr.6: 54870470-54870470 on Build GRCh38
rs35109988 Chr.6: 54870320-54870321 on Build GRCh38
rs768263411 Chr.6: 54870501-54870501 on Build GRCh38
rs746263850 Chr.6: 54870487-54870487 on Build GRCh38
rs756319912 Chr.6: 54870450-54870450 on Build GRCh38
rs775004920 Chr.6: 54870274-54870274 on Build GRCh38
rs552508963 Chr.6: 54870094-54870094 on Build GRCh38
rs756977112 Chr.6: 54870234-54870234 on Build GRCh38
rs897529010 Chr.6: 54870221-54870221 on Build GRCh38
rs767250037 Chr.6: 54870198-54870198 on Build GRCh38
rs768101945 Chr.6: 54870391-54870391 on Build GRCh38
rs796137406 Chr.6: 54870468-54870468 on Build GRCh38
rs207466978 Chr.6: 54870255-54870255 on Build GRCh38
rs765204933 Chr.6: 54870296-54870296 on Build GRCh38
rs770393974 Chr.6: 54870359-54870359 on Build GRCh38
rs16886105 Chr.6: 54870546-54870546 on Build GRCh38
rs192523695 Chr.6: 54870184-54870184 on Build GRCh38
rs758521772 Chr.6: 54870242-54870242 on Build GRCh38
rs549809239 Chr.6: 54870239-54870239 on Build GRCh38
rs749602760 Chr.6: 54870369-54870369 on Build GRCh38

View Full Product Details