Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 55758853-55759390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP5
Entrez Gene ID
653
Gene Name
bone morphogenetic protein 5
Gene Aliases
-
UniGene
Hs.296648
Gene Chromosome Location
Chr.6: 55753653-55875625 on Build GRCh38
Transcripts
NM_021073.2 XM_005249304.3 XM_011514816.2 XM_017011198.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1621793 c.1202T>G p.I401R Chr.6: 55759018-55759018 on Build GRCh38
742455 c.1108T>C p.W370R Chr.6: 55759112-55759112 on Build GRCh38
1621794 c.1151G>T p.G384V Chr.6: 55759069-55759069 on Build GRCh38
3158226 c.1147G>C p.D383H Chr.6: 55759073-55759073 on Build GRCh38
3874884 c.1211C>T p.T404I Chr.6: 55759009-55759009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202201755 Chr.6: 55759146-55759146 on Build GRCh38
rs933639698 Chr.6: 55759244-55759244 on Build GRCh38
rs944442339 Chr.6: 55759289-55759289 on Build GRCh38
rs201357101 Chr.6: 55759184-55759184 on Build GRCh38
rs780413486 Chr.6: 55759148-55759148 on Build GRCh38
rs766304070 Chr.6: 55759120-55759120 on Build GRCh38
rs896968214 Chr.6: 55759348-55759348 on Build GRCh38
rs765588364 Chr.6: 55759159-55759184 on Build GRCh38
rs200559487 Chr.6: 55758980-55758980 on Build GRCh38
rs112492283 Chr.6: 55759146-55759147 on Build GRCh38
rs144501971 Chr.6: 55758940-55758940 on Build GRCh38
rs754365141 Chr.6: 55759145-55759146 on Build GRCh38
rs750263775 Chr.6: 55758986-55758986 on Build GRCh38
rs781661428 Chr.6: 55759197-55759197 on Build GRCh38
rs767486670 Chr.6: 55759131-55759131 on Build GRCh38
rs779328371 Chr.6: 55759059-55759059 on Build GRCh38
rs753026421 Chr.6: 55759034-55759034 on Build GRCh38
rs1026315132 Chr.6: 55758886-55758886 on Build GRCh38
rs1041672491 Chr.6: 55759072-55759072 on Build GRCh38
rs184900087 Chr.6: 55759089-55759089 on Build GRCh38
rs779462976 Chr.6: 55759144-55759144 on Build GRCh38
rs756256577 Chr.6: 55759042-55759042 on Build GRCh38
rs952749558 Chr.6: 55758895-55758895 on Build GRCh38
rs746210301 Chr.6: 55759067-55759067 on Build GRCh38
rs556717308 Chr.6: 55759097-55759097 on Build GRCh38
rs769515411 Chr.6: 55759152-55759152 on Build GRCh38
rs935679456 Chr.6: 55759209-55759209 on Build GRCh38
rs888885620 Chr.6: 55759262-55759262 on Build GRCh38
rs990425669 Chr.6: 55758994-55758994 on Build GRCh38
rs773493883 Chr.6: 55759134-55759144 on Build GRCh38
rs745806206 Chr.6: 55759123-55759124 on Build GRCh38
rs564913582 Chr.6: 55759341-55759341 on Build GRCh38
rs756371439 Chr.6: 55759134-55759134 on Build GRCh38
rs769785414 Chr.6: 55759122-55759123 on Build GRCh38
rs925850772 Chr.6: 55759206-55759206 on Build GRCh38
rs879148488 Chr.6: 55759147-55759147 on Build GRCh38
rs946216383 Chr.6: 55759212-55759212 on Build GRCh38
rs770244836 Chr.6: 55759270-55759270 on Build GRCh38
rs529346654 Chr.6: 55758913-55758913 on Build GRCh38
rs752897944 Chr.6: 55759132-55759132 on Build GRCh38
rs747746901 Chr.6: 55759150-55759150 on Build GRCh38
rs927408984 Chr.6: 55758950-55758950 on Build GRCh38
rs757330989 Chr.6: 55759140-55759140 on Build GRCh38
rs796641932 Chr.6: 55759174-55759174 on Build GRCh38
rs753925688 Chr.6: 55759139-55759139 on Build GRCh38
rs759390981 Chr.6: 55759130-55759130 on Build GRCh38
rs1038301133 Chr.6: 55759345-55759345 on Build GRCh38
rs749711064 Chr.6: 55759047-55759047 on Build GRCh38
rs571202197 Chr.6: 55759088-55759088 on Build GRCh38
rs545018311 Chr.6: 55759313-55759313 on Build GRCh38
rs180770505 Chr.6: 55759060-55759060 on Build GRCh38
rs202085587 Chr.6: 55759023-55759023 on Build GRCh38
rs771503011 Chr.6: 55759053-55759053 on Build GRCh38
rs749127959 Chr.6: 55759124-55759125 on Build GRCh38
rs748144239 Chr.6: 55759172-55759184 on Build GRCh38
rs760281938 Chr.6: 55758965-55758965 on Build GRCh38
rs755292747 Chr.6: 55758932-55758932 on Build GRCh38
rs189414201 Chr.6: 55759273-55759273 on Build GRCh38
rs866204283 Chr.6: 55759172-55759172 on Build GRCh38
rs368744034 Chr.6: 55759184-55759190 on Build GRCh38
rs889458669 Chr.6: 55759293-55759293 on Build GRCh38
rs780694955 Chr.6: 55759118-55759119 on Build GRCh38
rs762957997 Chr.6: 55758995-55758995 on Build GRCh38
rs772486595 Chr.6: 55759077-55759077 on Build GRCh38
rs772775350 Chr.6: 55759154-55759154 on Build GRCh38
rs111885475 Chr.6: 55759181-55759181 on Build GRCh38
rs993084827 Chr.6: 55758981-55758981 on Build GRCh38
rs594910 Chr.6: 55759173-55759173 on Build GRCh38
rs760824407 Chr.6: 55759142-55759144 on Build GRCh38
rs72868839 Chr.6: 55759219-55759219 on Build GRCh38
rs994392969 Chr.6: 55758877-55758877 on Build GRCh38
rs775390057 Chr.6: 55759125-55759126 on Build GRCh38
rs370552267 Chr.6: 55758970-55758970 on Build GRCh38
rs764042132 Chr.6: 55759138-55759138 on Build GRCh38
rs766037115 Chr.6: 55759019-55759019 on Build GRCh38
rs765004095 Chr.6: 55759188-55759197 on Build GRCh38
rs762757308 Chr.6: 55759112-55759112 on Build GRCh38
rs767043356 Chr.6: 55759145-55759172 on Build GRCh38
rs879042530 Chr.6: 55759172-55759173 on Build GRCh38
rs774803268 Chr.6: 55758962-55758962 on Build GRCh38
rs534656598 Chr.6: 55759046-55759046 on Build GRCh38
rs754857723 Chr.6: 55759028-55759028 on Build GRCh38
rs772798463 Chr.6: 55759108-55759108 on Build GRCh38
rs977819665 Chr.6: 55759215-55759215 on Build GRCh38
rs761303484 Chr.6: 55758973-55758973 on Build GRCh38
rs774475300 Chr.6: 55759127-55759127 on Build GRCh38

View Full Product Details