Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 56457880-56458417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
UniGene
Hs.604915
Gene Chromosome Location
Chr.6: 56457987-56954671 on Build GRCh38
Transcripts
NM_001144769.2 NM_001144770.1 NM_015548.4 NM_183380.3 XM_005249310.3 XM_005249315.3 XM_005249316.2 XM_005249318.2 XM_005249319.2 XM_005249320.4 XM_005249322.4 XM_005249323.3 XM_005249324.2 XM_011514824.2 XM_011514825.2 XM_011514826.2 XM_017011205.1 XM_017011206.1 XM_017011207.1 XM_017011208.1 XM_017011209.1 XM_017011210.1 XM_017011211.1 XM_017011212.1 XM_017011213.1 XM_017011214.1 XM_017011215.1 XM_017011216.1 XM_017011217.1 XM_017011218.1 XM_017011219.1 XM_017011220.1 XM_017011221.1 XM_017011222.1 XM_017011223.1 XM_017011224.1

SNP Detail

SNP ID Chromosome Location
rs974139402 Chr.6: 56458042-56458045 on Build GRCh38
rs899607989 Chr.6: 56458197-56458197 on Build GRCh38
rs543055153 Chr.6: 56458323-56458323 on Build GRCh38
rs1031082207 Chr.6: 56458273-56458273 on Build GRCh38
rs534574076 Chr.6: 56458126-56458126 on Build GRCh38
rs948880918 Chr.6: 56458083-56458083 on Build GRCh38
rs1045930248 Chr.6: 56458088-56458091 on Build GRCh38
rs554743209 Chr.6: 56458337-56458337 on Build GRCh38
rs921867708 Chr.6: 56458365-56458365 on Build GRCh38
rs765337905 Chr.6: 56458289-56458290 on Build GRCh38
rs1045179656 Chr.6: 56458203-56458203 on Build GRCh38
rs955350820 Chr.6: 56457973-56457973 on Build GRCh38
rs572339946 Chr.6: 56458304-56458304 on Build GRCh38
rs896703351 Chr.6: 56458163-56458163 on Build GRCh38
rs566132688 Chr.6: 56458025-56458025 on Build GRCh38
rs556410193 Chr.6: 56458184-56458184 on Build GRCh38
rs916054830 Chr.6: 56458057-56458057 on Build GRCh38
rs1022073568 Chr.6: 56458194-56458194 on Build GRCh38
rs982015177 Chr.6: 56458384-56458384 on Build GRCh38
rs908552911 Chr.6: 56457994-56457994 on Build GRCh38
rs545265415 Chr.6: 56458181-56458182 on Build GRCh38
rs982743900 Chr.6: 56457985-56457985 on Build GRCh38
rs765860167 Chr.6: 56457989-56457989 on Build GRCh38
rs941412574 Chr.6: 56458010-56458010 on Build GRCh38
rs183822595 Chr.6: 56458093-56458093 on Build GRCh38
rs571540607 Chr.6: 56458188-56458188 on Build GRCh38
rs386701456 Chr.6: 56458183-56458184 on Build GRCh38
rs41271848 Chr.6: 56458363-56458363 on Build GRCh38
rs565221572 Chr.6: 56457922-56457922 on Build GRCh38
rs1048783725 Chr.6: 56458111-56458111 on Build GRCh38
rs997074324 Chr.6: 56458214-56458215 on Build GRCh38
rs537632090 Chr.6: 56458183-56458183 on Build GRCh38
rs957041226 Chr.6: 56458279-56458279 on Build GRCh38
rs778570158 Chr.6: 56458186-56458186 on Build GRCh38
rs930499751 Chr.6: 56458097-56458097 on Build GRCh38
rs191122945 Chr.6: 56457950-56457950 on Build GRCh38
rs922496428 Chr.6: 56457954-56457954 on Build GRCh38
rs777743507 Chr.6: 56457998-56457998 on Build GRCh38
rs567385337 Chr.6: 56458133-56458133 on Build GRCh38
rs576515609 Chr.6: 56458346-56458346 on Build GRCh38
rs944615516 Chr.6: 56458005-56458005 on Build GRCh38
rs1010663796 Chr.6: 56457905-56457905 on Build GRCh38
rs550931171 Chr.6: 56458024-56458024 on Build GRCh38
rs962702376 Chr.6: 56458333-56458333 on Build GRCh38
rs965930333 Chr.6: 56458359-56458359 on Build GRCh38
rs564559694 Chr.6: 56458188-56458188 on Build GRCh38
rs907653796 Chr.6: 56458116-56458116 on Build GRCh38
rs1056541102 Chr.6: 56458144-56458144 on Build GRCh38
rs1012471205 Chr.6: 56458177-56458177 on Build GRCh38
rs974535967 Chr.6: 56458091-56458091 on Build GRCh38
rs1001464929 Chr.6: 56458265-56458265 on Build GRCh38
rs188367907 Chr.6: 56458251-56458251 on Build GRCh38
rs1052061342 Chr.6: 56458128-56458128 on Build GRCh38
rs147960759 Chr.6: 56458252-56458252 on Build GRCh38
rs911806412 Chr.6: 56457983-56457983 on Build GRCh38
rs1022104384 Chr.6: 56457915-56457915 on Build GRCh38
rs1008197550 Chr.6: 56458287-56458287 on Build GRCh38
rs951908413 Chr.6: 56458377-56458377 on Build GRCh38

View Full Product Details