Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 56458690-56459187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
UniGene
Hs.604915
Gene Chromosome Location
Chr.6: 56457987-56954671 on Build GRCh38
Transcripts
NM_001144769.2 NM_001144770.1 NM_015548.4 NM_183380.3 XM_005249310.3 XM_005249315.3 XM_005249316.2 XM_005249318.2 XM_005249319.2 XM_005249320.4 XM_005249322.4 XM_005249323.3 XM_005249324.2 XM_011514824.2 XM_011514825.2 XM_011514826.2 XM_017011205.1 XM_017011206.1 XM_017011207.1 XM_017011208.1 XM_017011209.1 XM_017011210.1 XM_017011211.1 XM_017011212.1 XM_017011213.1 XM_017011214.1 XM_017011215.1 XM_017011216.1 XM_017011217.1 XM_017011218.1 XM_017011219.1 XM_017011220.1 XM_017011221.1 XM_017011222.1 XM_017011223.1 XM_017011224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1621804 c.15464C>T p.T5155M Chr.6: 56459057-56459057 on Build GRCh38
4387992 c.15432G>T p.R5144R Chr.6: 56459089-56459089 on Build GRCh38
4619292 c.22542A>G p.R7514R Chr.6: 56459101-56459101 on Build GRCh38
4989433 c.15416C>T p.P5139L Chr.6: 56459105-56459105 on Build GRCh38
3628945 c.22463C>T p.S7488L Chr.6: 56459180-56459180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023353247 Chr.6: 56458874-56458874 on Build GRCh38
rs747867774 Chr.6: 56459027-56459027 on Build GRCh38
rs917690539 Chr.6: 56458852-56458852 on Build GRCh38
rs1036786598 Chr.6: 56459152-56459152 on Build GRCh38
rs925524863 Chr.6: 56458921-56458921 on Build GRCh38
rs761116868 Chr.6: 56459090-56459090 on Build GRCh38
rs375978950 Chr.6: 56459102-56459102 on Build GRCh38
rs769636857 Chr.6: 56459109-56459109 on Build GRCh38
rs372672868 Chr.6: 56459033-56459033 on Build GRCh38
rs761169233 Chr.6: 56458993-56458993 on Build GRCh38
rs549957580 Chr.6: 56458717-56458717 on Build GRCh38
rs952061663 Chr.6: 56458748-56458748 on Build GRCh38
rs747080141 Chr.6: 56458756-56458756 on Build GRCh38
rs996888275 Chr.6: 56458729-56458729 on Build GRCh38
rs936924815 Chr.6: 56458922-56458922 on Build GRCh38
rs767445108 Chr.6: 56458979-56458979 on Build GRCh38
rs995386312 Chr.6: 56458849-56458849 on Build GRCh38
rs576431308 Chr.6: 56458938-56458938 on Build GRCh38
rs779478186 Chr.6: 56458975-56458975 on Build GRCh38
rs781243197 Chr.6: 56459020-56459020 on Build GRCh38
rs932364292 Chr.6: 56458794-56458794 on Build GRCh38
rs771266784 Chr.6: 56459141-56459141 on Build GRCh38
rs1014870620 Chr.6: 56459146-56459146 on Build GRCh38
rs920945556 Chr.6: 56458786-56458786 on Build GRCh38
rs763211663 Chr.6: 56459041-56459041 on Build GRCh38
rs367598583 Chr.6: 56459057-56459057 on Build GRCh38
rs753285303 Chr.6: 56459078-56459078 on Build GRCh38
rs554105582 Chr.6: 56458804-56458804 on Build GRCh38
rs774765327 Chr.6: 56459151-56459151 on Build GRCh38
rs988975062 Chr.6: 56459028-56459028 on Build GRCh38
rs947722160 Chr.6: 56458902-56458902 on Build GRCh38
rs577012766 Chr.6: 56459107-56459107 on Build GRCh38
rs750052381 Chr.6: 56458964-56458964 on Build GRCh38
rs1043745825 Chr.6: 56458757-56458757 on Build GRCh38
rs961791072 Chr.6: 56458799-56458799 on Build GRCh38
rs768572908 Chr.6: 56459000-56459000 on Build GRCh38
rs1036860215 Chr.6: 56458835-56458836 on Build GRCh38
rs201891015 Chr.6: 56459030-56459030 on Build GRCh38
rs759749631 Chr.6: 56459063-56459063 on Build GRCh38
rs183265598 Chr.6: 56458906-56458906 on Build GRCh38
rs956139209 Chr.6: 56459025-56459025 on Build GRCh38
rs371464814 Chr.6: 56459077-56459077 on Build GRCh38
rs777558951 Chr.6: 56459114-56459114 on Build GRCh38
rs79455288 Chr.6: 56458752-56458752 on Build GRCh38
rs964020701 Chr.6: 56458724-56458724 on Build GRCh38
rs980439707 Chr.6: 56458904-56458904 on Build GRCh38
rs1003347545 Chr.6: 56458951-56458951 on Build GRCh38
rs939814173 Chr.6: 56459137-56459137 on Build GRCh38
rs892642190 Chr.6: 56458940-56458940 on Build GRCh38
rs536235549 Chr.6: 56458844-56458844 on Build GRCh38
rs199881276 Chr.6: 56459056-56459056 on Build GRCh38
rs375622195 Chr.6: 56458985-56458985 on Build GRCh38
rs565992623 Chr.6: 56458831-56458831 on Build GRCh38
rs758066256 Chr.6: 56458967-56458967 on Build GRCh38
rs754164327 Chr.6: 56459091-56459091 on Build GRCh38
rs776966911 Chr.6: 56458991-56458991 on Build GRCh38
rs1028904918 Chr.6: 56458741-56458741 on Build GRCh38
rs780600756 Chr.6: 56459108-56459108 on Build GRCh38
rs779746657 Chr.6: 56459103-56459103 on Build GRCh38
rs769595354 Chr.6: 56459029-56459029 on Build GRCh38
rs749163618 Chr.6: 56459128-56459128 on Build GRCh38
rs751052704 Chr.6: 56459104-56459104 on Build GRCh38
rs558022526 Chr.6: 56459097-56459097 on Build GRCh38
rs372784136 Chr.6: 56458939-56458939 on Build GRCh38

View Full Product Details