Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70232393-70232934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A1
Entrez Gene ID
1297
Gene Name
collagen type IX alpha 1
Gene Aliases
DJ149L1.1.2, EDM6, MED, STL4
UniGene
Hs.590892
Gene Chromosome Location
Chr.6: 70216040-70303083 on Build GRCh38
Transcripts
NM_001851.4 NM_078485.3 XM_011535429.2 XM_011535430.2 XM_017010246.1 XM_017010247.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3366586 c.1735A>T p.K579* Chr.6: 70232622-70232622 on Build GRCh38
5996873 c.1757G>C p.G586A Chr.6: 70232600-70232600 on Build GRCh38
451744 c.2402C>T p.P801L Chr.6: 70232684-70232684 on Build GRCh38
4786705 c.1674C>T p.P558P Chr.6: 70232683-70232683 on Build GRCh38
5699397 c.1678C>A p.P560T Chr.6: 70232679-70232679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896106778 Chr.6: 70232529-70232529 on Build GRCh38
rs34119578 Chr.6: 70232616-70232616 on Build GRCh38
rs757620185 Chr.6: 70232629-70232629 on Build GRCh38
rs554442935 Chr.6: 70232490-70232490 on Build GRCh38
rs375684014 Chr.6: 70232738-70232738 on Build GRCh38
rs749231567 Chr.6: 70232823-70232823 on Build GRCh38
rs972916458 Chr.6: 70232767-70232767 on Build GRCh38
rs182052281 Chr.6: 70232834-70232834 on Build GRCh38
rs923041425 Chr.6: 70232889-70232889 on Build GRCh38
rs1049217906 Chr.6: 70232541-70232541 on Build GRCh38
rs961616754 Chr.6: 70232444-70232444 on Build GRCh38
rs930272823 Chr.6: 70232528-70232528 on Build GRCh38
rs961499099 Chr.6: 70232751-70232751 on Build GRCh38
rs764624908 Chr.6: 70232594-70232594 on Build GRCh38
rs375886220 Chr.6: 70232555-70232555 on Build GRCh38
rs910283521 Chr.6: 70232542-70232542 on Build GRCh38
rs777634301 Chr.6: 70232819-70232819 on Build GRCh38
rs527945609 Chr.6: 70232688-70232688 on Build GRCh38
rs752197248 Chr.6: 70232783-70232783 on Build GRCh38
rs751254245 Chr.6: 70232635-70232635 on Build GRCh38
rs771141590 Chr.6: 70232817-70232818 on Build GRCh38
rs74699744 Chr.6: 70232514-70232514 on Build GRCh38
rs1057494383 Chr.6: 70232520-70232520 on Build GRCh38
rs144581626 Chr.6: 70232630-70232630 on Build GRCh38
rs566003668 Chr.6: 70232750-70232750 on Build GRCh38
rs775466585 Chr.6: 70232687-70232687 on Build GRCh38
rs767229176 Chr.6: 70232753-70232753 on Build GRCh38
rs755614368 Chr.6: 70232797-70232797 on Build GRCh38
rs201159365 Chr.6: 70232591-70232591 on Build GRCh38
rs780851685 Chr.6: 70232640-70232640 on Build GRCh38
rs774610828 Chr.6: 70232682-70232682 on Build GRCh38
rs1014122856 Chr.6: 70232726-70232726 on Build GRCh38
rs368426763 Chr.6: 70232537-70232537 on Build GRCh38
rs752717596 Chr.6: 70232563-70232563 on Build GRCh38
rs867488468 Chr.6: 70232517-70232517 on Build GRCh38
rs373877078 Chr.6: 70232721-70232721 on Build GRCh38
rs947055932 Chr.6: 70232840-70232840 on Build GRCh38
rs777744864 Chr.6: 70232665-70232665 on Build GRCh38
rs771103997 Chr.6: 70232543-70232543 on Build GRCh38
rs754596057 Chr.6: 70232639-70232639 on Build GRCh38
rs1022948354 Chr.6: 70232534-70232534 on Build GRCh38
rs1043172574 Chr.6: 70232852-70232852 on Build GRCh38
rs530239619 Chr.6: 70232886-70232886 on Build GRCh38
rs754706930 Chr.6: 70232746-70232746 on Build GRCh38
rs749074294 Chr.6: 70232679-70232679 on Build GRCh38
rs747652928 Chr.6: 70232647-70232647 on Build GRCh38
rs770705023 Chr.6: 70232680-70232680 on Build GRCh38
rs975062720 Chr.6: 70232798-70232798 on Build GRCh38
rs945181616 Chr.6: 70232897-70232897 on Build GRCh38
rs747230904 Chr.6: 70232777-70232778 on Build GRCh38
rs762152784 Chr.6: 70232697-70232697 on Build GRCh38
rs759764758 Chr.6: 70232683-70232683 on Build GRCh38
rs561098568 Chr.6: 70232854-70232854 on Build GRCh38
rs1012860761 Chr.6: 70232531-70232531 on Build GRCh38
rs147095719 Chr.6: 70232685-70232685 on Build GRCh38
rs141830060 Chr.6: 70232650-70232650 on Build GRCh38
rs748515682 Chr.6: 70232442-70232442 on Build GRCh38
rs372626522 Chr.6: 70232545-70232545 on Build GRCh38
rs142257977 Chr.6: 70232513-70232513 on Build GRCh38
rs1040487573 Chr.6: 70232862-70232863 on Build GRCh38
rs992324331 Chr.6: 70232441-70232441 on Build GRCh38
rs765684676 Chr.6: 70232705-70232705 on Build GRCh38
rs770189715 Chr.6: 70232860-70232860 on Build GRCh38
rs959746959 Chr.6: 70232440-70232440 on Build GRCh38

View Full Product Details