Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70955738-70956244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GAT2
Entrez Gene ID
135152
Gene Name
beta-1,3-glucuronyltransferase 2
Gene Aliases
GLCATS
UniGene
Hs.713609
Gene Chromosome Location
Chr.6: 70861366-70957085 on Build GRCh38
Transcripts
NM_080742.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5634393 c.507G>C p.Q169H Chr.6: 70955923-70955923 on Build GRCh38
6547409 c.286G>A p.A96T Chr.6: 70956144-70956144 on Build GRCh38
5695204 c.478C>G p.R160G Chr.6: 70955952-70955952 on Build GRCh38
4542758 c.324G>A p.V108V Chr.6: 70956106-70956106 on Build GRCh38
6609286 c.549C>T p.F183F Chr.6: 70955881-70955881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774281144 Chr.6: 70956178-70956178 on Build GRCh38
rs995151563 Chr.6: 70956221-70956221 on Build GRCh38
rs767334400 Chr.6: 70956192-70956192 on Build GRCh38
rs758035011 Chr.6: 70955875-70955875 on Build GRCh38
rs752093504 Chr.6: 70956193-70956194 on Build GRCh38
rs781459405 Chr.6: 70955883-70955883 on Build GRCh38
rs973447037 Chr.6: 70955803-70955803 on Build GRCh38
rs1034177636 Chr.6: 70955760-70955760 on Build GRCh38
rs1040279371 Chr.6: 70955971-70955971 on Build GRCh38
rs751495356 Chr.6: 70956097-70956097 on Build GRCh38
rs752952453 Chr.6: 70955994-70955994 on Build GRCh38
rs745827101 Chr.6: 70955907-70955907 on Build GRCh38
rs755698058 Chr.6: 70956131-70956131 on Build GRCh38
rs759422862 Chr.6: 70956132-70956132 on Build GRCh38
rs760430051 Chr.6: 70955798-70955798 on Build GRCh38
rs762810318 Chr.6: 70956093-70956093 on Build GRCh38
rs759798543 Chr.6: 70955906-70955907 on Build GRCh38
rs753613802 Chr.6: 70955886-70955886 on Build GRCh38
rs764968027 Chr.6: 70956003-70956003 on Build GRCh38
rs373218616 Chr.6: 70956056-70956056 on Build GRCh38
rs62420336 Chr.6: 70955776-70955776 on Build GRCh38
rs778874580 Chr.6: 70955898-70955898 on Build GRCh38
rs747788346 Chr.6: 70956030-70956030 on Build GRCh38
rs752716799 Chr.6: 70956195-70956195 on Build GRCh38
rs112409269 Chr.6: 70956032-70956032 on Build GRCh38
rs746595710 Chr.6: 70955927-70955927 on Build GRCh38
rs1053255527 Chr.6: 70956021-70956021 on Build GRCh38
rs767627778 Chr.6: 70956126-70956126 on Build GRCh38
rs754177309 Chr.6: 70955881-70955883 on Build GRCh38
rs752787225 Chr.6: 70956138-70956138 on Build GRCh38
rs774796873 Chr.6: 70956074-70956074 on Build GRCh38
rs747583290 Chr.6: 70956222-70956222 on Build GRCh38
rs758815977 Chr.6: 70956217-70956217 on Build GRCh38
rs373530810 Chr.6: 70956022-70956022 on Build GRCh38
rs777256563 Chr.6: 70956141-70956141 on Build GRCh38
rs765020213 Chr.6: 70956039-70956039 on Build GRCh38
rs1024358868 Chr.6: 70955827-70955827 on Build GRCh38
rs768419572 Chr.6: 70955937-70955937 on Build GRCh38
rs751590012 Chr.6: 70956054-70956054 on Build GRCh38
rs751723321 Chr.6: 70955876-70955876 on Build GRCh38
rs763680241 Chr.6: 70956206-70956206 on Build GRCh38
rs770096506 Chr.6: 70955959-70955959 on Build GRCh38
rs750778011 Chr.6: 70956215-70956215 on Build GRCh38
rs752135630 Chr.6: 70956130-70956130 on Build GRCh38
rs201430647 Chr.6: 70956162-70956162 on Build GRCh38
rs778605827 Chr.6: 70956212-70956212 on Build GRCh38
rs879653030 Chr.6: 70956007-70956007 on Build GRCh38
rs940633060 Chr.6: 70955801-70955801 on Build GRCh38
rs780861676 Chr.6: 70956060-70956060 on Build GRCh38
rs753554542 Chr.6: 70956147-70956147 on Build GRCh38
rs370336662 Chr.6: 70956048-70956048 on Build GRCh38
rs760326264 Chr.6: 70955865-70955867 on Build GRCh38
rs568523673 Chr.6: 70956210-70956210 on Build GRCh38
rs556286650 Chr.6: 70956186-70956186 on Build GRCh38
rs369211351 Chr.6: 70956154-70956154 on Build GRCh38
rs767702737 Chr.6: 70955993-70955993 on Build GRCh38
rs778085009 Chr.6: 70956191-70956192 on Build GRCh38
rs777142818 Chr.6: 70955816-70955816 on Build GRCh38
rs757032901 Chr.6: 70955995-70955995 on Build GRCh38
rs776486029 Chr.6: 70955938-70955938 on Build GRCh38
rs771414127 Chr.6: 70956035-70956035 on Build GRCh38
rs765762864 Chr.6: 70955838-70955838 on Build GRCh38
rs1002276626 Chr.6: 70955963-70955963 on Build GRCh38
rs765433016 Chr.6: 70956057-70956060 on Build GRCh38
rs762149503 Chr.6: 70956167-70956167 on Build GRCh38
rs757565890 Chr.6: 70956150-70956150 on Build GRCh38
rs757048328 Chr.6: 70955892-70955892 on Build GRCh38
rs766413949 Chr.6: 70955974-70955974 on Build GRCh38
rs766309638 Chr.6: 70956094-70956094 on Build GRCh38
rs746394122 Chr.6: 70956077-70956077 on Build GRCh38
rs896531028 Chr.6: 70956177-70956177 on Build GRCh38
rs1044005381 Chr.6: 70956153-70956153 on Build GRCh38
rs199757620 Chr.6: 70956073-70956073 on Build GRCh38
rs750941214 Chr.6: 70955872-70955872 on Build GRCh38
rs758953378 Chr.6: 70956165-70956166 on Build GRCh38
rs944850761 Chr.6: 70955896-70955896 on Build GRCh38
rs951776976 Chr.6: 70956158-70956158 on Build GRCh38
rs764212379 Chr.6: 70956090-70956090 on Build GRCh38
rs766181134 Chr.6: 70955869-70955871 on Build GRCh38
rs771519115 Chr.6: 70955788-70955788 on Build GRCh38
rs376563227 Chr.6: 70956087-70956087 on Build GRCh38
rs762215470 Chr.6: 70955955-70955955 on Build GRCh38
rs532126324 Chr.6: 70955769-70955769 on Build GRCh38
rs890603320 Chr.6: 70955888-70955888 on Build GRCh38
rs777538824 Chr.6: 70956068-70956068 on Build GRCh38
rs758276244 Chr.6: 70956015-70956015 on Build GRCh38
rs747768660 Chr.6: 70956061-70956061 on Build GRCh38
rs9446312 Chr.6: 70956148-70956148 on Build GRCh38
rs771566480 Chr.6: 70955912-70955912 on Build GRCh38
rs534598813 Chr.6: 70956159-70956159 on Build GRCh38
rs767612928 Chr.6: 70956037-70956037 on Build GRCh38
rs527522804 Chr.6: 70956031-70956031 on Build GRCh38
rs779726717 Chr.6: 70955917-70955917 on Build GRCh38
rs200162847 Chr.6: 70955977-70955977 on Build GRCh38
rs775171456 Chr.6: 70955792-70955792 on Build GRCh38
rs769087175 Chr.6: 70955800-70955800 on Build GRCh38
rs951809246 Chr.6: 70955952-70955952 on Build GRCh38
rs776679570 Chr.6: 70956165-70956165 on Build GRCh38
rs755195344 Chr.6: 70955881-70955881 on Build GRCh38
rs760058633 Chr.6: 70956205-70956205 on Build GRCh38
rs775439822 Chr.6: 70956086-70956086 on Build GRCh38
rs753427344 Chr.6: 70956207-70956207 on Build GRCh38
rs147151504 Chr.6: 70956172-70956172 on Build GRCh38
rs755717028 Chr.6: 70956067-70956067 on Build GRCh38
rs780211141 Chr.6: 70956019-70956019 on Build GRCh38
rs9455275 Chr.6: 70955770-70955770 on Build GRCh38
rs762337236 Chr.6: 70955828-70955828 on Build GRCh38
rs529749468 Chr.6: 70955787-70955787 on Build GRCh38
rs552429904 Chr.6: 70956055-70956055 on Build GRCh38
rs149577161 Chr.6: 70956028-70956028 on Build GRCh38
rs372158787 Chr.6: 70956155-70956155 on Build GRCh38
rs750320042 Chr.6: 70956006-70956006 on Build GRCh38
rs759666660 Chr.6: 70955980-70955980 on Build GRCh38
rs904629150 Chr.6: 70955772-70955772 on Build GRCh38
rs983222335 Chr.6: 70955953-70955953 on Build GRCh38
rs773719748 Chr.6: 70955961-70955961 on Build GRCh38
rs993084237 Chr.6: 70955820-70955820 on Build GRCh38
rs759550215 Chr.6: 70956109-70956109 on Build GRCh38
rs759462366 Chr.6: 70956182-70956182 on Build GRCh38
rs780395953 Chr.6: 70956218-70956218 on Build GRCh38
rs956253849 Chr.6: 70955811-70955811 on Build GRCh38
rs766397393 Chr.6: 70956050-70956050 on Build GRCh38
rs749590540 Chr.6: 70956070-70956070 on Build GRCh38
rs143511495 Chr.6: 70956079-70956079 on Build GRCh38
rs760823778 Chr.6: 70956038-70956038 on Build GRCh38

View Full Product Details