Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70956133-70956635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GAT2
Entrez Gene ID
135152
Gene Name
beta-1,3-glucuronyltransferase 2
Gene Aliases
GLCATS
UniGene
Hs.713609
Gene Chromosome Location
Chr.6: 70861366-70957085 on Build GRCh38
Transcripts
NM_080742.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1445719 c.174C>T p.G58G Chr.6: 70956256-70956256 on Build GRCh38
4437238 c.11C>T p.A4V Chr.6: 70956419-70956419 on Build GRCh38
6547409 c.286G>A p.A96T Chr.6: 70956144-70956144 on Build GRCh38
5589780 c.63C>T p.I21I Chr.6: 70956367-70956367 on Build GRCh38
5596341 c.124C>T p.P42S Chr.6: 70956306-70956306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044296547 Chr.6: 70956475-70956475 on Build GRCh38
rs774281144 Chr.6: 70956178-70956178 on Build GRCh38
rs995151563 Chr.6: 70956221-70956221 on Build GRCh38
rs767334400 Chr.6: 70956192-70956192 on Build GRCh38
rs762681415 Chr.6: 70956347-70956347 on Build GRCh38
rs1056442323 Chr.6: 70956583-70956583 on Build GRCh38
rs752093504 Chr.6: 70956193-70956194 on Build GRCh38
rs921863953 Chr.6: 70956250-70956250 on Build GRCh38
rs775237359 Chr.6: 70956260-70956260 on Build GRCh38
rs371411394 Chr.6: 70956451-70956451 on Build GRCh38
rs867164665 Chr.6: 70956423-70956423 on Build GRCh38
rs760672632 Chr.6: 70956262-70956262 on Build GRCh38
rs906078505 Chr.6: 70956419-70956419 on Build GRCh38
rs9446313 Chr.6: 70956524-70956524 on Build GRCh38
rs768106390 Chr.6: 70956382-70956382 on Build GRCh38
rs761375136 Chr.6: 70956271-70956271 on Build GRCh38
rs1002214080 Chr.6: 70956420-70956420 on Build GRCh38
rs752716799 Chr.6: 70956195-70956195 on Build GRCh38
rs781346844 Chr.6: 70956240-70956240 on Build GRCh38
rs765259453 Chr.6: 70956269-70956269 on Build GRCh38
rs763443623 Chr.6: 70956265-70956265 on Build GRCh38
rs747583290 Chr.6: 70956222-70956222 on Build GRCh38
rs758815977 Chr.6: 70956217-70956217 on Build GRCh38
rs935967414 Chr.6: 70956595-70956595 on Build GRCh38
rs764758181 Chr.6: 70956336-70956336 on Build GRCh38
rs763680241 Chr.6: 70956206-70956206 on Build GRCh38
rs750778011 Chr.6: 70956215-70956215 on Build GRCh38
rs201430647 Chr.6: 70956162-70956162 on Build GRCh38
rs778605827 Chr.6: 70956212-70956212 on Build GRCh38
rs766587551 Chr.6: 70956350-70956350 on Build GRCh38
rs535966421 Chr.6: 70956249-70956249 on Build GRCh38
rs756301520 Chr.6: 70956293-70956293 on Build GRCh38
rs1320315 Chr.6: 70956443-70956443 on Build GRCh38
rs568523673 Chr.6: 70956210-70956210 on Build GRCh38
rs758650135 Chr.6: 70956282-70956282 on Build GRCh38
rs556286650 Chr.6: 70956186-70956186 on Build GRCh38
rs369211351 Chr.6: 70956154-70956154 on Build GRCh38
rs753287916 Chr.6: 70956421-70956421 on Build GRCh38
rs778085009 Chr.6: 70956191-70956192 on Build GRCh38
rs1574490 Chr.6: 70956283-70956283 on Build GRCh38
rs762149503 Chr.6: 70956167-70956167 on Build GRCh38
rs773913885 Chr.6: 70956470-70956470 on Build GRCh38
rs771703237 Chr.6: 70956314-70956314 on Build GRCh38
rs772803317 Chr.6: 70956342-70956342 on Build GRCh38
rs757495022 Chr.6: 70956437-70956437 on Build GRCh38
rs755345165 Chr.6: 70956369-70956369 on Build GRCh38
rs200678168 Chr.6: 70956279-70956279 on Build GRCh38
rs1029245325 Chr.6: 70956538-70956538 on Build GRCh38
rs896531028 Chr.6: 70956177-70956177 on Build GRCh38
rs775357510 Chr.6: 70956326-70956326 on Build GRCh38
rs1044005381 Chr.6: 70956153-70956153 on Build GRCh38
rs768381308 Chr.6: 70956446-70956446 on Build GRCh38
rs748304217 Chr.6: 70956245-70956245 on Build GRCh38
rs758953378 Chr.6: 70956165-70956166 on Build GRCh38
rs757632050 Chr.6: 70956265-70956288 on Build GRCh38
rs951776976 Chr.6: 70956158-70956158 on Build GRCh38
rs781591720 Chr.6: 70956365-70956367 on Build GRCh38
rs554377076 Chr.6: 70956273-70956273 on Build GRCh38
rs755504105 Chr.6: 70956284-70956284 on Build GRCh38
rs988847690 Chr.6: 70956598-70956598 on Build GRCh38
rs925217871 Chr.6: 70956563-70956563 on Build GRCh38
rs771928642 Chr.6: 70956258-70956258 on Build GRCh38
rs768571781 Chr.6: 70956229-70956229 on Build GRCh38
rs559736877 Chr.6: 70956574-70956574 on Build GRCh38
rs778098066 Chr.6: 70956297-70956297 on Build GRCh38
rs768628314 Chr.6: 70956332-70956332 on Build GRCh38
rs896520916 Chr.6: 70956593-70956593 on Build GRCh38
rs534598813 Chr.6: 70956159-70956159 on Build GRCh38
rs562160390 Chr.6: 70956511-70956511 on Build GRCh38
rs951703720 Chr.6: 70956449-70956449 on Build GRCh38
rs377152159 Chr.6: 70956555-70956555 on Build GRCh38
rs756209487 Chr.6: 70956431-70956431 on Build GRCh38
rs199861185 Chr.6: 70956247-70956247 on Build GRCh38
rs776679570 Chr.6: 70956165-70956165 on Build GRCh38
rs760058633 Chr.6: 70956205-70956205 on Build GRCh38
rs753427344 Chr.6: 70956207-70956207 on Build GRCh38
rs762426765 Chr.6: 70956453-70956453 on Build GRCh38
rs147151504 Chr.6: 70956172-70956172 on Build GRCh38
rs913309287 Chr.6: 70956606-70956606 on Build GRCh38
rs770593773 Chr.6: 70956467-70956467 on Build GRCh38
rs778009460 Chr.6: 70956434-70956434 on Build GRCh38
rs907659442 Chr.6: 70956225-70956225 on Build GRCh38
rs1048297438 Chr.6: 70956616-70956616 on Build GRCh38
rs776603346 Chr.6: 70956335-70956335 on Build GRCh38
rs779064100 Chr.6: 70956439-70956439 on Build GRCh38
rs745627538 Chr.6: 70956256-70956256 on Build GRCh38
rs965833167 Chr.6: 70956227-70956227 on Build GRCh38
rs1014625223 Chr.6: 70956506-70956506 on Build GRCh38
rs372158787 Chr.6: 70956155-70956155 on Build GRCh38
rs143347651 Chr.6: 70956334-70956334 on Build GRCh38
rs746848628 Chr.6: 70956331-70956331 on Build GRCh38
rs781778600 Chr.6: 70956286-70956286 on Build GRCh38
rs1046330445 Chr.6: 70956361-70956361 on Build GRCh38
rs769237161 Chr.6: 70956452-70956452 on Build GRCh38
rs979770442 Chr.6: 70956558-70956558 on Build GRCh38
rs780395953 Chr.6: 70956218-70956218 on Build GRCh38
rs759462366 Chr.6: 70956182-70956182 on Build GRCh38
rs751938366 Chr.6: 70956356-70956356 on Build GRCh38
rs773616950 Chr.6: 70956253-70956253 on Build GRCh38
rs748216440 Chr.6: 70956292-70956292 on Build GRCh38

View Full Product Details