Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73362585-73363086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDC3L
Entrez Gene ID
154288
Gene Name
KH domain containing 3 like, subcortical maternal complex member
Gene Aliases
C6orf221, ECAT1, HYDM2
UniGene
Hs.128326
Gene Chromosome Location
Chr.6: 73362677-73364175 on Build GRCh38
Transcripts
NM_001017361.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318813 c.124C>T p.P42S Chr.6: 73362853-73362853 on Build GRCh38
1487911 c.117G>T p.L39L Chr.6: 73362846-73362846 on Build GRCh38
7427502 c.138C>A p.R46R Chr.6: 73362867-73362867 on Build GRCh38
5888900 c.4G>A p.D2N Chr.6: 73362733-73362733 on Build GRCh38
6290417 c.89G>A p.R30Q Chr.6: 73362818-73362818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778300226 Chr.6: 73362921-73362921 on Build GRCh38
rs774504582 Chr.6: 73362817-73362817 on Build GRCh38
rs776478308 Chr.6: 73362835-73362835 on Build GRCh38
rs144291287 Chr.6: 73362743-73362743 on Build GRCh38
rs762515244 Chr.6: 73362876-73362876 on Build GRCh38
rs547174523 Chr.6: 73362918-73362918 on Build GRCh38
rs750335286 Chr.6: 73362859-73362859 on Build GRCh38
rs1056551148 Chr.6: 73362979-73362979 on Build GRCh38
rs200684575 Chr.6: 73363052-73363052 on Build GRCh38
rs763895578 Chr.6: 73362739-73362739 on Build GRCh38
rs11757433 Chr.6: 73362820-73362820 on Build GRCh38
rs968324083 Chr.6: 73362740-73362740 on Build GRCh38
rs778928764 Chr.6: 73362929-73362929 on Build GRCh38
rs770253703 Chr.6: 73362686-73362686 on Build GRCh38
rs993114329 Chr.6: 73363033-73363033 on Build GRCh38
rs749778733 Chr.6: 73362925-73362925 on Build GRCh38
rs372943893 Chr.6: 73362760-73362760 on Build GRCh38
rs199603559 Chr.6: 73362883-73362883 on Build GRCh38
rs368756507 Chr.6: 73362685-73362685 on Build GRCh38
rs992667093 Chr.6: 73362778-73362778 on Build GRCh38
rs371916561 Chr.6: 73362721-73362721 on Build GRCh38
rs953883035 Chr.6: 73362758-73362758 on Build GRCh38
rs879587586 Chr.6: 73362736-73362736 on Build GRCh38
rs771473640 Chr.6: 73362926-73362926 on Build GRCh38
rs143024400 Chr.6: 73362891-73362891 on Build GRCh38
rs778156260 Chr.6: 73362806-73362806 on Build GRCh38
rs954877750 Chr.6: 73362666-73362666 on Build GRCh38
rs531574387 Chr.6: 73363013-73363013 on Build GRCh38
rs560452001 Chr.6: 73362912-73362912 on Build GRCh38
rs766529626 Chr.6: 73362725-73362725 on Build GRCh38
rs1021767604 Chr.6: 73362991-73362991 on Build GRCh38
rs1022527171 Chr.6: 73362674-73362674 on Build GRCh38
rs757863032 Chr.6: 73362752-73362752 on Build GRCh38
rs192258859 Chr.6: 73362744-73362744 on Build GRCh38
rs778501993 Chr.6: 73362667-73362667 on Build GRCh38
rs948834870 Chr.6: 73362812-73362812 on Build GRCh38
rs535114156 Chr.6: 73362662-73362662 on Build GRCh38
rs775593039 Chr.6: 73363044-73363044 on Build GRCh38
rs376214417 Chr.6: 73363062-73363062 on Build GRCh38
rs549596218 Chr.6: 73363020-73363020 on Build GRCh38
rs761576316 Chr.6: 73362843-73362843 on Build GRCh38
rs772272761 Chr.6: 73362948-73362948 on Build GRCh38
rs80203576 Chr.6: 73362654-73362654 on Build GRCh38
rs606231235 Chr.6: 73362730-73362730 on Build GRCh38
rs968377813 Chr.6: 73363001-73363001 on Build GRCh38
rs996620238 Chr.6: 73362987-73362987 on Build GRCh38
rs781673395 Chr.6: 73362769-73362769 on Build GRCh38
rs75347618 Chr.6: 73362748-73362748 on Build GRCh38
rs369951888 Chr.6: 73362745-73362745 on Build GRCh38
rs577251355 Chr.6: 73363057-73363057 on Build GRCh38
rs765167528 Chr.6: 73362849-73362849 on Build GRCh38
rs1003543000 Chr.6: 73362645-73362645 on Build GRCh38
rs751166286 Chr.6: 73362884-73362884 on Build GRCh38
rs929269698 Chr.6: 73362895-73362895 on Build GRCh38
rs754813321 Chr.6: 73362651-73362651 on Build GRCh38
rs916019714 Chr.6: 73362784-73362784 on Build GRCh38
rs751290906 Chr.6: 73362756-73362756 on Build GRCh38
rs148734762 Chr.6: 73362753-73362753 on Build GRCh38
rs773337596 Chr.6: 73362694-73362694 on Build GRCh38
rs1019814442 Chr.6: 73362706-73362706 on Build GRCh38
rs606231233 Chr.6: 73362732-73362732 on Build GRCh38
rs147578781 Chr.6: 73362822-73362822 on Build GRCh38
rs900631294 Chr.6: 73362985-73362985 on Build GRCh38
rs568175839 Chr.6: 73363040-73363040 on Build GRCh38
rs748502124 Chr.6: 73362795-73362795 on Build GRCh38
rs774559307 Chr.6: 73362731-73362731 on Build GRCh38
rs753622103 Chr.6: 73362742-73362742 on Build GRCh38
rs749321389 Chr.6: 73362809-73362809 on Build GRCh38
rs895246177 Chr.6: 73362983-73362983 on Build GRCh38
rs149498621 Chr.6: 73362737-73362737 on Build GRCh38
rs959944700 Chr.6: 73363025-73363025 on Build GRCh38
rs999795233 Chr.6: 73362710-73362710 on Build GRCh38
rs767338149 Chr.6: 73362759-73362760 on Build GRCh38
rs527591826 Chr.6: 73362913-73362913 on Build GRCh38
rs750001427 Chr.6: 73363020-73363022 on Build GRCh38
rs770321628 Chr.6: 73362802-73362802 on Build GRCh38
rs746009624 Chr.6: 73362946-73362946 on Build GRCh38
rs370624464 Chr.6: 73362827-73362827 on Build GRCh38
rs9446890 Chr.6: 73362958-73362958 on Build GRCh38
rs977043184 Chr.6: 73363008-73363008 on Build GRCh38
rs201914230 Chr.6: 73363049-73363049 on Build GRCh38
rs759991776 Chr.6: 73362810-73362810 on Build GRCh38
rs897180339 Chr.6: 73362610-73362610 on Build GRCh38

View Full Product Details