Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73400027-73400558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX43
Entrez Gene ID
55510
Gene Name
DEAD-box helicase 43
Gene Aliases
CT13, HAGE
UniGene
Hs.125507
Gene Chromosome Location
Chr.6: 73394562-73417566 on Build GRCh38
Transcripts
NM_018665.2 XM_011535926.1 XM_011535927.2 XM_011535928.2 XM_017011003.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1445828 c.343T>G p.F115V Chr.6: 73400270-73400270 on Build GRCh38
6641855 c.366G>T p.T122T Chr.6: 73400293-73400293 on Build GRCh38
3629687 c.366G>A p.T122T Chr.6: 73400293-73400293 on Build GRCh38
6641844 c.429C>T p.C143C Chr.6: 73400356-73400356 on Build GRCh38
3629688 c.431G>A p.G144E Chr.6: 73400358-73400358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139888487 Chr.6: 73400257-73400257 on Build GRCh38
rs759816988 Chr.6: 73400411-73400411 on Build GRCh38
rs751858556 Chr.6: 73400257-73400259 on Build GRCh38
rs771115450 Chr.6: 73400394-73400394 on Build GRCh38
rs778716709 Chr.6: 73400313-73400313 on Build GRCh38
rs867983264 Chr.6: 73400072-73400072 on Build GRCh38
rs768985390 Chr.6: 73400241-73400241 on Build GRCh38
rs773285227 Chr.6: 73400266-73400266 on Build GRCh38
rs191599374 Chr.6: 73400284-73400284 on Build GRCh38
rs112342050 Chr.6: 73400383-73400383 on Build GRCh38
rs758679939 Chr.6: 73400185-73400185 on Build GRCh38
rs763133859 Chr.6: 73400272-73400272 on Build GRCh38
rs962351288 Chr.6: 73400245-73400245 on Build GRCh38
rs753598699 Chr.6: 73400415-73400415 on Build GRCh38
rs757925559 Chr.6: 73400324-73400324 on Build GRCh38
rs1043949848 Chr.6: 73400429-73400429 on Build GRCh38
rs141778577 Chr.6: 73400464-73400464 on Build GRCh38
rs970313056 Chr.6: 73400218-73400218 on Build GRCh38
rs3757106 Chr.6: 73400489-73400489 on Build GRCh38
rs143268677 Chr.6: 73400265-73400265 on Build GRCh38
rs112063528 Chr.6: 73400357-73400357 on Build GRCh38
rs777525978 Chr.6: 73400215-73400215 on Build GRCh38
rs1022767963 Chr.6: 73400216-73400216 on Build GRCh38
rs776901081 Chr.6: 73400250-73400250 on Build GRCh38
rs983061443 Chr.6: 73400333-73400333 on Build GRCh38
rs779742597 Chr.6: 73400394-73400394 on Build GRCh38
rs755781085 Chr.6: 73400284-73400284 on Build GRCh38
rs199574627 Chr.6: 73400389-73400389 on Build GRCh38
rs574240075 Chr.6: 73400060-73400060 on Build GRCh38
rs752478523 Chr.6: 73400201-73400201 on Build GRCh38
rs1056515919 Chr.6: 73400174-73400174 on Build GRCh38
rs749303498 Chr.6: 73400447-73400447 on Build GRCh38
rs140241372 Chr.6: 73400293-73400293 on Build GRCh38
rs182480480 Chr.6: 73400384-73400384 on Build GRCh38
rs766723144 Chr.6: 73400187-73400187 on Build GRCh38
rs757156103 Chr.6: 73400309-73400309 on Build GRCh38
rs146682107 Chr.6: 73400290-73400290 on Build GRCh38
rs267601111 Chr.6: 73400256-73400256 on Build GRCh38
rs746631522 Chr.6: 73400346-73400346 on Build GRCh38
rs368151627 Chr.6: 73400498-73400498 on Build GRCh38
rs756006388 Chr.6: 73400207-73400207 on Build GRCh38
rs763942872 Chr.6: 73400287-73400287 on Build GRCh38
rs775154392 Chr.6: 73400230-73400230 on Build GRCh38
rs997694902 Chr.6: 73400180-73400180 on Build GRCh38
rs112589417 Chr.6: 73400222-73400222 on Build GRCh38
rs779586725 Chr.6: 73400343-73400343 on Build GRCh38
rs745768493 Chr.6: 73400381-73400381 on Build GRCh38
rs745379819 Chr.6: 73400225-73400225 on Build GRCh38
rs374678810 Chr.6: 73400232-73400232 on Build GRCh38
rs374879594 Chr.6: 73400350-73400350 on Build GRCh38
rs200562580 Chr.6: 73400306-73400306 on Build GRCh38
rs749895377 Chr.6: 73400321-73400321 on Build GRCh38
rs999516442 Chr.6: 73400108-73400108 on Build GRCh38
rs755304318 Chr.6: 73400285-73400285 on Build GRCh38
rs946885235 Chr.6: 73400400-73400400 on Build GRCh38
rs747323658 Chr.6: 73400239-73400239 on Build GRCh38
rs911588171 Chr.6: 73400356-73400356 on Build GRCh38
rs201151024 Chr.6: 73400159-73400159 on Build GRCh38
rs774636655 Chr.6: 73400403-73400403 on Build GRCh38
rs776408319 Chr.6: 73400354-73400354 on Build GRCh38
rs901969517 Chr.6: 73400114-73400114 on Build GRCh38
rs561655846 Chr.6: 73400387-73400387 on Build GRCh38
rs764398180 Chr.6: 73400215-73400216 on Build GRCh38
rs960777478 Chr.6: 73400270-73400270 on Build GRCh38
rs186760672 Chr.6: 73400443-73400443 on Build GRCh38
rs771645231 Chr.6: 73400229-73400229 on Build GRCh38
rs935591218 Chr.6: 73400458-73400458 on Build GRCh38
rs191359925 Chr.6: 73400506-73400506 on Build GRCh38
rs370171205 Chr.6: 73400413-73400413 on Build GRCh38
rs770939328 Chr.6: 73400224-73400224 on Build GRCh38
rs547564467 Chr.6: 73400420-73400420 on Build GRCh38
rs145803970 Chr.6: 73400064-73400064 on Build GRCh38
rs766672479 Chr.6: 73400273-73400273 on Build GRCh38
rs768261109 Chr.6: 73400349-73400349 on Build GRCh38
rs757205645 Chr.6: 73400223-73400223 on Build GRCh38
rs530834767 Chr.6: 73400314-73400314 on Build GRCh38
rs901856492 Chr.6: 73400456-73400456 on Build GRCh38
rs1006410866 Chr.6: 73400508-73400508 on Build GRCh38

View Full Product Details