Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75308307-75308849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FILIP1
Entrez Gene ID
27145
Gene Name
filamin A interacting protein 1
Gene Aliases
FILIP
UniGene
Hs.696158
Gene Chromosome Location
Chr.6: 75294895-75493829 on Build GRCh38
Transcripts
NM_001289987.1 NM_001300866.1 NM_015687.3 XM_005248713.3 XM_005248715.4 XM_011535756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5748454 c.3493+4T>C p.? Chr.6: 75308836-75308836 on Build GRCh38
7163648 c.3585A>G p.I1195M Chr.6: 75308748-75308748 on Build GRCh38
5748453 c.3497T>C p.M1166T Chr.6: 75308836-75308836 on Build GRCh38
5061947 c.3526C>A p.P1176T Chr.6: 75308807-75308807 on Build GRCh38
3175442 c.3611G>A p.S1204N Chr.6: 75308722-75308722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920912640 Chr.6: 75308677-75308677 on Build GRCh38
rs772003968 Chr.6: 75308713-75308713 on Build GRCh38
rs376160760 Chr.6: 75308646-75308646 on Build GRCh38
rs756278620 Chr.6: 75308795-75308795 on Build GRCh38
rs200732415 Chr.6: 75308703-75308703 on Build GRCh38
rs942977029 Chr.6: 75308472-75308472 on Build GRCh38
rs748920264 Chr.6: 75308700-75308700 on Build GRCh38
rs771811265 Chr.6: 75308805-75308805 on Build GRCh38
rs763807570 Chr.6: 75308675-75308675 on Build GRCh38
rs760134089 Chr.6: 75308661-75308661 on Build GRCh38
rs750754722 Chr.6: 75308700-75308700 on Build GRCh38
rs925456506 Chr.6: 75308366-75308366 on Build GRCh38
rs370874218 Chr.6: 75308469-75308469 on Build GRCh38
rs143184461 Chr.6: 75308356-75308356 on Build GRCh38
rs140845898 Chr.6: 75308337-75308337 on Build GRCh38
rs1000682949 Chr.6: 75308349-75308349 on Build GRCh38
rs777887172 Chr.6: 75308798-75308798 on Build GRCh38
rs986778338 Chr.6: 75308426-75308426 on Build GRCh38
rs779900343 Chr.6: 75308672-75308673 on Build GRCh38
rs986742538 Chr.6: 75308768-75308768 on Build GRCh38
rs776219952 Chr.6: 75308821-75308821 on Build GRCh38
rs562704815 Chr.6: 75308451-75308451 on Build GRCh38
rs143331565 Chr.6: 75308760-75308760 on Build GRCh38
rs780551073 Chr.6: 75308709-75308709 on Build GRCh38
rs781184540 Chr.6: 75308789-75308789 on Build GRCh38
rs375290523 Chr.6: 75308781-75308781 on Build GRCh38
rs952553713 Chr.6: 75308492-75308492 on Build GRCh38
rs76996233 Chr.6: 75308583-75308583 on Build GRCh38
rs768717241 Chr.6: 75308816-75308816 on Build GRCh38
rs577167054 Chr.6: 75308434-75308434 on Build GRCh38
rs753590213 Chr.6: 75308676-75308676 on Build GRCh38
rs574738063 Chr.6: 75308497-75308497 on Build GRCh38
rs766890609 Chr.6: 75308774-75308774 on Build GRCh38
rs905311100 Chr.6: 75308440-75308440 on Build GRCh38
rs1002264347 Chr.6: 75308465-75308465 on Build GRCh38
rs559194990 Chr.6: 75308433-75308433 on Build GRCh38
rs138460165 Chr.6: 75308705-75308705 on Build GRCh38
rs936801792 Chr.6: 75308369-75308369 on Build GRCh38
rs544508334 Chr.6: 75308435-75308435 on Build GRCh38
rs775329549 Chr.6: 75308718-75308718 on Build GRCh38
rs747006280 Chr.6: 75308813-75308813 on Build GRCh38
rs201566310 Chr.6: 75308647-75308647 on Build GRCh38
rs768079202 Chr.6: 75308760-75308761 on Build GRCh38
rs745645384 Chr.6: 75308799-75308799 on Build GRCh38
rs145901023 Chr.6: 75308704-75308704 on Build GRCh38
rs752199966 Chr.6: 75308650-75308650 on Build GRCh38
rs965665947 Chr.6: 75308669-75308669 on Build GRCh38
rs112839775 Chr.6: 75308400-75308400 on Build GRCh38
rs750762407 Chr.6: 75308765-75308765 on Build GRCh38
rs1050605779 Chr.6: 75308357-75308357 on Build GRCh38
rs1004411356 Chr.6: 75308540-75308540 on Build GRCh38
rs763750918 Chr.6: 75308744-75308744 on Build GRCh38
rs748241916 Chr.6: 75308793-75308793 on Build GRCh38
rs775482186 Chr.6: 75308809-75308809 on Build GRCh38
rs185802665 Chr.6: 75308335-75308335 on Build GRCh38
rs905214360 Chr.6: 75308824-75308824 on Build GRCh38
rs1038498313 Chr.6: 75308491-75308491 on Build GRCh38
rs73453730 Chr.6: 75308468-75308468 on Build GRCh38
rs896253621 Chr.6: 75308620-75308620 on Build GRCh38
rs776414914 Chr.6: 75308748-75308748 on Build GRCh38
rs373457338 Chr.6: 75308778-75308778 on Build GRCh38
rs554786241 Chr.6: 75308390-75308390 on Build GRCh38
rs556119817 Chr.6: 75308555-75308555 on Build GRCh38
rs868766269 Chr.6: 75308561-75308561 on Build GRCh38
rs796442315 Chr.6: 75308686-75308687 on Build GRCh38
rs533974355 Chr.6: 75308407-75308407 on Build GRCh38
rs757664489 Chr.6: 75308652-75308652 on Build GRCh38
rs1047212581 Chr.6: 75308608-75308608 on Build GRCh38
rs148016136 Chr.6: 75308773-75308773 on Build GRCh38
rs770772524 Chr.6: 75308643-75308643 on Build GRCh38
rs531595086 Chr.6: 75308756-75308756 on Build GRCh38
rs368516482 Chr.6: 75308724-75308724 on Build GRCh38
rs955534453 Chr.6: 75308418-75308418 on Build GRCh38
rs758893145 Chr.6: 75308702-75308702 on Build GRCh38

View Full Product Details