Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75315047-75315574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FILIP1
Entrez Gene ID
27145
Gene Name
filamin A interacting protein 1
Gene Aliases
FILIP
UniGene
Hs.696158
Gene Chromosome Location
Chr.6: 75294895-75493829 on Build GRCh38
Transcripts
NM_001289987.1 NM_001300866.1 NM_015687.3 XM_005248713.3 XM_005248715.4 XM_011535756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3922179 c.775G>A p.E259K Chr.6: 75315057-75315057 on Build GRCh38
268392 c.754G>A p.D252N Chr.6: 75315078-75315078 on Build GRCh38
4912079 c.778C>T p.Q260* Chr.6: 75315054-75315054 on Build GRCh38
484361 c.740C>A p.A247E Chr.6: 75315092-75315092 on Build GRCh38
7368299 c.695G>A p.R232Q Chr.6: 75315137-75315137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757888492 Chr.6: 75315399-75315399 on Build GRCh38
rs150628166 Chr.6: 75315190-75315190 on Build GRCh38
rs764835248 Chr.6: 75315161-75315161 on Build GRCh38
rs752410890 Chr.6: 75315291-75315291 on Build GRCh38
rs71561424 Chr.6: 75315194-75315194 on Build GRCh38
rs905935862 Chr.6: 75315506-75315506 on Build GRCh38
rs373722500 Chr.6: 75315126-75315126 on Build GRCh38
rs753365729 Chr.6: 75315149-75315149 on Build GRCh38
rs756860523 Chr.6: 75315151-75315151 on Build GRCh38
rs1015383347 Chr.6: 75315505-75315505 on Build GRCh38
rs1052507040 Chr.6: 75315314-75315314 on Build GRCh38
rs1020473217 Chr.6: 75315546-75315546 on Build GRCh38
rs149636488 Chr.6: 75315445-75315445 on Build GRCh38
rs762079751 Chr.6: 75315099-75315099 on Build GRCh38
rs750776513 Chr.6: 75315162-75315162 on Build GRCh38
rs777307438 Chr.6: 75315439-75315439 on Build GRCh38
rs982439252 Chr.6: 75315493-75315493 on Build GRCh38
rs745674282 Chr.6: 75315247-75315247 on Build GRCh38
rs759690675 Chr.6: 75315217-75315217 on Build GRCh38
rs1050672197 Chr.6: 75315252-75315252 on Build GRCh38
rs936646228 Chr.6: 75315290-75315290 on Build GRCh38
rs771700997 Chr.6: 75315253-75315253 on Build GRCh38
rs367834294 Chr.6: 75315191-75315191 on Build GRCh38
rs759301903 Chr.6: 75315118-75315118 on Build GRCh38
rs988800815 Chr.6: 75315516-75315516 on Build GRCh38
rs952521528 Chr.6: 75315478-75315478 on Build GRCh38
rs773367637 Chr.6: 75315235-75315235 on Build GRCh38
rs776864651 Chr.6: 75315089-75315089 on Build GRCh38
rs146166109 Chr.6: 75315510-75315510 on Build GRCh38
rs540515856 Chr.6: 75315508-75315508 on Build GRCh38
rs748279846 Chr.6: 75315198-75315198 on Build GRCh38
rs530349181 Chr.6: 75315514-75315514 on Build GRCh38
rs374036208 Chr.6: 75315339-75315339 on Build GRCh38
rs575611903 Chr.6: 75315300-75315300 on Build GRCh38
rs778538012 Chr.6: 75315223-75315223 on Build GRCh38
rs758746237 Chr.6: 75315169-75315169 on Build GRCh38
rs61300362 Chr.6: 75315549-75315549 on Build GRCh38
rs180746912 Chr.6: 75315455-75315455 on Build GRCh38
rs1038537103 Chr.6: 75315271-75315272 on Build GRCh38
rs577000841 Chr.6: 75315544-75315544 on Build GRCh38
rs866664915 Chr.6: 75315098-75315098 on Build GRCh38
rs544411358 Chr.6: 75315146-75315146 on Build GRCh38
rs12164038 Chr.6: 75315080-75315080 on Build GRCh38
rs892537679 Chr.6: 75315346-75315346 on Build GRCh38
rs376789528 Chr.6: 75315216-75315216 on Build GRCh38
rs910957863 Chr.6: 75315221-75315221 on Build GRCh38
rs952377712 Chr.6: 75315143-75315143 on Build GRCh38
rs780605345 Chr.6: 75315185-75315185 on Build GRCh38
rs747071603 Chr.6: 75315078-75315078 on Build GRCh38
rs904994835 Chr.6: 75315437-75315437 on Build GRCh38
rs562708916 Chr.6: 75315309-75315309 on Build GRCh38
rs539166309 Chr.6: 75315116-75315116 on Build GRCh38
rs74842817 Chr.6: 75315411-75315411 on Build GRCh38
rs1033075254 Chr.6: 75315509-75315509 on Build GRCh38
rs200258569 Chr.6: 75315138-75315138 on Build GRCh38
rs766157987 Chr.6: 75315087-75315087 on Build GRCh38
rs932244987 Chr.6: 75315226-75315226 on Build GRCh38
rs770037594 Chr.6: 75315220-75315220 on Build GRCh38
rs200643558 Chr.6: 75315137-75315137 on Build GRCh38
rs139648537 Chr.6: 75315140-75315140 on Build GRCh38
rs573505909 Chr.6: 75315476-75315476 on Build GRCh38
rs769942534 Chr.6: 75315105-75315105 on Build GRCh38

View Full Product Details