Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 80167561-80168004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCKDHB
Entrez Gene ID
594
Gene Name
branched chain keto acid dehydrogenase E1 subunit beta
Gene Aliases
BCKDE1B, BCKDH E1-beta, E1B
UniGene
Hs.654441
Gene Chromosome Location
Chr.6: 80106610-80469088 on Build GRCh38
Transcripts
NM_000056.4 NM_001318975.1 NM_183050.3 NR_134945.1 XM_005248756.4 XM_011536023.2 XM_011536024.2 XM_011536025.2 XM_011536026.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5484871 c.372G>A p.L124L Chr.6: 80167706-80167706 on Build GRCh38
6610573 c.427G>A p.A143T Chr.6: 80167761-80167761 on Build GRCh38
5964347 c.410C>T p.A137V Chr.6: 80167744-80167744 on Build GRCh38
5163020 c.402C>T p.I134I Chr.6: 80167736-80167736 on Build GRCh38
1081945 c.435G>A p.A145A Chr.6: 80167769-80167769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs398124574 Chr.6: 80167677-80167677 on Build GRCh38
rs934559515 Chr.6: 80167727-80167727 on Build GRCh38
rs779594181 Chr.6: 80167650-80167650 on Build GRCh38
rs574122134 Chr.6: 80167768-80167768 on Build GRCh38
rs757865717 Chr.6: 80167645-80167645 on Build GRCh38
rs781586403 Chr.6: 80167680-80167680 on Build GRCh38
rs984092629 Chr.6: 80167830-80167830 on Build GRCh38
rs869312130 Chr.6: 80167735-80167735 on Build GRCh38
rs148586471 Chr.6: 80167588-80167588 on Build GRCh38
rs765097275 Chr.6: 80167751-80167751 on Build GRCh38
rs879163635 Chr.6: 80167952-80167952 on Build GRCh38
rs958989070 Chr.6: 80167884-80167884 on Build GRCh38
rs187604700 Chr.6: 80167860-80167860 on Build GRCh38
rs73479953 Chr.6: 80167654-80167654 on Build GRCh38
rs762398486 Chr.6: 80167761-80167761 on Build GRCh38
rs905623126 Chr.6: 80167806-80167806 on Build GRCh38
rs142858149 Chr.6: 80167745-80167745 on Build GRCh38
rs1035778890 Chr.6: 80167819-80167819 on Build GRCh38
rs921986157 Chr.6: 80167867-80167867 on Build GRCh38
rs749902388 Chr.6: 80167643-80167643 on Build GRCh38
rs765953398 Chr.6: 80167764-80167764 on Build GRCh38
rs73479950 Chr.6: 80167590-80167590 on Build GRCh38
rs774472182 Chr.6: 80167726-80167726 on Build GRCh38
rs748479257 Chr.6: 80167656-80167656 on Build GRCh38
rs891015474 Chr.6: 80167703-80167703 on Build GRCh38
rs778168544 Chr.6: 80167801-80167801 on Build GRCh38
rs765214300 Chr.6: 80167632-80167632 on Build GRCh38
rs1054076746 Chr.6: 80167622-80167622 on Build GRCh38
rs879089153 Chr.6: 80167954-80167954 on Build GRCh38
rs924160428 Chr.6: 80167599-80167599 on Build GRCh38
rs974908766 Chr.6: 80167858-80167858 on Build GRCh38
rs923918870 Chr.6: 80167944-80167944 on Build GRCh38
rs772426371 Chr.6: 80167834-80167834 on Build GRCh38
rs772850899 Chr.6: 80167847-80167847 on Build GRCh38
rs112504381 Chr.6: 80167825-80167825 on Build GRCh38
rs759881863 Chr.6: 80167736-80167736 on Build GRCh38
rs974814086 Chr.6: 80167616-80167616 on Build GRCh38
rs751953459 Chr.6: 80167737-80167737 on Build GRCh38
rs770996331 Chr.6: 80167712-80167712 on Build GRCh38
rs745934384 Chr.6: 80167828-80167828 on Build GRCh38
rs757318276 Chr.6: 80167631-80167631 on Build GRCh38
rs1002472211 Chr.6: 80167812-80167812 on Build GRCh38
rs374642431 Chr.6: 80167770-80167770 on Build GRCh38
rs867033248 Chr.6: 80167783-80167783 on Build GRCh38
rs73750071 Chr.6: 80167881-80167881 on Build GRCh38
rs370267903 Chr.6: 80167769-80167769 on Build GRCh38
rs35969420 Chr.6: 80167781-80167781 on Build GRCh38
rs112844382 Chr.6: 80167843-80167843 on Build GRCh38
rs74599962 Chr.6: 80167750-80167750 on Build GRCh38
rs1011884367 Chr.6: 80167941-80167941 on Build GRCh38
rs748499543 Chr.6: 80167683-80167683 on Build GRCh38
rs749766896 Chr.6: 80167802-80167802 on Build GRCh38
rs775794152 Chr.6: 80167836-80167836 on Build GRCh38
rs867276414 Chr.6: 80167797-80167797 on Build GRCh38
rs369090338 Chr.6: 80167854-80167854 on Build GRCh38
rs776631396 Chr.6: 80167744-80167744 on Build GRCh38
rs770355362 Chr.6: 80167694-80167694 on Build GRCh38
rs754593273 Chr.6: 80167670-80167670 on Build GRCh38
rs572122909 Chr.6: 80167872-80167872 on Build GRCh38
rs762688386 Chr.6: 80167600-80167600 on Build GRCh38
rs398124575 Chr.6: 80167699-80167699 on Build GRCh38
rs749343269 Chr.6: 80167700-80167700 on Build GRCh38
rs987601673 Chr.6: 80167938-80167938 on Build GRCh38
rs933267402 Chr.6: 80167655-80167655 on Build GRCh38
rs935384723 Chr.6: 80167972-80167972 on Build GRCh38
rs750324428 Chr.6: 80167760-80167760 on Build GRCh38
rs121965005 Chr.6: 80167690-80167690 on Build GRCh38
rs936918468 Chr.6: 80167604-80167604 on Build GRCh38

View Full Product Details