Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 80272905-80273407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCKDHB
Entrez Gene ID
594
Gene Name
branched chain keto acid dehydrogenase E1 subunit beta
Gene Aliases
BCKDE1B, BCKDH E1-beta, E1B
UniGene
Hs.654441
Gene Chromosome Location
Chr.6: 80106610-80469088 on Build GRCh38
Transcripts
NM_000056.4 NM_001318975.1 NM_183050.3 NR_134945.1 XM_005248756.4 XM_011536023.2 XM_011536024.2 XM_011536025.2 XM_011536026.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
270688 c.971G>A p.R324Q Chr.6: 80273154-80273154 on Build GRCh38
1446102 c.1016C>T p.S339L Chr.6: 80273199-80273199 on Build GRCh38
1446101 c.970C>A p.R324R Chr.6: 80273153-80273153 on Build GRCh38
5177003 c.1028C>T p.S343F Chr.6: 80273211-80273211 on Build GRCh38
1081953 c.1005C>T p.G335G Chr.6: 80273188-80273188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139758968 Chr.6: 80273339-80273339 on Build GRCh38
rs767009413 Chr.6: 80273116-80273116 on Build GRCh38
rs750591348 Chr.6: 80273234-80273234 on Build GRCh38
rs533298369 Chr.6: 80273161-80273161 on Build GRCh38
rs890103324 Chr.6: 80273116-80273117 on Build GRCh38
rs398124604 Chr.6: 80273157-80273157 on Build GRCh38
rs372013960 Chr.6: 80273144-80273144 on Build GRCh38
rs773855234 Chr.6: 80273200-80273200 on Build GRCh38
rs973553024 Chr.6: 80273052-80273052 on Build GRCh38
rs557193350 Chr.6: 80273340-80273340 on Build GRCh38
rs769446376 Chr.6: 80273172-80273172 on Build GRCh38
rs138670449 Chr.6: 80273170-80273170 on Build GRCh38
rs113577567 Chr.6: 80273308-80273308 on Build GRCh38
rs759040971 Chr.6: 80273208-80273208 on Build GRCh38
rs753170463 Chr.6: 80273143-80273143 on Build GRCh38
rs761765227 Chr.6: 80273186-80273186 on Build GRCh38
rs148902222 Chr.6: 80273215-80273215 on Build GRCh38
rs150651504 Chr.6: 80273321-80273321 on Build GRCh38
rs1042771488 Chr.6: 80273195-80273195 on Build GRCh38
rs755178361 Chr.6: 80273264-80273264 on Build GRCh38
rs761138547 Chr.6: 80273214-80273214 on Build GRCh38
rs398124561 Chr.6: 80273199-80273199 on Build GRCh38
rs869312131 Chr.6: 80273147-80273147 on Build GRCh38
rs759198446 Chr.6: 80273109-80273109 on Build GRCh38
rs879506080 Chr.6: 80272974-80272974 on Build GRCh38
rs931968192 Chr.6: 80273081-80273081 on Build GRCh38
rs34844836 Chr.6: 80273321-80273322 on Build GRCh38
rs758386968 Chr.6: 80273241-80273241 on Build GRCh38
rs570748969 Chr.6: 80273192-80273192 on Build GRCh38
rs116771533 Chr.6: 80273303-80273303 on Build GRCh38
rs774981415 Chr.6: 80273213-80273213 on Build GRCh38
rs141838243 Chr.6: 80273267-80273267 on Build GRCh38
rs73463529 Chr.6: 80273248-80273248 on Build GRCh38
rs796051939 Chr.6: 80273205-80273205 on Build GRCh38
rs539676910 Chr.6: 80273315-80273315 on Build GRCh38
rs563589042 Chr.6: 80272938-80272938 on Build GRCh38
rs749246466 Chr.6: 80273181-80273181 on Build GRCh38
rs772951591 Chr.6: 80273179-80273179 on Build GRCh38
rs779150907 Chr.6: 80273154-80273154 on Build GRCh38
rs534395254 Chr.6: 80273313-80273313 on Build GRCh38
rs765905155 Chr.6: 80273087-80273087 on Build GRCh38
rs936491510 Chr.6: 80272963-80272963 on Build GRCh38
rs776270090 Chr.6: 80273206-80273206 on Build GRCh38
rs1040842793 Chr.6: 80273169-80273169 on Build GRCh38
rs973988775 Chr.6: 80273329-80273329 on Build GRCh38
rs926058880 Chr.6: 80273324-80273324 on Build GRCh38
rs764707388 Chr.6: 80273152-80273152 on Build GRCh38
rs751761225 Chr.6: 80273252-80273252 on Build GRCh38
rs559388556 Chr.6: 80272997-80272997 on Build GRCh38
rs565119070 Chr.6: 80273096-80273096 on Build GRCh38
rs953605887 Chr.6: 80273361-80273361 on Build GRCh38
rs16891628 Chr.6: 80273298-80273298 on Build GRCh38
rs528490227 Chr.6: 80273006-80273006 on Build GRCh38
rs931831898 Chr.6: 80273366-80273366 on Build GRCh38
rs748534284 Chr.6: 80273102-80273102 on Build GRCh38
rs904277226 Chr.6: 80273197-80273197 on Build GRCh38
rs398124560 Chr.6: 80273189-80273189 on Build GRCh38
rs996013555 Chr.6: 80273048-80273048 on Build GRCh38
rs762419044 Chr.6: 80273223-80273223 on Build GRCh38
rs960268058 Chr.6: 80273295-80273295 on Build GRCh38
rs772386574 Chr.6: 80273188-80273189 on Build GRCh38
rs762215900 Chr.6: 80273321-80273321 on Build GRCh38
rs527503644 Chr.6: 80273353-80273357 on Build GRCh38
rs398124602 Chr.6: 80273134-80273134 on Build GRCh38
rs1050398834 Chr.6: 80273085-80273085 on Build GRCh38
rs398124603 Chr.6: 80273153-80273153 on Build GRCh38
rs9341811 Chr.6: 80272984-80272984 on Build GRCh38
rs112736231 Chr.6: 80272977-80272977 on Build GRCh38
rs569868468 Chr.6: 80273226-80273226 on Build GRCh38
rs545413403 Chr.6: 80273069-80273069 on Build GRCh38

View Full Product Details