Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87702143-87702635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKIRIN2
Entrez Gene ID
55122
Gene Name
akirin 2
Gene Aliases
C6orf166, FBI1, dJ486L4.2
UniGene
Hs.485915
Gene Chromosome Location
Chr.6: 87674860-87702267 on Build GRCh38
Transcripts
NM_018064.3

SNP Detail

SNP ID Chromosome Location
rs949477455 Chr.6: 87702319-87702320 on Build GRCh38
rs146893469 Chr.6: 87702586-87702586 on Build GRCh38
rs28384515 Chr.6: 87702489-87702489 on Build GRCh38
rs909929932 Chr.6: 87702205-87702205 on Build GRCh38
rs573214846 Chr.6: 87702243-87702243 on Build GRCh38
rs998326744 Chr.6: 87702474-87702474 on Build GRCh38
rs373733514 Chr.6: 87702404-87702404 on Build GRCh38
rs562450785 Chr.6: 87702377-87702377 on Build GRCh38
rs977405326 Chr.6: 87702245-87702245 on Build GRCh38
rs979626675 Chr.6: 87702263-87702263 on Build GRCh38
rs558358656 Chr.6: 87702365-87702365 on Build GRCh38
rs1030000609 Chr.6: 87702394-87702394 on Build GRCh38
rs965641506 Chr.6: 87702241-87702241 on Build GRCh38
rs530698871 Chr.6: 87702272-87702272 on Build GRCh38
rs527564866 Chr.6: 87702534-87702534 on Build GRCh38
rs193183154 Chr.6: 87702209-87702209 on Build GRCh38
rs557214864 Chr.6: 87702208-87702208 on Build GRCh38
rs925507622 Chr.6: 87702504-87702505 on Build GRCh38
rs894794251 Chr.6: 87702218-87702218 on Build GRCh38
rs115813023 Chr.6: 87702239-87702239 on Build GRCh38
rs773353861 Chr.6: 87702592-87702592 on Build GRCh38
rs944242316 Chr.6: 87702176-87702176 on Build GRCh38
rs1054839826 Chr.6: 87702217-87702217 on Build GRCh38
rs954292362 Chr.6: 87702498-87702498 on Build GRCh38
rs74987104 Chr.6: 87702195-87702195 on Build GRCh38
rs749247599 Chr.6: 87702202-87702202 on Build GRCh38
rs560226279 Chr.6: 87702514-87702514 on Build GRCh38
rs1010536076 Chr.6: 87702225-87702225 on Build GRCh38
rs925307263 Chr.6: 87702275-87702275 on Build GRCh38
rs939876146 Chr.6: 87702206-87702206 on Build GRCh38
rs1015949572 Chr.6: 87702232-87702232 on Build GRCh38
rs896600756 Chr.6: 87702290-87702290 on Build GRCh38
rs942975642 Chr.6: 87702262-87702262 on Build GRCh38
rs895022389 Chr.6: 87702373-87702373 on Build GRCh38
rs977544827 Chr.6: 87702167-87702167 on Build GRCh38
rs1003399962 Chr.6: 87702213-87702213 on Build GRCh38
rs1052623 Chr.6: 87702172-87702172 on Build GRCh38
rs541223516 Chr.6: 87702224-87702224 on Build GRCh38
rs954478101 Chr.6: 87702244-87702244 on Build GRCh38
rs1029011498 Chr.6: 87702477-87702477 on Build GRCh38
rs142540829 Chr.6: 87702585-87702585 on Build GRCh38
rs921493665 Chr.6: 87702247-87702247 on Build GRCh38
rs903851789 Chr.6: 87702166-87702166 on Build GRCh38
rs370153786 Chr.6: 87702546-87702546 on Build GRCh38
rs1022463659 Chr.6: 87702233-87702233 on Build GRCh38
rs769067253 Chr.6: 87702330-87702330 on Build GRCh38
rs940098276 Chr.6: 87702283-87702283 on Build GRCh38
rs909970959 Chr.6: 87702278-87702278 on Build GRCh38
rs906490891 Chr.6: 87702210-87702210 on Build GRCh38
rs527452655 Chr.6: 87702163-87702163 on Build GRCh38
rs544283693 Chr.6: 87702366-87702366 on Build GRCh38
rs938781065 Chr.6: 87702237-87702237 on Build GRCh38
rs757255487 Chr.6: 87702313-87702314 on Build GRCh38
rs1036851818 Chr.6: 87702342-87702342 on Build GRCh38
rs1018872785 Chr.6: 87702468-87702468 on Build GRCh38
rs190117179 Chr.6: 87702319-87702319 on Build GRCh38
rs965526930 Chr.6: 87702472-87702472 on Build GRCh38
rs966424183 Chr.6: 87702234-87702234 on Build GRCh38
rs185256635 Chr.6: 87702238-87702238 on Build GRCh38
rs555518258 Chr.6: 87702363-87702363 on Build GRCh38
rs984116029 Chr.6: 87702277-87702277 on Build GRCh38
rs976777637 Chr.6: 87702188-87702188 on Build GRCh38
rs147711957 Chr.6: 87702516-87702516 on Build GRCh38
rs143718343 Chr.6: 87702442-87702442 on Build GRCh38
rs993297695 Chr.6: 87702391-87702391 on Build GRCh38
rs954188993 Chr.6: 87702269-87702269 on Build GRCh38
rs961482997 Chr.6: 87702578-87702578 on Build GRCh38
rs747370988 Chr.6: 87702294-87702294 on Build GRCh38
rs148950888 Chr.6: 87702354-87702354 on Build GRCh38
rs911377158 Chr.6: 87702248-87702248 on Build GRCh38
rs769712932 Chr.6: 87702515-87702515 on Build GRCh38
rs77622916 Chr.6: 87702204-87702204 on Build GRCh38
rs1031881803 Chr.6: 87702449-87702449 on Build GRCh38
rs1045130355 Chr.6: 87702593-87702593 on Build GRCh38
rs371992214 Chr.6: 87702507-87702508 on Build GRCh38
rs902291695 Chr.6: 87702401-87702401 on Build GRCh38
rs935580509 Chr.6: 87702532-87702532 on Build GRCh38

View Full Product Details