Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89180380-89180874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR1
Entrez Gene ID
2569
Gene Name
gamma-aminobutyric acid type A receptor rho1 subunit
Gene Aliases
-
UniGene
Hs.99927
Gene Chromosome Location
Chr.6: 89177504-89231288 on Build GRCh38
Transcripts
NM_001256703.1 NM_001256704.1 NM_001267582.1 NM_002042.4 XM_017010689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668721 c.1010G>A p.R337H Chr.6: 89180428-89180428 on Build GRCh38
1082445 c.959C>T p.T320M Chr.6: 89180479-89180479 on Build GRCh38
3178059 c.1009C>T p.R337C Chr.6: 89180429-89180429 on Build GRCh38
7106931 c.1001C>G p.S334C Chr.6: 89180437-89180437 on Build GRCh38
6668720 c.1011C>T p.R337R Chr.6: 89180427-89180427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777978678 Chr.6: 89180417-89180417 on Build GRCh38
rs192148367 Chr.6: 89180426-89180426 on Build GRCh38
rs750447301 Chr.6: 89180829-89180829 on Build GRCh38
rs1040889451 Chr.6: 89180670-89180670 on Build GRCh38
rs990103810 Chr.6: 89180680-89180680 on Build GRCh38
rs781005829 Chr.6: 89180505-89180505 on Build GRCh38
rs565116159 Chr.6: 89180758-89180758 on Build GRCh38
rs779163204 Chr.6: 89180481-89180481 on Build GRCh38
rs767876225 Chr.6: 89180458-89180458 on Build GRCh38
rs201172276 Chr.6: 89180487-89180487 on Build GRCh38
rs368010272 Chr.6: 89180461-89180461 on Build GRCh38
rs920848363 Chr.6: 89180546-89180546 on Build GRCh38
rs62414648 Chr.6: 89180712-89180712 on Build GRCh38
rs373071538 Chr.6: 89180514-89180514 on Build GRCh38
rs746432349 Chr.6: 89180430-89180430 on Build GRCh38
rs759994922 Chr.6: 89180455-89180455 on Build GRCh38
rs371702531 Chr.6: 89180408-89180408 on Build GRCh38
rs561192861 Chr.6: 89180827-89180827 on Build GRCh38
rs757338894 Chr.6: 89180479-89180479 on Build GRCh38
rs187926567 Chr.6: 89180428-89180428 on Build GRCh38
rs376086704 Chr.6: 89180478-89180478 on Build GRCh38
rs185458142 Chr.6: 89180427-89180427 on Build GRCh38
rs973988974 Chr.6: 89180837-89180837 on Build GRCh38
rs1020615212 Chr.6: 89180748-89180748 on Build GRCh38
rs770390345 Chr.6: 89180528-89180528 on Build GRCh38
rs567561851 Chr.6: 89180452-89180452 on Build GRCh38
rs145000276 Chr.6: 89180451-89180451 on Build GRCh38
rs972881325 Chr.6: 89180732-89180732 on Build GRCh38
rs893721063 Chr.6: 89180714-89180714 on Build GRCh38
rs754918547 Chr.6: 89180490-89180490 on Build GRCh38
rs371833286 Chr.6: 89180466-89180466 on Build GRCh38
rs35577195 Chr.6: 89180699-89180699 on Build GRCh38
rs578118381 Chr.6: 89180560-89180560 on Build GRCh38
rs931448567 Chr.6: 89180774-89180774 on Build GRCh38
rs970841048 Chr.6: 89180593-89180593 on Build GRCh38
rs772700915 Chr.6: 89180447-89180447 on Build GRCh38
rs756314695 Chr.6: 89180409-89180409 on Build GRCh38
rs148675578 Chr.6: 89180522-89180522 on Build GRCh38
rs867633132 Chr.6: 89180445-89180445 on Build GRCh38
rs764478832 Chr.6: 89180467-89180467 on Build GRCh38
rs545253286 Chr.6: 89180603-89180603 on Build GRCh38
rs1053669024 Chr.6: 89180700-89180700 on Build GRCh38
rs776857470 Chr.6: 89180697-89180697 on Build GRCh38
rs376401079 Chr.6: 89180429-89180429 on Build GRCh38
rs192974440 Chr.6: 89180727-89180727 on Build GRCh38
rs748947115 Chr.6: 89180526-89180526 on Build GRCh38
rs1031534147 Chr.6: 89180652-89180652 on Build GRCh38
rs755754709 Chr.6: 89180515-89180515 on Build GRCh38
rs773758178 Chr.6: 89180533-89180533 on Build GRCh38
rs407221 Chr.6: 89180656-89180656 on Build GRCh38
rs769472188 Chr.6: 89180440-89180440 on Build GRCh38
rs372297904 Chr.6: 89180463-89180463 on Build GRCh38
rs953720230 Chr.6: 89180698-89180698 on Build GRCh38
rs754011759 Chr.6: 89180419-89180419 on Build GRCh38
rs772547976 Chr.6: 89180431-89180431 on Build GRCh38
rs762512117 Chr.6: 89180448-89180448 on Build GRCh38
rs761897322 Chr.6: 89180740-89180740 on Build GRCh38

View Full Product Details