Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89199073-89199571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR1
Entrez Gene ID
2569
Gene Name
gamma-aminobutyric acid type A receptor rho1 subunit
Gene Aliases
-
UniGene
Hs.99927
Gene Chromosome Location
Chr.6: 89177504-89231288 on Build GRCh38
Transcripts
NM_001256703.1 NM_001256704.1 NM_001267582.1 NM_002042.4 XM_017010689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
743905 c.298G>T p.G100C Chr.6: 89199412-89199412 on Build GRCh38
5756905 c.289A>G p.I97V Chr.6: 89199421-89199421 on Build GRCh38
6529886 c.284C>G p.P95R Chr.6: 89199426-89199426 on Build GRCh38
6175095 c.312G>C p.Q104H Chr.6: 89199398-89199398 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs578183944 Chr.6: 89199441-89199441 on Build GRCh38
rs915529537 Chr.6: 89199191-89199191 on Build GRCh38
rs754036460 Chr.6: 89199337-89199337 on Build GRCh38
rs538547613 Chr.6: 89199256-89199256 on Build GRCh38
rs374191683 Chr.6: 89199463-89199463 on Build GRCh38
rs1048732123 Chr.6: 89199254-89199254 on Build GRCh38
rs768058445 Chr.6: 89199343-89199343 on Build GRCh38
rs766170340 Chr.6: 89199345-89199345 on Build GRCh38
rs377459490 Chr.6: 89199312-89199312 on Build GRCh38
rs747773649 Chr.6: 89199383-89199383 on Build GRCh38
rs549472603 Chr.6: 89199126-89199126 on Build GRCh38
rs939388369 Chr.6: 89199548-89199548 on Build GRCh38
rs954186212 Chr.6: 89199438-89199438 on Build GRCh38
rs751519021 Chr.6: 89199364-89199364 on Build GRCh38
rs748777031 Chr.6: 89199273-89199273 on Build GRCh38
rs565351901 Chr.6: 89199286-89199286 on Build GRCh38
rs777029761 Chr.6: 89199270-89199270 on Build GRCh38
rs770067646 Chr.6: 89199422-89199426 on Build GRCh38
rs566930673 Chr.6: 89199120-89199120 on Build GRCh38
rs770491596 Chr.6: 89199429-89199429 on Build GRCh38
rs755801569 Chr.6: 89199386-89199386 on Build GRCh38
rs945656941 Chr.6: 89199210-89199210 on Build GRCh38
rs547261363 Chr.6: 89199258-89199258 on Build GRCh38
rs954220760 Chr.6: 89199486-89199486 on Build GRCh38
rs111740397 Chr.6: 89199160-89199160 on Build GRCh38
rs146927089 Chr.6: 89199315-89199315 on Build GRCh38
rs561210292 Chr.6: 89199161-89199161 on Build GRCh38
rs149010982 Chr.6: 89199290-89199290 on Build GRCh38
rs754685769 Chr.6: 89199366-89199366 on Build GRCh38
rs55999329 Chr.6: 89199485-89199485 on Build GRCh38
rs912640944 Chr.6: 89199114-89199114 on Build GRCh38
rs531563097 Chr.6: 89199184-89199184 on Build GRCh38
rs746371349 Chr.6: 89199371-89199372 on Build GRCh38
rs771761605 Chr.6: 89199504-89199504 on Build GRCh38
rs1004152872 Chr.6: 89199395-89199395 on Build GRCh38
rs989612898 Chr.6: 89199460-89199460 on Build GRCh38
rs954217125 Chr.6: 89199107-89199107 on Build GRCh38
rs571067962 Chr.6: 89199226-89199226 on Build GRCh38
rs927940785 Chr.6: 89199495-89199495 on Build GRCh38
rs545740462 Chr.6: 89199481-89199481 on Build GRCh38
rs145851293 Chr.6: 89199100-89199100 on Build GRCh38
rs777264011 Chr.6: 89199404-89199404 on Build GRCh38
rs978378147 Chr.6: 89199491-89199491 on Build GRCh38
rs887398905 Chr.6: 89199255-89199255 on Build GRCh38
rs764248228 Chr.6: 89199336-89199336 on Build GRCh38
rs965477656 Chr.6: 89199529-89199529 on Build GRCh38
rs1020139862 Chr.6: 89199512-89199512 on Build GRCh38
rs538470381 Chr.6: 89199403-89199403 on Build GRCh38
rs576051305 Chr.6: 89199326-89199326 on Build GRCh38
rs929195289 Chr.6: 89199213-89199213 on Build GRCh38
rs549563141 Chr.6: 89199189-89199189 on Build GRCh38
rs534739356 Chr.6: 89199299-89199299 on Build GRCh38
rs772959773 Chr.6: 89199549-89199549 on Build GRCh38
rs1042741086 Chr.6: 89199240-89199240 on Build GRCh38
rs1012740109 Chr.6: 89199311-89199311 on Build GRCh38
rs572439298 Chr.6: 89199539-89199539 on Build GRCh38
rs780983775 Chr.6: 89199374-89199374 on Build GRCh38
rs1015648008 Chr.6: 89199397-89199397 on Build GRCh38
rs998325900 Chr.6: 89199415-89199415 on Build GRCh38
rs201073532 Chr.6: 89199420-89199420 on Build GRCh38
rs759765552 Chr.6: 89199314-89199314 on Build GRCh38
rs965103034 Chr.6: 89199398-89199398 on Build GRCh38

View Full Product Details