Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89271549-89272057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR2
Entrez Gene ID
2570
Gene Name
gamma-aminobutyric acid type A receptor rho2 subunit
Gene Aliases
-
UniGene
Hs.379330
Gene Chromosome Location
Chr.6: 89254464-89315299 on Build GRCh38
Transcripts
NM_002043.4 XM_011535713.2 XM_011535714.2 XM_011535715.2 XM_011535716.2 XM_011535717.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5665319 c.296-3C>T p.? Chr.6: 89271725-89271725 on Build GRCh38
3762217 c.324A>G p.V108V Chr.6: 89271694-89271694 on Build GRCh38
4357403 c.316G>A p.V106M Chr.6: 89271702-89271702 on Build GRCh38
4413226 c.246C>T p.D82D Chr.6: 89271697-89271697 on Build GRCh38
5665320 c.221-3C>T p.? Chr.6: 89271725-89271725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894165152 Chr.6: 89271887-89271907 on Build GRCh38
rs761043912 Chr.6: 89271770-89271770 on Build GRCh38
rs772652570 Chr.6: 89271645-89271645 on Build GRCh38
rs1001327458 Chr.6: 89272008-89272008 on Build GRCh38
rs1052674796 Chr.6: 89271976-89271976 on Build GRCh38
rs772423350 Chr.6: 89271759-89271759 on Build GRCh38
rs748706268 Chr.6: 89271733-89271733 on Build GRCh38
rs747786550 Chr.6: 89271719-89271719 on Build GRCh38
rs780056404 Chr.6: 89271612-89271612 on Build GRCh38
rs960378743 Chr.6: 89271942-89271942 on Build GRCh38
rs757588172 Chr.6: 89271710-89271710 on Build GRCh38
rs572646003 Chr.6: 89271934-89271934 on Build GRCh38
rs886459836 Chr.6: 89271638-89271638 on Build GRCh38
rs929755506 Chr.6: 89271601-89271601 on Build GRCh38
rs778044729 Chr.6: 89271628-89271628 on Build GRCh38
rs892484452 Chr.6: 89271977-89271977 on Build GRCh38
rs200580267 Chr.6: 89271666-89271666 on Build GRCh38
rs753805707 Chr.6: 89271698-89271698 on Build GRCh38
rs539554043 Chr.6: 89271798-89271798 on Build GRCh38
rs148711691 Chr.6: 89271686-89271686 on Build GRCh38
rs147754574 Chr.6: 89271680-89271680 on Build GRCh38
rs907047968 Chr.6: 89271786-89271786 on Build GRCh38
rs979125039 Chr.6: 89271742-89271742 on Build GRCh38
rs373888602 Chr.6: 89271624-89271624 on Build GRCh38
rs35608866 Chr.6: 89271700-89271700 on Build GRCh38
rs745815166 Chr.6: 89271608-89271608 on Build GRCh38
rs912291430 Chr.6: 89271873-89271873 on Build GRCh38
rs779819646 Chr.6: 89271702-89271702 on Build GRCh38
rs1006422019 Chr.6: 89271855-89271855 on Build GRCh38
rs566494692 Chr.6: 89271819-89271819 on Build GRCh38
rs1001585343 Chr.6: 89271918-89271918 on Build GRCh38
rs201908910 Chr.6: 89271889-89271889 on Build GRCh38
rs1013448187 Chr.6: 89271967-89271967 on Build GRCh38
rs773018276 Chr.6: 89271898-89271900 on Build GRCh38
rs905836539 Chr.6: 89271891-89271891 on Build GRCh38
rs282115 Chr.6: 89271993-89271993 on Build GRCh38
rs747677562 Chr.6: 89271850-89271850 on Build GRCh38
rs554050275 Chr.6: 89271839-89271839 on Build GRCh38
rs367685537 Chr.6: 89271609-89271609 on Build GRCh38
rs140504422 Chr.6: 89271703-89271703 on Build GRCh38
rs759844416 Chr.6: 89271678-89271678 on Build GRCh38
rs925437014 Chr.6: 89271903-89271903 on Build GRCh38
rs1034185341 Chr.6: 89272028-89272028 on Build GRCh38
rs762536064 Chr.6: 89271651-89271651 on Build GRCh38
rs367792893 Chr.6: 89271803-89271803 on Build GRCh38
rs936938255 Chr.6: 89271914-89271914 on Build GRCh38
rs758472754 Chr.6: 89271627-89271627 on Build GRCh38
rs746717830 Chr.6: 89271620-89271620 on Build GRCh38
rs970151310 Chr.6: 89271688-89271688 on Build GRCh38
rs774697752 Chr.6: 89271734-89271734 on Build GRCh38
rs756396951 Chr.6: 89271696-89271696 on Build GRCh38
rs764222221 Chr.6: 89271697-89271697 on Build GRCh38
rs760077115 Chr.6: 89271753-89271753 on Build GRCh38
rs770302267 Chr.6: 89271799-89271799 on Build GRCh38
rs933480795 Chr.6: 89271802-89271802 on Build GRCh38
rs774952454 Chr.6: 89271669-89271669 on Build GRCh38
rs1037707155 Chr.6: 89271869-89271869 on Build GRCh38
rs975538970 Chr.6: 89271899-89271899 on Build GRCh38
rs781571990 Chr.6: 89271722-89271722 on Build GRCh38
rs1046929633 Chr.6: 89271992-89271992 on Build GRCh38
rs745472082 Chr.6: 89271761-89271761 on Build GRCh38
rs150834111 Chr.6: 89271797-89271797 on Build GRCh38
rs144730602 Chr.6: 89271709-89271709 on Build GRCh38
rs945270685 Chr.6: 89271713-89271713 on Build GRCh38
rs141403078 Chr.6: 89271721-89271721 on Build GRCh38
rs979303392 Chr.6: 89272009-89272009 on Build GRCh38
rs282116 Chr.6: 89271767-89271767 on Build GRCh38
rs553709575 Chr.6: 89271911-89271911 on Build GRCh38
rs771527470 Chr.6: 89271985-89271985 on Build GRCh38
rs1039257128 Chr.6: 89271572-89271572 on Build GRCh38
rs534423050 Chr.6: 89271830-89271830 on Build GRCh38
rs282117 Chr.6: 89271694-89271694 on Build GRCh38
rs528344587 Chr.6: 89271623-89271623 on Build GRCh38
rs766728215 Chr.6: 89271662-89271662 on Build GRCh38

View Full Product Details