Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89856731-89857252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP8AP2
Entrez Gene ID
9994
Gene Name
caspase 8 associated protein 2
Gene Aliases
CED-4, FLASH, RIP25
UniGene
Hs.558218
Gene Chromosome Location
Chr.6: 89829900-89874436 on Build GRCh38
Transcripts
NM_001137667.1 NM_001137668.1 NM_012115.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3831221 c.368G>T p.R123I Chr.6: 89857151-89857151 on Build GRCh38
3831220 c.306C>T p.F102F Chr.6: 89857089-89857089 on Build GRCh38
4491537 c.381C>T p.N127N Chr.6: 89857164-89857164 on Build GRCh38
7185919 c.334A>C p.K112Q Chr.6: 89857117-89857117 on Build GRCh38
6107550 c.357C>G p.I119M Chr.6: 89857140-89857140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914931835 Chr.6: 89856916-89856916 on Build GRCh38
rs969975257 Chr.6: 89856938-89856938 on Build GRCh38
rs78957214 Chr.6: 89856964-89856964 on Build GRCh38
rs752852169 Chr.6: 89856884-89856884 on Build GRCh38
rs767371851 Chr.6: 89857176-89857176 on Build GRCh38
rs534459577 Chr.6: 89857133-89857133 on Build GRCh38
rs1042690998 Chr.6: 89856929-89856929 on Build GRCh38
rs1004824621 Chr.6: 89857110-89857110 on Build GRCh38
rs980742161 Chr.6: 89856940-89856940 on Build GRCh38
rs950573819 Chr.6: 89856900-89856900 on Build GRCh38
rs567727300 Chr.6: 89857212-89857212 on Build GRCh38
rs939541143 Chr.6: 89856932-89856932 on Build GRCh38
rs759528308 Chr.6: 89857167-89857167 on Build GRCh38
rs548197473 Chr.6: 89856949-89856949 on Build GRCh38
rs866813659 Chr.6: 89857199-89857199 on Build GRCh38
rs76588369 Chr.6: 89856931-89856931 on Build GRCh38
rs375845338 Chr.6: 89857131-89857131 on Build GRCh38
rs753565969 Chr.6: 89857216-89857216 on Build GRCh38
rs901579882 Chr.6: 89856773-89856773 on Build GRCh38
rs776010982 Chr.6: 89857093-89857093 on Build GRCh38
rs992247583 Chr.6: 89857014-89857014 on Build GRCh38
rs373042680 Chr.6: 89857225-89857225 on Build GRCh38
rs765402525 Chr.6: 89857164-89857164 on Build GRCh38
rs530637913 Chr.6: 89857011-89857011 on Build GRCh38
rs760543553 Chr.6: 89856986-89856986 on Build GRCh38
rs960746078 Chr.6: 89856953-89856953 on Build GRCh38
rs552313762 Chr.6: 89857022-89857022 on Build GRCh38
rs188884572 Chr.6: 89856862-89856862 on Build GRCh38
rs755851282 Chr.6: 89857213-89857213 on Build GRCh38
rs746454800 Chr.6: 89857038-89857038 on Build GRCh38
rs895272311 Chr.6: 89856975-89856975 on Build GRCh38
rs773164983 Chr.6: 89857166-89857166 on Build GRCh38
rs747311012 Chr.6: 89857097-89857097 on Build GRCh38
rs111244010 Chr.6: 89856922-89856922 on Build GRCh38
rs921053147 Chr.6: 89856813-89856813 on Build GRCh38
rs914012397 Chr.6: 89857075-89857075 on Build GRCh38
rs570533855 Chr.6: 89857068-89857068 on Build GRCh38
rs373992341 Chr.6: 89856774-89856774 on Build GRCh38
rs569884270 Chr.6: 89856973-89856973 on Build GRCh38
rs1026535568 Chr.6: 89856810-89856810 on Build GRCh38
rs1049587672 Chr.6: 89857064-89857064 on Build GRCh38
rs769032180 Chr.6: 89857101-89857101 on Build GRCh38
rs370454326 Chr.6: 89857134-89857134 on Build GRCh38
rs552640964 Chr.6: 89857181-89857181 on Build GRCh38
rs1015444491 Chr.6: 89856789-89856789 on Build GRCh38
rs1044314416 Chr.6: 89857211-89857211 on Build GRCh38
rs965211638 Chr.6: 89856798-89856798 on Build GRCh38

View Full Product Details