Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 93245303-93245811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA7
Entrez Gene ID
2045
Gene Name
EPH receptor A7
Gene Aliases
EHK-3, EHK3, EK11, HEK11
UniGene
Hs.73962
Gene Chromosome Location
Chr.6: 93240020-93419687 on Build GRCh38
Transcripts
NM_001288629.1 NM_004440.3 XM_005248669.2 XM_017010365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7268659 c.2869A>G p.R957G Chr.6: 93245311-93245311 on Build GRCh38
6655114 c.2858A>G p.E953G Chr.6: 93245322-93245322 on Build GRCh38
4786995 c.2788G>T p.E930* Chr.6: 93245392-93245392 on Build GRCh38
4874715 c.2772C>T p.T924T Chr.6: 93245408-93245408 on Build GRCh38
6655122 c.2758C>T p.P920S Chr.6: 93245422-93245422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018327906 Chr.6: 93245581-93245581 on Build GRCh38
rs902269577 Chr.6: 93245716-93245716 on Build GRCh38
rs530355686 Chr.6: 93245623-93245623 on Build GRCh38
rs932900354 Chr.6: 93245644-93245644 on Build GRCh38
rs1007281136 Chr.6: 93245575-93245575 on Build GRCh38
rs763374008 Chr.6: 93245380-93245380 on Build GRCh38
rs114769374 Chr.6: 93245720-93245720 on Build GRCh38
rs1003404422 Chr.6: 93245489-93245489 on Build GRCh38
rs756054614 Chr.6: 93245451-93245451 on Build GRCh38
rs542986426 Chr.6: 93245575-93245575 on Build GRCh38
rs151105732 Chr.6: 93245346-93245346 on Build GRCh38
rs752942156 Chr.6: 93245409-93245409 on Build GRCh38
rs1024889723 Chr.6: 93245787-93245787 on Build GRCh38
rs367774379 Chr.6: 93245714-93245714 on Build GRCh38
rs879735300 Chr.6: 93245605-93245605 on Build GRCh38
rs752140433 Chr.6: 93245500-93245500 on Build GRCh38
rs1020425332 Chr.6: 93245786-93245786 on Build GRCh38
rs764861058 Chr.6: 93245364-93245364 on Build GRCh38
rs1044738144 Chr.6: 93245789-93245789 on Build GRCh38
rs1044853276 Chr.6: 93245459-93245459 on Build GRCh38
rs890770968 Chr.6: 93245377-93245377 on Build GRCh38
rs866764000 Chr.6: 93245602-93245602 on Build GRCh38
rs567859207 Chr.6: 93245569-93245569 on Build GRCh38
rs530679092 Chr.6: 93245603-93245603 on Build GRCh38
rs766896709 Chr.6: 93245382-93245382 on Build GRCh38
rs538780514 Chr.6: 93245741-93245741 on Build GRCh38
rs1022601350 Chr.6: 93245563-93245563 on Build GRCh38
rs947671764 Chr.6: 93245452-93245452 on Build GRCh38
rs768600630 Chr.6: 93245477-93245477 on Build GRCh38
rs769504187 Chr.6: 93245479-93245479 on Build GRCh38
rs976861296 Chr.6: 93245345-93245345 on Build GRCh38
rs749890930 Chr.6: 93245376-93245376 on Build GRCh38
rs139990635 Chr.6: 93245413-93245413 on Build GRCh38
rs368531054 Chr.6: 93245407-93245407 on Build GRCh38
rs943717916 Chr.6: 93245707-93245707 on Build GRCh38
rs182795817 Chr.6: 93245673-93245673 on Build GRCh38
rs1032102827 Chr.6: 93245484-93245484 on Build GRCh38
rs547685753 Chr.6: 93245511-93245511 on Build GRCh38
rs1010818852 Chr.6: 93245518-93245518 on Build GRCh38
rs374288532 Chr.6: 93245331-93245331 on Build GRCh38
rs918752834 Chr.6: 93245675-93245675 on Build GRCh38
rs141581954 Chr.6: 93245328-93245328 on Build GRCh38
rs776388575 Chr.6: 93245337-93245337 on Build GRCh38
rs749385662 Chr.6: 93245415-93245415 on Build GRCh38
rs751918950 Chr.6: 93245403-93245403 on Build GRCh38
rs565168538 Chr.6: 93245586-93245586 on Build GRCh38
rs1012142808 Chr.6: 93245781-93245781 on Build GRCh38
rs912239979 Chr.6: 93245687-93245688 on Build GRCh38
rs1023415571 Chr.6: 93245551-93245551 on Build GRCh38
rs763886229 Chr.6: 93245336-93245336 on Build GRCh38
rs981972502 Chr.6: 93245571-93245571 on Build GRCh38
rs746817774 Chr.6: 93245470-93245470 on Build GRCh38
rs762503385 Chr.6: 93245494-93245494 on Build GRCh38
rs1048575750 Chr.6: 93245423-93245423 on Build GRCh38
rs1053568700 Chr.6: 93245721-93245721 on Build GRCh38
rs369268179 Chr.6: 93245689-93245690 on Build GRCh38
rs771983362 Chr.6: 93245327-93245327 on Build GRCh38
rs551414699 Chr.6: 93245482-93245482 on Build GRCh38
rs760501056 Chr.6: 93245333-93245333 on Build GRCh38
rs976912132 Chr.6: 93245671-93245671 on Build GRCh38
rs781498240 Chr.6: 93245408-93245408 on Build GRCh38
rs910100612 Chr.6: 93245762-93245762 on Build GRCh38
rs149864613 Chr.6: 93245430-93245430 on Build GRCh38
rs564553276 Chr.6: 93245493-93245493 on Build GRCh38
rs766837679 Chr.6: 93245498-93245498 on Build GRCh38
rs551033335 Chr.6: 93245478-93245478 on Build GRCh38
rs190816224 Chr.6: 93245510-93245510 on Build GRCh38
rs188957280 Chr.6: 93245772-93245772 on Build GRCh38

View Full Product Details