Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 96206431-96206969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT9
Entrez Gene ID
10690
Gene Name
fucosyltransferase 9
Gene Aliases
Fuc-TIX
UniGene
Hs.49117
Gene Chromosome Location
Chr.6: 96015969-96215612 on Build GRCh38
Transcripts
NM_006581.3 XM_011535383.2 XM_011535385.2 XM_017010188.1 XM_017010190.1

SNP Detail

SNP ID Chromosome Location
rs988046201 Chr.6: 96206852-96206852 on Build GRCh38
rs540346251 Chr.6: 96206534-96206534 on Build GRCh38
rs1053252710 Chr.6: 96206698-96206698 on Build GRCh38
rs950044474 Chr.6: 96206550-96206550 on Build GRCh38
rs911221055 Chr.6: 96206903-96206903 on Build GRCh38
rs867332131 Chr.6: 96206751-96206751 on Build GRCh38
rs531118592 Chr.6: 96206642-96206642 on Build GRCh38
rs570599769 Chr.6: 96206863-96206863 on Build GRCh38
rs769702755 Chr.6: 96206923-96206923 on Build GRCh38
rs149131395 Chr.6: 96206629-96206629 on Build GRCh38
rs548491276 Chr.6: 96206575-96206575 on Build GRCh38
rs566831406 Chr.6: 96206888-96206888 on Build GRCh38
rs979320551 Chr.6: 96206502-96206502 on Build GRCh38
rs746202102 Chr.6: 96206713-96206713 on Build GRCh38
rs919949514 Chr.6: 96206585-96206585 on Build GRCh38
rs574839680 Chr.6: 96206608-96206608 on Build GRCh38
rs986487073 Chr.6: 96206497-96206497 on Build GRCh38
rs770194123 Chr.6: 96206803-96206803 on Build GRCh38
rs575608140 Chr.6: 96206457-96206457 on Build GRCh38
rs375939430 Chr.6: 96206775-96206775 on Build GRCh38
rs934437443 Chr.6: 96206695-96206695 on Build GRCh38
rs1001102693 Chr.6: 96206806-96206806 on Build GRCh38
rs570532869 Chr.6: 96206792-96206792 on Build GRCh38
rs189732252 Chr.6: 96206673-96206673 on Build GRCh38
rs550814687 Chr.6: 96206711-96206711 on Build GRCh38
rs1011848246 Chr.6: 96206458-96206458 on Build GRCh38
rs186378254 Chr.6: 96206532-96206532 on Build GRCh38
rs1040247861 Chr.6: 96206640-96206640 on Build GRCh38
rs760216742 Chr.6: 96206740-96206740 on Build GRCh38
rs985407680 Chr.6: 96206628-96206628 on Build GRCh38
rs952784022 Chr.6: 96206889-96206889 on Build GRCh38
rs961814040 Chr.6: 96206859-96206859 on Build GRCh38
rs560322379 Chr.6: 96206647-96206647 on Build GRCh38
rs1023200577 Chr.6: 96206504-96206504 on Build GRCh38
rs961486731 Chr.6: 96206554-96206554 on Build GRCh38
rs952688916 Chr.6: 96206605-96206609 on Build GRCh38
rs1036264225 Chr.6: 96206547-96206547 on Build GRCh38
rs935301066 Chr.6: 96206531-96206531 on Build GRCh38
rs544268094 Chr.6: 96206484-96206484 on Build GRCh38
rs146552186 Chr.6: 96206558-96206558 on Build GRCh38
rs955050453 Chr.6: 96206463-96206463 on Build GRCh38
rs985458560 Chr.6: 96206898-96206898 on Build GRCh38
rs763122556 Chr.6: 96206499-96206499 on Build GRCh38
rs564082847 Chr.6: 96206511-96206511 on Build GRCh38
rs750976769 Chr.6: 96206676-96206676 on Build GRCh38
rs9373717 Chr.6: 96206929-96206929 on Build GRCh38
rs956752189 Chr.6: 96206845-96206845 on Build GRCh38
rs1044072453 Chr.6: 96206757-96206757 on Build GRCh38
rs994397903 Chr.6: 96206635-96206635 on Build GRCh38
rs543899190 Chr.6: 96206455-96206455 on Build GRCh38
rs553467911 Chr.6: 96206880-96206880 on Build GRCh38
rs374180376 Chr.6: 96206607-96206607 on Build GRCh38
rs1011272441 Chr.6: 96206715-96206715 on Build GRCh38
rs927385697 Chr.6: 96206588-96206588 on Build GRCh38
rs528711485 Chr.6: 96206571-96206571 on Build GRCh38
rs753509934 Chr.6: 96206827-96206827 on Build GRCh38
rs748434999 Chr.6: 96206630-96206630 on Build GRCh38
rs769041092 Chr.6: 96206510-96206510 on Build GRCh38
rs963829663 Chr.6: 96206501-96206501 on Build GRCh38

View Full Product Details