Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 98873929-98874436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL4
Entrez Gene ID
26235
Gene Name
F-box and leucine rich repeat protein 4
Gene Aliases
FBL4, FBL5, MTDPS13
UniGene
Hs.536850
Gene Chromosome Location
Chr.6: 98868535-98948006 on Build GRCh38
Transcripts
NM_001278716.1 NM_012160.4 NR_103836.1 XM_005266930.2 XM_017010726.1 XM_017010727.1 XM_017010728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6175003 c.1865G>T p.*622L Chr.6: 98874279-98874279 on Build GRCh38
5678615 c.1733G>T p.R578I Chr.6: 98874411-98874411 on Build GRCh38
1206782 c.1787C>T p.S596L Chr.6: 98874357-98874357 on Build GRCh38
451957 c.1775T>C p.V592A Chr.6: 98874369-98874369 on Build GRCh38
5026603 c.1782C>G p.F594L Chr.6: 98874362-98874362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532388048 Chr.6: 98873990-98873990 on Build GRCh38
rs944639669 Chr.6: 98874193-98874193 on Build GRCh38
rs1034559631 Chr.6: 98874319-98874319 on Build GRCh38
rs910199513 Chr.6: 98874142-98874142 on Build GRCh38
rs757279926 Chr.6: 98874348-98874348 on Build GRCh38
rs201989042 Chr.6: 98874354-98874354 on Build GRCh38
rs924006630 Chr.6: 98874223-98874223 on Build GRCh38
rs746650044 Chr.6: 98874365-98874365 on Build GRCh38
rs971653815 Chr.6: 98874016-98874016 on Build GRCh38
rs115816653 Chr.6: 98874393-98874393 on Build GRCh38
rs929163891 Chr.6: 98874175-98874175 on Build GRCh38
rs770326964 Chr.6: 98874380-98874380 on Build GRCh38
rs745713189 Chr.6: 98874356-98874356 on Build GRCh38
rs548253136 Chr.6: 98874095-98874095 on Build GRCh38
rs565821494 Chr.6: 98873996-98873996 on Build GRCh38
rs933243665 Chr.6: 98874112-98874112 on Build GRCh38
rs767709670 Chr.6: 98874285-98874285 on Build GRCh38
rs929088736 Chr.6: 98874192-98874192 on Build GRCh38
rs980125673 Chr.6: 98874033-98874033 on Build GRCh38
rs142988015 Chr.6: 98874273-98874273 on Build GRCh38
rs756228059 Chr.6: 98874415-98874415 on Build GRCh38
rs185850413 Chr.6: 98874269-98874269 on Build GRCh38
rs386704156 Chr.6: 98874273-98874275 on Build GRCh38
rs201574566 Chr.6: 98874206-98874206 on Build GRCh38
rs918645020 Chr.6: 98874150-98874150 on Build GRCh38
rs9388792 Chr.6: 98874260-98874260 on Build GRCh38
rs768243264 Chr.6: 98874366-98874366 on Build GRCh38
rs550510374 Chr.6: 98873993-98873993 on Build GRCh38
rs891542397 Chr.6: 98874254-98874254 on Build GRCh38
rs776141903 Chr.6: 98874371-98874371 on Build GRCh38
rs773599829 Chr.6: 98874267-98874267 on Build GRCh38
rs751656896 Chr.6: 98874308-98874308 on Build GRCh38
rs962676565 Chr.6: 98874120-98874120 on Build GRCh38
rs1034362494 Chr.6: 98874052-98874052 on Build GRCh38
rs762322035 Chr.6: 98874235-98874235 on Build GRCh38
rs115180578 Chr.6: 98874275-98874275 on Build GRCh38
rs757154231 Chr.6: 98874405-98874405 on Build GRCh38
rs536394975 Chr.6: 98874021-98874021 on Build GRCh38
rs751716397 Chr.6: 98874274-98874274 on Build GRCh38
rs1025018280 Chr.6: 98874004-98874004 on Build GRCh38
rs774709441 Chr.6: 98874399-98874399 on Build GRCh38
rs900180092 Chr.6: 98874214-98874214 on Build GRCh38
rs377712959 Chr.6: 98873956-98873956 on Build GRCh38
rs1032686842 Chr.6: 98873988-98873988 on Build GRCh38
rs148574319 Chr.6: 98874218-98874218 on Build GRCh38
rs747618415 Chr.6: 98874372-98874372 on Build GRCh38
rs371875653 Chr.6: 98874318-98874318 on Build GRCh38
rs9492614 Chr.6: 98873959-98873959 on Build GRCh38
rs986038735 Chr.6: 98874141-98874141 on Build GRCh38
rs962710974 Chr.6: 98874332-98874332 on Build GRCh38
rs974157707 Chr.6: 98874149-98874149 on Build GRCh38
rs911827902 Chr.6: 98874190-98874190 on Build GRCh38
rs111226417 Chr.6: 98874188-98874188 on Build GRCh38
rs1037340287 Chr.6: 98874177-98874177 on Build GRCh38
rs566557659 Chr.6: 98874117-98874117 on Build GRCh38
rs766646887 Chr.6: 98874396-98874396 on Build GRCh38
rs188288630 Chr.6: 98874006-98874006 on Build GRCh38
rs773572240 Chr.6: 98874384-98874384 on Build GRCh38
rs542778224 Chr.6: 98874357-98874357 on Build GRCh38

View Full Product Details