Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99277602-99278100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAXC
Entrez Gene ID
84553
Gene Name
failed axon connections homolog
Gene Aliases
C6orf168, dJ273F20
UniGene
Hs.573245
Gene Chromosome Location
Chr.6: 99272915-99350081 on Build GRCh38
Transcripts
NM_032511.2 XM_006715582.2 XM_011536188.2

SNP Detail

SNP ID Chromosome Location
rs894818181 Chr.6: 99277644-99277664 on Build GRCh38
rs901294290 Chr.6: 99277872-99277872 on Build GRCh38
rs535989975 Chr.6: 99277688-99277688 on Build GRCh38
rs141509092 Chr.6: 99277886-99277886 on Build GRCh38
rs573438107 Chr.6: 99277799-99277799 on Build GRCh38
rs548367652 Chr.6: 99278032-99278032 on Build GRCh38
rs959812194 Chr.6: 99278053-99278053 on Build GRCh38
rs937469757 Chr.6: 99277694-99277694 on Build GRCh38
rs977264421 Chr.6: 99278035-99278035 on Build GRCh38
rs1028769577 Chr.6: 99277900-99277900 on Build GRCh38
rs927428413 Chr.6: 99277675-99277675 on Build GRCh38
rs1056750368 Chr.6: 99277689-99277689 on Build GRCh38
rs761298625 Chr.6: 99277823-99277823 on Build GRCh38
rs540836576 Chr.6: 99277809-99277809 on Build GRCh38
rs758869040 Chr.6: 99278075-99278075 on Build GRCh38
rs1002559474 Chr.6: 99278048-99278048 on Build GRCh38
rs1038616718 Chr.6: 99277938-99277938 on Build GRCh38
rs990737860 Chr.6: 99277732-99277732 on Build GRCh38
rs562779879 Chr.6: 99277948-99277948 on Build GRCh38
rs671973 Chr.6: 99278045-99278045 on Build GRCh38
rs894129260 Chr.6: 99277757-99277757 on Build GRCh38
rs914628645 Chr.6: 99277756-99277756 on Build GRCh38
rs540793591 Chr.6: 99278018-99278018 on Build GRCh38
rs138513818 Chr.6: 99277860-99277860 on Build GRCh38
rs948359465 Chr.6: 99277795-99277795 on Build GRCh38
rs968601009 Chr.6: 99278028-99278028 on Build GRCh38
rs759854253 Chr.6: 99277929-99277929 on Build GRCh38
rs554669127 Chr.6: 99277782-99277782 on Build GRCh38
rs763011265 Chr.6: 99277623-99277623 on Build GRCh38
rs898204636 Chr.6: 99277939-99277939 on Build GRCh38
rs372364407 Chr.6: 99277672-99277672 on Build GRCh38
rs12195299 Chr.6: 99277821-99277821 on Build GRCh38

View Full Product Details