Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99380069-99380606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ3
Entrez Gene ID
51805
Gene Name
coenzyme Q3, methyltransferase
Gene Aliases
DHHBMT, DHHBMTASE, UG0215E05, bA9819.1
UniGene
Hs.713623
Gene Chromosome Location
Chr.6: 99369401-99394223 on Build GRCh38
Transcripts
NM_017421.3 XM_005267018.4 XM_006715500.3 XM_011535898.2 XM_011535899.1 XM_017010933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082788 c.370A>G p.R124G Chr.6: 99380205-99380205 on Build GRCh38
7362394 c.245C>T p.A82V Chr.6: 99380330-99380330 on Build GRCh38
6634140 c.236A>G p.Y79C Chr.6: 99380339-99380339 on Build GRCh38
4799855 c.263C>A p.S88Y Chr.6: 99380312-99380312 on Build GRCh38
389586 c.267A>T p.Q89H Chr.6: 99380308-99380308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548896136 Chr.6: 99380329-99380329 on Build GRCh38
rs986351965 Chr.6: 99380565-99380565 on Build GRCh38
rs750880593 Chr.6: 99380173-99380173 on Build GRCh38
rs781566655 Chr.6: 99380315-99380315 on Build GRCh38
rs559211257 Chr.6: 99380532-99380532 on Build GRCh38
rs140743578 Chr.6: 99380288-99380288 on Build GRCh38
rs894301577 Chr.6: 99380410-99380410 on Build GRCh38
rs779285077 Chr.6: 99380157-99380157 on Build GRCh38
rs111455620 Chr.6: 99380330-99380330 on Build GRCh38
rs1047220123 Chr.6: 99380184-99380184 on Build GRCh38
rs528276781 Chr.6: 99380391-99380391 on Build GRCh38
rs757841060 Chr.6: 99380155-99380155 on Build GRCh38
rs530259664 Chr.6: 99380294-99380294 on Build GRCh38
rs761237295 Chr.6: 99380279-99380279 on Build GRCh38
rs140709506 Chr.6: 99380531-99380531 on Build GRCh38
rs769845397 Chr.6: 99380229-99380229 on Build GRCh38
rs976225059 Chr.6: 99380113-99380113 on Build GRCh38
rs370276772 Chr.6: 99380581-99380581 on Build GRCh38
rs745768147 Chr.6: 99380236-99380236 on Build GRCh38
rs930279296 Chr.6: 99380495-99380495 on Build GRCh38
rs183814578 Chr.6: 99380180-99380180 on Build GRCh38
rs1003019860 Chr.6: 99380562-99380562 on Build GRCh38
rs770996339 Chr.6: 99380141-99380141 on Build GRCh38
rs1006208016 Chr.6: 99380462-99380462 on Build GRCh38
rs200544814 Chr.6: 99380222-99380222 on Build GRCh38
rs202070978 Chr.6: 99380217-99380217 on Build GRCh38
rs1038005094 Chr.6: 99380483-99380483 on Build GRCh38
rs201503654 Chr.6: 99380262-99380262 on Build GRCh38
rs113428143 Chr.6: 99380527-99380527 on Build GRCh38
rs562947006 Chr.6: 99380293-99380293 on Build GRCh38
rs775324416 Chr.6: 99380342-99380342 on Build GRCh38
rs753325156 Chr.6: 99380148-99380148 on Build GRCh38
rs9494272 Chr.6: 99380459-99380459 on Build GRCh38
rs923447747 Chr.6: 99380166-99380166 on Build GRCh38
rs768373988 Chr.6: 99380363-99380363 on Build GRCh38
rs771976517 Chr.6: 99380339-99380339 on Build GRCh38
rs755226826 Chr.6: 99380202-99380202 on Build GRCh38
rs777038091 Chr.6: 99380307-99380307 on Build GRCh38
rs547078973 Chr.6: 99380102-99380102 on Build GRCh38
rs953670806 Chr.6: 99380551-99380551 on Build GRCh38
rs986536432 Chr.6: 99380350-99380350 on Build GRCh38
rs575887811 Chr.6: 99380575-99380575 on Build GRCh38
rs1048781308 Chr.6: 99380420-99380420 on Build GRCh38
rs771884404 Chr.6: 99380252-99380252 on Build GRCh38
rs761480052 Chr.6: 99380369-99380369 on Build GRCh38
rs538807612 Chr.6: 99380435-99380435 on Build GRCh38
rs763723644 Chr.6: 99380258-99380258 on Build GRCh38
rs4612169 Chr.6: 99380387-99380387 on Build GRCh38
rs748343680 Chr.6: 99380319-99380319 on Build GRCh38
rs764750523 Chr.6: 99380290-99380290 on Build GRCh38
rs897884755 Chr.6: 99380498-99380498 on Build GRCh38
rs751957396 Chr.6: 99380299-99380299 on Build GRCh38
rs1056931443 Chr.6: 99380360-99380360 on Build GRCh38
rs780520402 Chr.6: 99380185-99380185 on Build GRCh38
rs150244593 Chr.6: 99380484-99380484 on Build GRCh38
rs377313779 Chr.6: 99380292-99380292 on Build GRCh38
rs948619841 Chr.6: 99380411-99380411 on Build GRCh38
rs756169853 Chr.6: 99380326-99380326 on Build GRCh38
rs776165532 Chr.6: 99380364-99380364 on Build GRCh38
rs267601176 Chr.6: 99380241-99380241 on Build GRCh38
rs775236131 Chr.6: 99380255-99380255 on Build GRCh38
rs371967196 Chr.6: 99380196-99380196 on Build GRCh38
rs746803032 Chr.6: 99380343-99380343 on Build GRCh38
rs755314655 Chr.6: 99380302-99380302 on Build GRCh38
rs188980651 Chr.6: 99380233-99380233 on Build GRCh38

View Full Product Details