Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99550856-99551355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNC
Entrez Gene ID
892
Gene Name
cyclin C
Gene Aliases
CycC
UniGene
Hs.430646
Gene Chromosome Location
Chr.6: 99542387-99568973 on Build GRCh38
Transcripts
NM_001013399.1 NM_005190.3 XM_006715594.1 XM_011536232.2 XM_017011436.1

Gene Symbol
TSTD3
Entrez Gene ID
100130890
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 3
Gene Aliases
-
UniGene
Hs.634506
Gene Chromosome Location
Chr.6: 99424854-99579288 on Build GRCh38
Transcripts
NM_001013399.1 NM_005190.3 XM_006715594.1 XM_011536232.2 XM_017011436.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
306932 c.438+1G>A p.? Chr.6: 99550992-99550992 on Build GRCh38
4945635 c.433C>G p.L145V Chr.6: 99550998-99550998 on Build GRCh38
3430977 c.420C>A p.F140L Chr.6: 99551011-99551011 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1020818835 Chr.6: 99551151-99551151 on Build GRCh38
rs1001922786 Chr.6: 99551122-99551122 on Build GRCh38
rs755976826 Chr.6: 99551040-99551041 on Build GRCh38
rs758953575 Chr.6: 99551079-99551079 on Build GRCh38
rs56223909 Chr.6: 99551149-99551149 on Build GRCh38
rs764375418 Chr.6: 99550973-99550973 on Build GRCh38
rs1005492627 Chr.6: 99551099-99551099 on Build GRCh38
rs1016919663 Chr.6: 99551116-99551116 on Build GRCh38
rs966918937 Chr.6: 99551164-99551164 on Build GRCh38
rs1056290604 Chr.6: 99550942-99550942 on Build GRCh38
rs562642599 Chr.6: 99551083-99551083 on Build GRCh38
rs772790820 Chr.6: 99550905-99550905 on Build GRCh38
rs779278405 Chr.6: 99551062-99551062 on Build GRCh38
rs2296155 Chr.6: 99551143-99551143 on Build GRCh38
rs976559337 Chr.6: 99551251-99551251 on Build GRCh38
rs767446951 Chr.6: 99550957-99550957 on Build GRCh38
rs114389754 Chr.6: 99550943-99550943 on Build GRCh38
rs754120370 Chr.6: 99551057-99551057 on Build GRCh38
rs897288370 Chr.6: 99551072-99551072 on Build GRCh38
rs968312202 Chr.6: 99551282-99551282 on Build GRCh38
rs578093763 Chr.6: 99551133-99551133 on Build GRCh38
rs756227048 Chr.6: 99550969-99550969 on Build GRCh38
rs922329241 Chr.6: 99551307-99551307 on Build GRCh38
rs757584147 Chr.6: 99551060-99551060 on Build GRCh38
rs1023945530 Chr.6: 99551009-99551009 on Build GRCh38
rs945149524 Chr.6: 99550940-99550940 on Build GRCh38
rs113112859 Chr.6: 99551118-99551118 on Build GRCh38
rs985658784 Chr.6: 99551175-99551175 on Build GRCh38
rs1037166812 Chr.6: 99551049-99551049 on Build GRCh38
rs189302354 Chr.6: 99550961-99550961 on Build GRCh38
rs961433840 Chr.6: 99551119-99551119 on Build GRCh38
rs911452882 Chr.6: 99551267-99551267 on Build GRCh38
rs184368587 Chr.6: 99550941-99550941 on Build GRCh38
rs746320904 Chr.6: 99551075-99551075 on Build GRCh38
rs910135708 Chr.6: 99551020-99551020 on Build GRCh38
rs201904580 Chr.6: 99551150-99551150 on Build GRCh38
rs748574723 Chr.6: 99551312-99551312 on Build GRCh38

View Full Product Details