Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 100517854-100518386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCC3
Entrez Gene ID
10973
Gene Name
activating signal cointegrator 1 complex subunit 3
Gene Aliases
ASC1p200, HELIC1, RNAH
UniGene
Hs.486031
Gene Chromosome Location
Chr.6: 100508194-100881372 on Build GRCh38
Transcripts
NM_006828.3 XM_011535394.2 XM_011535395.2 XM_011535396.2 XM_017010205.1 XM_017010206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7356576 c.5857G>A p.G1953S Chr.6: 100518061-100518061 on Build GRCh38
1161554 c.5842C>A p.Q1948K Chr.6: 100518076-100518076 on Build GRCh38
5910500 c.5884C>T p.L1962F Chr.6: 100518034-100518034 on Build GRCh38
7426049 c.5877T>G p.S1959S Chr.6: 100518041-100518041 on Build GRCh38
1697682 c.5860C>T p.R1954W Chr.6: 100518058-100518058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200821241 Chr.6: 100517986-100517986 on Build GRCh38
rs780283854 Chr.6: 100517895-100517895 on Build GRCh38
rs755402233 Chr.6: 100518036-100518036 on Build GRCh38
rs751806446 Chr.6: 100518138-100518138 on Build GRCh38
rs750640907 Chr.6: 100518015-100518015 on Build GRCh38
rs765358416 Chr.6: 100518011-100518011 on Build GRCh38
rs561142266 Chr.6: 100517967-100517967 on Build GRCh38
rs998048092 Chr.6: 100517883-100517883 on Build GRCh38
rs772029187 Chr.6: 100517941-100517941 on Build GRCh38
rs940178427 Chr.6: 100517881-100517881 on Build GRCh38
rs984686504 Chr.6: 100518030-100518030 on Build GRCh38
rs745565292 Chr.6: 100518192-100518192 on Build GRCh38
rs758660624 Chr.6: 100518027-100518027 on Build GRCh38
rs755387033 Chr.6: 100518145-100518145 on Build GRCh38
rs767520036 Chr.6: 100518003-100518003 on Build GRCh38
rs775957361 Chr.6: 100517952-100517952 on Build GRCh38
rs940188841 Chr.6: 100518175-100518175 on Build GRCh38
rs781726805 Chr.6: 100518038-100518038 on Build GRCh38
rs1037297395 Chr.6: 100517892-100517892 on Build GRCh38
rs550326463 Chr.6: 100518281-100518281 on Build GRCh38
rs777366197 Chr.6: 100518166-100518166 on Build GRCh38
rs759601879 Chr.6: 100518179-100518180 on Build GRCh38
rs760656462 Chr.6: 100518299-100518299 on Build GRCh38
rs984964709 Chr.6: 100518163-100518163 on Build GRCh38
rs771846135 Chr.6: 100518088-100518088 on Build GRCh38
rs773115452 Chr.6: 100517940-100517941 on Build GRCh38
rs769420460 Chr.6: 100518057-100518057 on Build GRCh38
rs937536294 Chr.6: 100518266-100518266 on Build GRCh38
rs747727761 Chr.6: 100517953-100517953 on Build GRCh38
rs753220760 Chr.6: 100518160-100518160 on Build GRCh38
rs571263714 Chr.6: 100518029-100518029 on Build GRCh38
rs747445208 Chr.6: 100517945-100517945 on Build GRCh38
rs780261391 Chr.6: 100518028-100518028 on Build GRCh38
rs759339081 Chr.6: 100517997-100517997 on Build GRCh38
rs535830855 Chr.6: 100518325-100518325 on Build GRCh38
rs757342455 Chr.6: 100518006-100518006 on Build GRCh38
rs927455007 Chr.6: 100518232-100518232 on Build GRCh38
rs376954806 Chr.6: 100518061-100518061 on Build GRCh38
rs3213542 Chr.6: 100518130-100518130 on Build GRCh38
rs530728412 Chr.6: 100517885-100517885 on Build GRCh38
rs999281194 Chr.6: 100518058-100518058 on Build GRCh38
rs549330173 Chr.6: 100517938-100517938 on Build GRCh38
rs766618626 Chr.6: 100518117-100518118 on Build GRCh38
rs532141712 Chr.6: 100518310-100518310 on Build GRCh38
rs745601584 Chr.6: 100517985-100517985 on Build GRCh38
rs1054564056 Chr.6: 100518287-100518287 on Build GRCh38
rs960611800 Chr.6: 100518091-100518091 on Build GRCh38
rs148599093 Chr.6: 100518298-100518298 on Build GRCh38
rs767319936 Chr.6: 100518229-100518229 on Build GRCh38
rs188706215 Chr.6: 100518355-100518355 on Build GRCh38
rs775484253 Chr.6: 100518089-100518089 on Build GRCh38
rs373611160 Chr.6: 100518168-100518168 on Build GRCh38
rs140237465 Chr.6: 100518047-100518047 on Build GRCh38
rs889910754 Chr.6: 100517976-100517976 on Build GRCh38
rs748923873 Chr.6: 100518080-100518080 on Build GRCh38
rs760673883 Chr.6: 100518105-100518105 on Build GRCh38
rs922887333 Chr.6: 100518333-100518333 on Build GRCh38
rs753791620 Chr.6: 100518005-100518005 on Build GRCh38
rs769508314 Chr.6: 100517955-100517955 on Build GRCh38
rs773015399 Chr.6: 100517966-100517966 on Build GRCh38
rs770727020 Chr.6: 100518084-100518084 on Build GRCh38
rs753866927 Chr.6: 100518162-100518163 on Build GRCh38
rs146657972 Chr.6: 100518018-100518018 on Build GRCh38
rs750365821 Chr.6: 100518285-100518285 on Build GRCh38
rs769255985 Chr.6: 100517948-100517948 on Build GRCh38
rs183405692 Chr.6: 100518336-100518336 on Build GRCh38
rs370541026 Chr.6: 100518147-100518147 on Build GRCh38
rs766657659 Chr.6: 100518131-100518131 on Build GRCh38
rs771895739 Chr.6: 100518193-100518193 on Build GRCh38
rs765270435 Chr.6: 100518106-100518106 on Build GRCh38
rs778639398 Chr.6: 100518190-100518190 on Build GRCh38
rs966113339 Chr.6: 100517882-100517882 on Build GRCh38
rs998188452 Chr.6: 100517957-100517957 on Build GRCh38
rs750553551 Chr.6: 100518110-100518110 on Build GRCh38
rs369548376 Chr.6: 100517971-100517971 on Build GRCh38
rs756730631 Chr.6: 100518162-100518162 on Build GRCh38

View Full Product Details