Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 105100622-105101133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BVES
Entrez Gene ID
11149
Gene Name
blood vessel epicardial substance
Gene Aliases
HBVES, LGMD2X, POP1, POPDC1
UniGene
Hs.221660
Gene Chromosome Location
Chr.6: 105096822-105137174 on Build GRCh38
Transcripts
NM_001199563.1 NM_007073.4 NM_147147.3 XM_011535398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817110 c.1047A>G p.A349A Chr.6: 105101125-105101125 on Build GRCh38
3410516 c.1043C>T p.P348L Chr.6: 105101129-105101129 on Build GRCh38
324841 c.1058C>T p.T353I Chr.6: 105101114-105101114 on Build GRCh38
4666436 c.1044G>A p.P348P Chr.6: 105101128-105101128 on Build GRCh38
7171260 c.1059A>G p.T353T Chr.6: 105101113-105101113 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs943138377 Chr.6: 105100778-105100778 on Build GRCh38
rs894494278 Chr.6: 105100875-105100875 on Build GRCh38
rs886731106 Chr.6: 105100822-105100822 on Build GRCh38
rs544697809 Chr.6: 105100829-105100829 on Build GRCh38
rs981940596 Chr.6: 105100933-105100933 on Build GRCh38
rs1005210736 Chr.6: 105100894-105100906 on Build GRCh38
rs1023450547 Chr.6: 105100655-105100655 on Build GRCh38
rs759766919 Chr.6: 105101055-105101055 on Build GRCh38
rs554721697 Chr.6: 105100834-105100835 on Build GRCh38
rs112964153 Chr.6: 105101060-105101060 on Build GRCh38
rs202148646 Chr.6: 105101061-105101061 on Build GRCh38
rs931697330 Chr.6: 105100765-105100765 on Build GRCh38
rs140313590 Chr.6: 105100989-105100989 on Build GRCh38
rs767606599 Chr.6: 105101058-105101058 on Build GRCh38
rs933535838 Chr.6: 105100800-105100800 on Build GRCh38
rs375241884 Chr.6: 105101041-105101041 on Build GRCh38
rs766510648 Chr.6: 105101040-105101040 on Build GRCh38
rs540292748 Chr.6: 105100868-105100868 on Build GRCh38
rs981971758 Chr.6: 105100684-105100684 on Build GRCh38
rs183390165 Chr.6: 105101104-105101104 on Build GRCh38
rs971506085 Chr.6: 105100904-105100904 on Build GRCh38
rs1039156863 Chr.6: 105100780-105100780 on Build GRCh38
rs914258986 Chr.6: 105101072-105101072 on Build GRCh38
rs145488013 Chr.6: 105100970-105100970 on Build GRCh38
rs907786580 Chr.6: 105100686-105100686 on Build GRCh38
rs1012338617 Chr.6: 105100853-105100853 on Build GRCh38
rs538785772 Chr.6: 105100715-105100715 on Build GRCh38
rs1024382740 Chr.6: 105100895-105100895 on Build GRCh38
rs183602594 Chr.6: 105100784-105100784 on Build GRCh38
rs944461281 Chr.6: 105101112-105101112 on Build GRCh38
rs188442096 Chr.6: 105100882-105100882 on Build GRCh38
rs1014769294 Chr.6: 105100951-105100951 on Build GRCh38
rs778111050 Chr.6: 105101066-105101066 on Build GRCh38
rs114541983 Chr.6: 105100649-105100649 on Build GRCh38
rs149722503 Chr.6: 105100678-105100678 on Build GRCh38
rs770465732 Chr.6: 105100653-105100653 on Build GRCh38
rs575558953 Chr.6: 105100823-105100823 on Build GRCh38
rs890230177 Chr.6: 105100809-105100809 on Build GRCh38
rs920284679 Chr.6: 105100750-105100750 on Build GRCh38
rs145232199 Chr.6: 105101033-105101033 on Build GRCh38
rs1041906145 Chr.6: 105100821-105100821 on Build GRCh38
rs974473204 Chr.6: 105101084-105101084 on Build GRCh38
rs376764408 Chr.6: 105101073-105101073 on Build GRCh38
rs920529160 Chr.6: 105101029-105101029 on Build GRCh38
rs944866959 Chr.6: 105100819-105100819 on Build GRCh38
rs17065481 Chr.6: 105100845-105100845 on Build GRCh38
rs867858037 Chr.6: 105101005-105101005 on Build GRCh38
rs775907092 Chr.6: 105101045-105101045 on Build GRCh38
rs973440681 Chr.6: 105100719-105100719 on Build GRCh38
rs989796624 Chr.6: 105100720-105100720 on Build GRCh38
rs221657 Chr.6: 105100975-105100975 on Build GRCh38
rs1013357484 Chr.6: 105100886-105100886 on Build GRCh38
rs971245165 Chr.6: 105100920-105100920 on Build GRCh38

View Full Product Details