Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 115942830-115943344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRK
Entrez Gene ID
2444
Gene Name
fyn related Src family tyrosine kinase
Gene Aliases
GTK, PTK5, RAK
UniGene
Hs.89426
Gene Chromosome Location
Chr.6: 115941031-116100739 on Build GRCh38
Transcripts
NM_002031.2 XM_005266880.4 XM_005266881.2 XM_005266882.4 XM_011535653.2 XM_011535654.2 XM_011535655.2 XM_011535656.2 XM_017010645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1725280 c.1231A>T p.S411C Chr.6: 115943095-115943095 on Build GRCh38
3619641 c.1242C>T p.S414S Chr.6: 115943084-115943084 on Build GRCh38
5404185 c.1173C>T p.H391H Chr.6: 115943153-115943153 on Build GRCh38
3619640 c.1305T>G p.S435R Chr.6: 115943021-115943021 on Build GRCh38
6307318 c.1187C>T p.P396L Chr.6: 115943139-115943139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055292712 Chr.6: 115942887-115942887 on Build GRCh38
rs980042768 Chr.6: 115943196-115943196 on Build GRCh38
rs74407477 Chr.6: 115943123-115943123 on Build GRCh38
rs773462469 Chr.6: 115943089-115943089 on Build GRCh38
rs765434724 Chr.6: 115942980-115942980 on Build GRCh38
rs372244528 Chr.6: 115942994-115942994 on Build GRCh38
rs922146806 Chr.6: 115943317-115943317 on Build GRCh38
rs758491803 Chr.6: 115942991-115942991 on Build GRCh38
rs751580135 Chr.6: 115943108-115943108 on Build GRCh38
rs989329507 Chr.6: 115943236-115943236 on Build GRCh38
rs775379239 Chr.6: 115942976-115942976 on Build GRCh38
rs754168620 Chr.6: 115942926-115942926 on Build GRCh38
rs772165246 Chr.6: 115942852-115942852 on Build GRCh38
rs750678775 Chr.6: 115942982-115942982 on Build GRCh38
rs774653455 Chr.6: 115942947-115942947 on Build GRCh38
rs919917624 Chr.6: 115942931-115942931 on Build GRCh38
rs146969929 Chr.6: 115943107-115943107 on Build GRCh38
rs185354663 Chr.6: 115943152-115943152 on Build GRCh38
rs376090182 Chr.6: 115943124-115943124 on Build GRCh38
rs938778402 Chr.6: 115942956-115942956 on Build GRCh38
rs748474763 Chr.6: 115943039-115943039 on Build GRCh38
rs755426355 Chr.6: 115942995-115942995 on Build GRCh38
rs377733382 Chr.6: 115943109-115943109 on Build GRCh38
rs770537162 Chr.6: 115943063-115943063 on Build GRCh38
rs550480 Chr.6: 115943191-115943191 on Build GRCh38
rs768646391 Chr.6: 115943237-115943237 on Build GRCh38
rs370495146 Chr.6: 115943075-115943075 on Build GRCh38
rs149341971 Chr.6: 115943148-115943148 on Build GRCh38
rs779562799 Chr.6: 115943057-115943059 on Build GRCh38
rs1043364003 Chr.6: 115943114-115943114 on Build GRCh38
rs773527826 Chr.6: 115942873-115942873 on Build GRCh38
rs113246501 Chr.6: 115943090-115943090 on Build GRCh38
rs57164993 Chr.6: 115943236-115943237 on Build GRCh38
rs913414438 Chr.6: 115943037-115943037 on Build GRCh38
rs781672641 Chr.6: 115943014-115943014 on Build GRCh38
rs758764121 Chr.6: 115943223-115943224 on Build GRCh38
rs11758059 Chr.6: 115943000-115943000 on Build GRCh38
rs763121310 Chr.6: 115943098-115943098 on Build GRCh38
rs766611219 Chr.6: 115943101-115943101 on Build GRCh38
rs200561287 Chr.6: 115943111-115943111 on Build GRCh38
rs143648469 Chr.6: 115943153-115943153 on Build GRCh38
rs756500618 Chr.6: 115943042-115943042 on Build GRCh38
rs760517045 Chr.6: 115943084-115943084 on Build GRCh38
rs1053267410 Chr.6: 115943003-115943003 on Build GRCh38
rs767597023 Chr.6: 115942974-115942974 on Build GRCh38
rs755604497 Chr.6: 115943047-115943047 on Build GRCh38
rs928644147 Chr.6: 115943200-115943200 on Build GRCh38
rs1038404150 Chr.6: 115943267-115943267 on Build GRCh38
rs11330350 Chr.6: 115943224-115943224 on Build GRCh38
rs180731108 Chr.6: 115943013-115943013 on Build GRCh38
rs150259692 Chr.6: 115943137-115943137 on Build GRCh38
rs17077409 Chr.6: 115942902-115942902 on Build GRCh38
rs766484707 Chr.6: 115942993-115942993 on Build GRCh38
rs772080212 Chr.6: 115943080-115943080 on Build GRCh38
rs201753000 Chr.6: 115943138-115943138 on Build GRCh38
rs756925110 Chr.6: 115943126-115943126 on Build GRCh38
rs745563720 Chr.6: 115943224-115943226 on Build GRCh38
rs760946377 Chr.6: 115942901-115942901 on Build GRCh38
rs374511452 Chr.6: 115943119-115943119 on Build GRCh38
rs1004687705 Chr.6: 115942899-115942899 on Build GRCh38
rs147717219 Chr.6: 115943120-115943120 on Build GRCh38
rs80305773 Chr.6: 115943296-115943296 on Build GRCh38
rs973319938 Chr.6: 115942918-115942918 on Build GRCh38
rs139523641 Chr.6: 115943262-115943262 on Build GRCh38
rs970069561 Chr.6: 115942946-115942946 on Build GRCh38
rs777947516 Chr.6: 115943224-115943225 on Build GRCh38
rs748939342 Chr.6: 115943057-115943057 on Build GRCh38
rs753213606 Chr.6: 115943217-115943217 on Build GRCh38
rs550507766 Chr.6: 115943212-115943212 on Build GRCh38
rs542576719 Chr.6: 115942951-115942951 on Build GRCh38
rs969283455 Chr.6: 115943194-115943194 on Build GRCh38
rs771073208 Chr.6: 115943168-115943168 on Build GRCh38
rs763853996 Chr.6: 115943085-115943085 on Build GRCh38
rs141208044 Chr.6: 115943073-115943073 on Build GRCh38
rs138957087 Chr.6: 115943139-115943139 on Build GRCh38
rs913393495 Chr.6: 115943261-115943261 on Build GRCh38
rs947651624 Chr.6: 115943106-115943106 on Build GRCh38
rs201918998 Chr.6: 115943224-115943224 on Build GRCh38
rs939641423 Chr.6: 115942874-115942874 on Build GRCh38
rs777464965 Chr.6: 115943046-115943046 on Build GRCh38
rs201637193 Chr.6: 115943211-115943211 on Build GRCh38
rs773962419 Chr.6: 115943072-115943072 on Build GRCh38

View Full Product Details