Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116003798-116004292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRK
Entrez Gene ID
2444
Gene Name
fyn related Src family tyrosine kinase
Gene Aliases
GTK, PTK5, RAK
UniGene
Hs.89426
Gene Chromosome Location
Chr.6: 115941031-116100739 on Build GRCh38
Transcripts
NM_002031.2 XM_005266880.4 XM_005266881.2 XM_005266882.4 XM_011535653.2 XM_011535654.2 XM_011535655.2 XM_011535656.2 XM_017010645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586162 c.411C>T p.T137T Chr.6: 116003932-116003932 on Build GRCh38
74703 c.361A>G p.I121V Chr.6: 116003982-116003982 on Build GRCh38
3156979 c.436G>T p.E146* Chr.6: 116003907-116003907 on Build GRCh38
1072348 c.412G>C p.G138R Chr.6: 116003931-116003931 on Build GRCh38
3156978 c.439A>C p.S147R Chr.6: 116003904-116003904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748623211 Chr.6: 116004011-116004011 on Build GRCh38
rs1006189651 Chr.6: 116004243-116004243 on Build GRCh38
rs958176769 Chr.6: 116004190-116004191 on Build GRCh38
rs1046988659 Chr.6: 116004153-116004153 on Build GRCh38
rs142770112 Chr.6: 116003987-116003987 on Build GRCh38
rs187351459 Chr.6: 116004092-116004092 on Build GRCh38
rs778978275 Chr.6: 116003860-116003860 on Build GRCh38
rs576981129 Chr.6: 116004128-116004128 on Build GRCh38
rs755959595 Chr.6: 116003852-116003852 on Build GRCh38
rs757813892 Chr.6: 116003826-116003826 on Build GRCh38
rs867690373 Chr.6: 116004261-116004261 on Build GRCh38
rs751000439 Chr.6: 116003946-116003946 on Build GRCh38
rs770221371 Chr.6: 116004013-116004013 on Build GRCh38
rs34064900 Chr.6: 116003945-116003945 on Build GRCh38
rs554429518 Chr.6: 116004057-116004057 on Build GRCh38
rs762257926 Chr.6: 116003927-116003927 on Build GRCh38
rs765870792 Chr.6: 116003831-116003831 on Build GRCh38
rs768917709 Chr.6: 116004009-116004009 on Build GRCh38
rs1016330218 Chr.6: 116003916-116003916 on Build GRCh38
rs781112476 Chr.6: 116003845-116003845 on Build GRCh38
rs375482461 Chr.6: 116004021-116004021 on Build GRCh38
rs990150126 Chr.6: 116004191-116004191 on Build GRCh38
rs752650800 Chr.6: 116003927-116003928 on Build GRCh38
rs3756772 Chr.6: 116003979-116003979 on Build GRCh38
rs760613388 Chr.6: 116004042-116004042 on Build GRCh38
rs150495043 Chr.6: 116003971-116003971 on Build GRCh38
rs763527245 Chr.6: 116003936-116003936 on Build GRCh38
rs1032291188 Chr.6: 116004186-116004186 on Build GRCh38
rs746144192 Chr.6: 116003861-116003861 on Build GRCh38
rs369216672 Chr.6: 116003998-116003998 on Build GRCh38
rs759112703 Chr.6: 116003958-116003958 on Build GRCh38
rs935989195 Chr.6: 116003844-116003844 on Build GRCh38
rs773564517 Chr.6: 116004020-116004020 on Build GRCh38
rs569656818 Chr.6: 116004078-116004078 on Build GRCh38
rs753599705 Chr.6: 116003982-116003982 on Build GRCh38
rs781759597 Chr.6: 116004010-116004010 on Build GRCh38
rs746935585 Chr.6: 116004076-116004076 on Build GRCh38
rs777585898 Chr.6: 116003980-116003980 on Build GRCh38
rs190939776 Chr.6: 116004127-116004127 on Build GRCh38
rs368746017 Chr.6: 116003841-116003841 on Build GRCh38
rs774047136 Chr.6: 116004034-116004034 on Build GRCh38
rs888349970 Chr.6: 116004219-116004219 on Build GRCh38
rs752484244 Chr.6: 116003849-116003849 on Build GRCh38
rs759755643 Chr.6: 116004181-116004181 on Build GRCh38
rs772704030 Chr.6: 116003877-116003877 on Build GRCh38
rs375756617 Chr.6: 116003932-116003932 on Build GRCh38
rs886479621 Chr.6: 116003829-116003829 on Build GRCh38
rs953217235 Chr.6: 116004117-116004117 on Build GRCh38
rs867784375 Chr.6: 116003972-116003972 on Build GRCh38
rs12173584 Chr.6: 116004191-116004191 on Build GRCh38
rs762614101 Chr.6: 116003943-116003943 on Build GRCh38
rs752237084 Chr.6: 116004040-116004040 on Build GRCh38
rs772793869 Chr.6: 116003978-116003978 on Build GRCh38
rs770700465 Chr.6: 116004039-116004039 on Build GRCh38
rs545141267 Chr.6: 116004209-116004209 on Build GRCh38
rs1037641702 Chr.6: 116004248-116004248 on Build GRCh38
rs748652398 Chr.6: 116003870-116003870 on Build GRCh38
rs1030175552 Chr.6: 116004109-116004109 on Build GRCh38
rs117605537 Chr.6: 116003968-116003968 on Build GRCh38
rs765412402 Chr.6: 116004252-116004252 on Build GRCh38
rs1040889060 Chr.6: 116004051-116004051 on Build GRCh38
rs772216083 Chr.6: 116003866-116003866 on Build GRCh38
rs751089081 Chr.6: 116003840-116003840 on Build GRCh38
rs151309696 Chr.6: 116003887-116003887 on Build GRCh38
rs769224017 Chr.6: 116003885-116003885 on Build GRCh38
rs770246096 Chr.6: 116003931-116003931 on Build GRCh38
rs770612128 Chr.6: 116004026-116004026 on Build GRCh38
rs181842829 Chr.6: 116003871-116003871 on Build GRCh38

View Full Product Details