Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116125059-116125588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL10A1
Entrez Gene ID
1300
Gene Name
collagen type X alpha 1 chain
Gene Aliases
-
UniGene
Hs.520339
Gene Chromosome Location
Chr.6: 116118909-116218698 on Build GRCh38
Transcripts
NM_000493.3 XM_006715333.3 XM_011535432.2 XM_011535433.2 XM_017010248.1 XM_017010249.1 NM_152729.2 XM_006715378.3

Gene Symbol
NT5DC1
Entrez Gene ID
221294
Gene Name
5'-nucleotidase domain containing 1
Gene Aliases
C6orf200, LP2642, NT5C2L1
UniGene
Hs.520341
Gene Chromosome Location
Chr.6: 116100833-116245690 on Build GRCh38
Transcripts
NM_000493.3 XM_006715333.3 XM_011535432.2 XM_011535433.2 XM_017010248.1 XM_017010249.1 NM_152729.2 XM_006715378.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6632284 c.54A>C p.G18G Chr.6: 116125439-116125439 on Build GRCh38
3857714 c.130A>G p.I44V Chr.6: 116125363-116125363 on Build GRCh38
1072362 c.4C>A p.L2M Chr.6: 116125489-116125489 on Build GRCh38
5991307 c.113C>A p.T38N Chr.6: 116125380-116125380 on Build GRCh38
3619653 c.71G>A p.R24Q Chr.6: 116125422-116125422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751978834 Chr.6: 116125348-116125348 on Build GRCh38
rs767889462 Chr.6: 116125429-116125429 on Build GRCh38
rs1039586029 Chr.6: 116125274-116125274 on Build GRCh38
rs755737324 Chr.6: 116125432-116125432 on Build GRCh38
rs371468765 Chr.6: 116125199-116125199 on Build GRCh38
rs901397217 Chr.6: 116125362-116125362 on Build GRCh38
rs749633212 Chr.6: 116125307-116125307 on Build GRCh38
rs556358438 Chr.6: 116125167-116125167 on Build GRCh38
rs752328564 Chr.6: 116125430-116125430 on Build GRCh38
rs534414782 Chr.6: 116125105-116125105 on Build GRCh38
rs745701381 Chr.6: 116125387-116125387 on Build GRCh38
rs2024995 Chr.6: 116125235-116125235 on Build GRCh38
rs777154342 Chr.6: 116125378-116125378 on Build GRCh38
rs745826013 Chr.6: 116125474-116125474 on Build GRCh38
rs780861272 Chr.6: 116125358-116125358 on Build GRCh38
rs927118438 Chr.6: 116125165-116125165 on Build GRCh38
rs1042750688 Chr.6: 116125287-116125287 on Build GRCh38
rs377425972 Chr.6: 116125517-116125517 on Build GRCh38
rs574499406 Chr.6: 116125204-116125204 on Build GRCh38
rs769964985 Chr.6: 116125501-116125501 on Build GRCh38
rs983995260 Chr.6: 116125153-116125153 on Build GRCh38
rs754619889 Chr.6: 116125352-116125352 on Build GRCh38
rs774469761 Chr.6: 116125480-116125481 on Build GRCh38
rs770035715 Chr.6: 116125121-116125121 on Build GRCh38
rs377375663 Chr.6: 116125315-116125315 on Build GRCh38
rs142411445 Chr.6: 116125343-116125343 on Build GRCh38
rs759940492 Chr.6: 116125428-116125428 on Build GRCh38
rs972369568 Chr.6: 116125098-116125098 on Build GRCh38
rs199808053 Chr.6: 116125423-116125423 on Build GRCh38
rs374560255 Chr.6: 116125482-116125482 on Build GRCh38
rs942448687 Chr.6: 116125495-116125495 on Build GRCh38
rs918355747 Chr.6: 116125107-116125107 on Build GRCh38
rs755612188 Chr.6: 116125372-116125372 on Build GRCh38
rs111033550 Chr.6: 116125441-116125441 on Build GRCh38
rs768980930 Chr.6: 116125390-116125390 on Build GRCh38
rs757234908 Chr.6: 116125463-116125463 on Build GRCh38
rs949403944 Chr.6: 116125247-116125247 on Build GRCh38
rs748339808 Chr.6: 116125491-116125491 on Build GRCh38
rs139701581 Chr.6: 116125532-116125532 on Build GRCh38
rs748219587 Chr.6: 116125541-116125542 on Build GRCh38
rs761969132 Chr.6: 116125396-116125396 on Build GRCh38
rs890883311 Chr.6: 116125205-116125205 on Build GRCh38
rs755842616 Chr.6: 116125477-116125478 on Build GRCh38
rs774301704 Chr.6: 116125301-116125301 on Build GRCh38
rs752123744 Chr.6: 116125366-116125366 on Build GRCh38
rs1064583 Chr.6: 116125413-116125413 on Build GRCh38
rs147612968 Chr.6: 116125450-116125450 on Build GRCh38
rs766668557 Chr.6: 116125345-116125345 on Build GRCh38
rs373709444 Chr.6: 116125219-116125219 on Build GRCh38
rs758830415 Chr.6: 116125344-116125344 on Build GRCh38
rs111033551 Chr.6: 116125440-116125440 on Build GRCh38
rs777324834 Chr.6: 116125433-116125433 on Build GRCh38
rs34539681 Chr.6: 116125470-116125470 on Build GRCh38
rs990852794 Chr.6: 116125355-116125355 on Build GRCh38
rs374225504 Chr.6: 116125296-116125296 on Build GRCh38
rs769843417 Chr.6: 116125412-116125412 on Build GRCh38
rs779858250 Chr.6: 116125479-116125480 on Build GRCh38
rs1053994343 Chr.6: 116125312-116125312 on Build GRCh38
rs191911355 Chr.6: 116125333-116125333 on Build GRCh38
rs776946569 Chr.6: 116125393-116125393 on Build GRCh38
rs147235308 Chr.6: 116125527-116125527 on Build GRCh38
rs749192483 Chr.6: 116125384-116125384 on Build GRCh38
rs774722396 Chr.6: 116125323-116125323 on Build GRCh38
rs552989772 Chr.6: 116125143-116125143 on Build GRCh38
rs745699172 Chr.6: 116125402-116125403 on Build GRCh38
rs992714969 Chr.6: 116125203-116125203 on Build GRCh38
rs199644615 Chr.6: 116125328-116125328 on Build GRCh38
rs1028048149 Chr.6: 116125159-116125159 on Build GRCh38
rs779715618 Chr.6: 116125486-116125486 on Build GRCh38
rs766938257 Chr.6: 116125422-116125422 on Build GRCh38
rs773337533 Chr.6: 116125502-116125502 on Build GRCh38
rs778850616 Chr.6: 116125386-116125386 on Build GRCh38
rs947248245 Chr.6: 116125559-116125562 on Build GRCh38
rs556661451 Chr.6: 116125269-116125269 on Build GRCh38
rs770693989 Chr.6: 116125297-116125297 on Build GRCh38
rs763500298 Chr.6: 116125420-116125420 on Build GRCh38
rs760042533 Chr.6: 116125539-116125539 on Build GRCh38
rs781272934 Chr.6: 116125399-116125400 on Build GRCh38
rs764481154 Chr.6: 116125317-116125317 on Build GRCh38
rs187987567 Chr.6: 116125326-116125326 on Build GRCh38

View Full Product Details