Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116462186-116462699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM26F
Entrez Gene ID
441168
Gene Name
family with sequence similarity 26 member F
Gene Aliases
C6orf187, dJ93H18.5
UniGene
Hs.381220
Gene Chromosome Location
Chr.6: 116461370-116463782 on Build GRCh38
Transcripts
NM_001010919.2 NM_001276460.1 XM_005266999.3 XM_011535845.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6836901 c.274G>C p.A92P Chr.6: 116462203-116462203 on Build GRCh38
3393776 c.424C>T p.R142C Chr.6: 116462353-116462353 on Build GRCh38
5450173 c.421G>A p.G141S Chr.6: 116462350-116462350 on Build GRCh38
5813331 c.417C>A p.C139* Chr.6: 116462346-116462346 on Build GRCh38
6049987 c.345C>G p.A115A Chr.6: 116462274-116462274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945045087 Chr.6: 116462603-116462603 on Build GRCh38
rs952320989 Chr.6: 116462295-116462295 on Build GRCh38
rs922586923 Chr.6: 116462623-116462623 on Build GRCh38
rs1027910899 Chr.6: 116462500-116462500 on Build GRCh38
rs553301411 Chr.6: 116462491-116462491 on Build GRCh38
rs61740843 Chr.6: 116462397-116462397 on Build GRCh38
rs992299039 Chr.6: 116462362-116462362 on Build GRCh38
rs945033930 Chr.6: 116462381-116462381 on Build GRCh38
rs1017398946 Chr.6: 116462575-116462575 on Build GRCh38
rs945318699 Chr.6: 116462211-116462211 on Build GRCh38
rs11544159 Chr.6: 116462259-116462259 on Build GRCh38
rs1024248444 Chr.6: 116462442-116462442 on Build GRCh38
rs979041204 Chr.6: 116462613-116462613 on Build GRCh38
rs746434125 Chr.6: 116462458-116462458 on Build GRCh38
rs961413323 Chr.6: 116462594-116462594 on Build GRCh38
rs11544161 Chr.6: 116462296-116462296 on Build GRCh38
rs891470152 Chr.6: 116462434-116462434 on Build GRCh38
rs570090638 Chr.6: 116462388-116462388 on Build GRCh38
rs1038183722 Chr.6: 116462319-116462319 on Build GRCh38
rs932498644 Chr.6: 116462412-116462412 on Build GRCh38
rs971234040 Chr.6: 116462265-116462265 on Build GRCh38
rs772718572 Chr.6: 116462488-116462488 on Build GRCh38
rs753344499 Chr.6: 116462212-116462212 on Build GRCh38
rs755445117 Chr.6: 116462390-116462390 on Build GRCh38
rs981416932 Chr.6: 116462554-116462554 on Build GRCh38
rs553402397 Chr.6: 116462402-116462402 on Build GRCh38
rs991902366 Chr.6: 116462576-116462576 on Build GRCh38
rs1049908436 Chr.6: 116462391-116462391 on Build GRCh38
rs779771068 Chr.6: 116462358-116462358 on Build GRCh38
rs909923041 Chr.6: 116462341-116462341 on Build GRCh38
rs766910203 Chr.6: 116462354-116462354 on Build GRCh38
rs897863429 Chr.6: 116462460-116462460 on Build GRCh38
rs743853 Chr.6: 116462596-116462596 on Build GRCh38
rs866767887 Chr.6: 116462443-116462443 on Build GRCh38
rs939975739 Chr.6: 116462350-116462350 on Build GRCh38
rs10784 Chr.6: 116462227-116462227 on Build GRCh38
rs927096558 Chr.6: 116462276-116462276 on Build GRCh38
rs1018987349 Chr.6: 116462273-116462273 on Build GRCh38
rs910939001 Chr.6: 116462202-116462202 on Build GRCh38
rs934178977 Chr.6: 116462386-116462386 on Build GRCh38
rs760961107 Chr.6: 116462302-116462302 on Build GRCh38
rs1033875095 Chr.6: 116462561-116462561 on Build GRCh38
rs952301336 Chr.6: 116462514-116462514 on Build GRCh38
rs758179055 Chr.6: 116462349-116462349 on Build GRCh38
rs999988021 Chr.6: 116462411-116462411 on Build GRCh38
rs4354189 Chr.6: 116462274-116462274 on Build GRCh38
rs1001442558 Chr.6: 116462463-116462463 on Build GRCh38
rs760974954 Chr.6: 116462511-116462511 on Build GRCh38
rs117361304 Chr.6: 116462456-116462456 on Build GRCh38
rs1010158433 Chr.6: 116462231-116462231 on Build GRCh38
rs751061740 Chr.6: 116462424-116462424 on Build GRCh38
rs903728897 Chr.6: 116462219-116462219 on Build GRCh38
rs889508770 Chr.6: 116462210-116462210 on Build GRCh38
rs951967505 Chr.6: 116462509-116462509 on Build GRCh38
rs745577148 Chr.6: 116462656-116462656 on Build GRCh38
rs574943823 Chr.6: 116462583-116462583 on Build GRCh38
rs934273728 Chr.6: 116462652-116462652 on Build GRCh38
rs750716446 Chr.6: 116462287-116462287 on Build GRCh38
rs556617380 Chr.6: 116462671-116462671 on Build GRCh38
rs913618524 Chr.6: 116462589-116462589 on Build GRCh38
rs534984948 Chr.6: 116462359-116462359 on Build GRCh38
rs913608647 Chr.6: 116462363-116462363 on Build GRCh38
rs926378531 Chr.6: 116462339-116462339 on Build GRCh38
rs61740836 Chr.6: 116462435-116462435 on Build GRCh38
rs1027902364 Chr.6: 116462292-116462292 on Build GRCh38
rs1029491856 Chr.6: 116462235-116462235 on Build GRCh38
rs76716752 Chr.6: 116462422-116462422 on Build GRCh38
rs973119944 Chr.6: 116462611-116462611 on Build GRCh38
rs1032713895 Chr.6: 116462220-116462220 on Build GRCh38
rs188970565 Chr.6: 116462566-116462566 on Build GRCh38
rs535196241 Chr.6: 116462660-116462660 on Build GRCh38
rs1009328907 Chr.6: 116462266-116462266 on Build GRCh38

View Full Product Details